Gene Transcripts

Gene Symbol
HOXD4
Entrez Gene ID
3233
Gene Name
homeobox D4
Gene Aliases
HHO.C13, HOX-5.1, HOX4, HOX4B, Hox-4.2
Chromosome Location
Chr. 2: 176150394 - 176156078 on Build GRCh38
UniGene ID
Hs.591609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014621.2 NP_055436.2 1, 2 670
GenBank mRNA AK313885.1 - 1, 2 590
BC016763.1 - 1, 2 488 4068219
BC074797.2 - 1, 2 468 30915295

View Full Product Details