Gene Transcripts
- Gene Symbol
- PAPOLG
- Entrez Gene ID
- 64895
- Gene Name
- poly(A) polymerase gamma
- Gene Aliases
- Chromosome Location
- Chr. 2: 60756230 - 60802086 on Build GRCh38
- UniGene ID
- Hs.387471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022894.3 |
NP_075045.2
|
17, 18 |
1939 |
|
XM_005264500.3 |
XP_005264557.1
|
17, 18 |
1939 |
|
XM_005264501.1 |
XP_005264558.1
|
17, 18 |
1757 |
|
XM_011533041.1 |
XP_011531343.1
|
16, 17 |
1645 |
|
XM_017004723.1 |
XP_016860212.1
|
16, 17 |
1801 |
|
XM_017004724.1 |
XP_016860213.1
|
16, 17 |
1801 |
|
GenBank mRNA |
AB209304.1 |
- |
7, 8 |
694 |
|
AF312211.1 |
- |
17, 18 |
1921 |
|
AJ308101.1 |
- |
17, 18 |
1690 |
|
AK023034.1 |
- |
6, 7 |
641 |
|
AK023544.1 |
- |
16, 17 |
1688 |
|
AK024249.1 |
- |
5, 6 |
556 |
|
AK314663.1 |
- |
17, 18 |
1903 |
|
AY029162.1 |
- |
17, 18 |
1699 |
|
BC111701.1 |
- |
17, 18 |
1813 |
40038554 |
BC113747.1 |
- |
17, 18 |
1813 |
40038563 |
BX647824.1 |
- |
16, 17 |
1659 |
|
View Full Product Details