Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41330785-41331321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB1
Entrez Gene ID
7040
Gene Name
transforming growth factor beta 1
Gene Aliases
CED, DPD1, LAP, TGFB, TGFbeta
UniGene
Hs.645227
Gene Chromosome Location
Chr.19: 41330531-41353933 on Build GRCh38
Transcripts
NM_000660.5 XM_011527242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4733343 c.1155C>T p.R385R Chr.19: 41331070-41331070 on Build GRCh38
6375057 c.1019T>G p.L340R Chr.19: 41331206-41331206 on Build GRCh38
6374629 c.1085A>G p.E362G Chr.19: 41331140-41331140 on Build GRCh38
7393659 c.1111G>T p.G371C Chr.19: 41331114-41331114 on Build GRCh38
6191004 c.1058C>T p.A353V Chr.19: 41331167-41331167 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933612071 Chr.19: 41331281-41331281 on Build GRCh38
rs747563417 Chr.19: 41331075-41331075 on Build GRCh38
rs780197769 Chr.19: 41331172-41331174 on Build GRCh38
rs944749403 Chr.19: 41331005-41331005 on Build GRCh38
rs1053119443 Chr.19: 41330862-41330862 on Build GRCh38
rs929927581 Chr.19: 41331292-41331292 on Build GRCh38
rs201889805 Chr.19: 41330946-41330946 on Build GRCh38
rs745683754 Chr.19: 41331024-41331025 on Build GRCh38
rs1014982176 Chr.19: 41331254-41331254 on Build GRCh38
rs988360047 Chr.19: 41331246-41331246 on Build GRCh38
rs202014722 Chr.19: 41330812-41330812 on Build GRCh38
rs199704340 Chr.19: 41330838-41330838 on Build GRCh38
rs780503471 Chr.19: 41331073-41331073 on Build GRCh38
rs768959025 Chr.19: 41331076-41331076 on Build GRCh38
rs183878618 Chr.19: 41331287-41331287 on Build GRCh38
rs1013236527 Chr.19: 41331279-41331279 on Build GRCh38
rs199636275 Chr.19: 41330916-41330916 on Build GRCh38
rs550662946 Chr.19: 41331070-41331070 on Build GRCh38
rs960386747 Chr.19: 41331257-41331257 on Build GRCh38
rs201700967 Chr.19: 41331071-41331071 on Build GRCh38
rs980524311 Chr.19: 41331204-41331204 on Build GRCh38
rs969095476 Chr.19: 41331203-41331203 on Build GRCh38
rs201584524 Chr.19: 41330836-41330836 on Build GRCh38
rs201564160 Chr.19: 41330915-41330915 on Build GRCh38
rs937001579 Chr.19: 41331101-41331101 on Build GRCh38
rs200729745 Chr.19: 41330923-41330923 on Build GRCh38
rs933310239 Chr.19: 41330895-41330895 on Build GRCh38
rs914198736 Chr.19: 41330879-41330879 on Build GRCh38
rs763943753 Chr.19: 41331186-41331186 on Build GRCh38
rs529010133 Chr.19: 41330885-41330885 on Build GRCh38
rs887658300 Chr.19: 41331049-41331049 on Build GRCh38
rs770027604 Chr.19: 41331042-41331043 on Build GRCh38
rs996008305 Chr.19: 41331035-41331035 on Build GRCh38
rs1004816931 Chr.19: 41331230-41331230 on Build GRCh38
rs1023331355 Chr.19: 41331285-41331285 on Build GRCh38
rs1040463951 Chr.19: 41331010-41331010 on Build GRCh38
rs200340652 Chr.19: 41330830-41330830 on Build GRCh38
rs943475580 Chr.19: 41331061-41331061 on Build GRCh38
rs775550741 Chr.19: 41331170-41331170 on Build GRCh38
rs771748887 Chr.19: 41331169-41331169 on Build GRCh38
rs199982059 Chr.19: 41331218-41331218 on Build GRCh38
rs1048889853 Chr.19: 41331040-41331040 on Build GRCh38
rs201103297 Chr.19: 41330856-41330856 on Build GRCh38
rs372269771 Chr.19: 41331217-41331217 on Build GRCh38
rs537671979 Chr.19: 41331261-41331261 on Build GRCh38
rs1051744180 Chr.19: 41330906-41330906 on Build GRCh38
rs987410076 Chr.19: 41331283-41331283 on Build GRCh38
rs1024663906 Chr.19: 41331199-41331199 on Build GRCh38
rs370517732 Chr.19: 41331121-41331121 on Build GRCh38
rs866738660 Chr.19: 41331129-41331129 on Build GRCh38
rs958640694 Chr.19: 41331284-41331284 on Build GRCh38
rs914127637 Chr.19: 41331256-41331256 on Build GRCh38
rs201096069 Chr.19: 41330959-41330959 on Build GRCh38
rs373910726 Chr.19: 41331163-41331163 on Build GRCh38
rs368054278 Chr.19: 41331178-41331178 on Build GRCh38
rs900617204 Chr.19: 41331030-41331030 on Build GRCh38
rs757277969 Chr.19: 41331025-41331034 on Build GRCh38
rs199849225 Chr.19: 41331155-41331155 on Build GRCh38
rs988979301 Chr.19: 41331020-41331020 on Build GRCh38
rs199517779 Chr.19: 41330872-41330872 on Build GRCh38
rs749364437 Chr.19: 41331229-41331230 on Build GRCh38
rs781566009 Chr.19: 41331083-41331083 on Build GRCh38
rs971141474 Chr.19: 41331051-41331051 on Build GRCh38
rs780991744 Chr.19: 41331025-41331029 on Build GRCh38
rs981119783 Chr.19: 41331072-41331072 on Build GRCh38
rs943424743 Chr.19: 41331288-41331288 on Build GRCh38
rs1046906328 Chr.19: 41331146-41331146 on Build GRCh38
rs200451143 Chr.19: 41330850-41330850 on Build GRCh38
rs541829714 Chr.19: 41331143-41331143 on Build GRCh38
rs974438258 Chr.19: 41331108-41331108 on Build GRCh38
rs971506325 Chr.19: 41331255-41331255 on Build GRCh38
rs758966134 Chr.19: 41331137-41331137 on Build GRCh38
rs1040473871 Chr.19: 41331196-41331196 on Build GRCh38
rs200179008 Chr.19: 41330954-41330954 on Build GRCh38
rs200673279 Chr.19: 41330880-41330880 on Build GRCh38
rs770505137 Chr.19: 41331111-41331111 on Build GRCh38
rs748901865 Chr.19: 41331091-41331091 on Build GRCh38
rs893399552 Chr.19: 41331226-41331226 on Build GRCh38

View Full Product Details