Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48211721-48212228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
UniGene
Hs.446146
Gene Chromosome Location
Chr.19: 48203148-48256269 on Build GRCh38
Transcripts
NM_001184900.1 NM_001184901.1 NM_001184902.1 NM_001184903.1 NM_014959.3 NR_033678.1 NR_033679.1 NR_033680.1 XM_006723091.3 XM_006723092.3 XM_006723093.3 XM_006723094.3 XM_006723095.3 XM_006723096.3 XM_006723097.3 XM_006723098.3 XM_006723099.3 XM_006723101.2 XM_006723102.3 XM_006723104.3 XM_006723106.3 XM_006723107.3 XM_006723109.3 XM_011526641.2 XM_011526642.2 XM_011526643.2 XM_011526644.2 XM_011526646.2 XM_011526647.2 XM_011526648.2 XM_011526650.2 XM_011526653.2 XM_017026482.1 XM_017026483.1 XM_017026484.1 XM_017026485.1 XM_017026486.1 XM_017026487.1 XM_017026488.1 XM_017026489.1 XM_017026490.1 XM_017026492.1 XM_017026493.1 XM_017026494.1 XM_017026495.1 XM_017026496.1 XM_017026497.1 XM_017026498.1 XM_017026499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129946 c.1276C>T p.L426F Chr.19: 48211730-48211730 on Build GRCh38
7201010 c.1593T>C p.Y531Y Chr.19: 48211731-48211731 on Build GRCh38
1712545 c.1601A>C p.Q534P Chr.19: 48211723-48211723 on Build GRCh38
1729866 c.1496A>G p.N499S Chr.19: 48211828-48211828 on Build GRCh38
7201056 c.1596T>C p.L532L Chr.19: 48211728-48211728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754980203 Chr.19: 48211985-48211986 on Build GRCh38
rs958035541 Chr.19: 48212139-48212139 on Build GRCh38
rs149162452 Chr.19: 48211840-48211840 on Build GRCh38
rs760744039 Chr.19: 48212066-48212066 on Build GRCh38
rs924363041 Chr.19: 48212092-48212093 on Build GRCh38
rs3745718 Chr.19: 48211896-48211896 on Build GRCh38
rs777814650 Chr.19: 48212000-48212000 on Build GRCh38
rs754890431 Chr.19: 48211866-48211866 on Build GRCh38
rs780016656 Chr.19: 48211866-48211868 on Build GRCh38
rs756786481 Chr.19: 48211751-48211751 on Build GRCh38
rs754800916 Chr.19: 48211778-48211778 on Build GRCh38
rs1013415417 Chr.19: 48211868-48211868 on Build GRCh38
rs745911441 Chr.19: 48211907-48211907 on Build GRCh38
rs751317554 Chr.19: 48211956-48211956 on Build GRCh38
rs756792561 Chr.19: 48211757-48211757 on Build GRCh38
rs1054554682 Chr.19: 48212142-48212142 on Build GRCh38
rs767068052 Chr.19: 48211765-48211766 on Build GRCh38
rs139483339 Chr.19: 48211950-48211950 on Build GRCh38
rs532121024 Chr.19: 48212000-48212001 on Build GRCh38
rs761074966 Chr.19: 48211822-48211822 on Build GRCh38
rs758006827 Chr.19: 48211766-48211766 on Build GRCh38
rs756028048 Chr.19: 48211987-48211987 on Build GRCh38
rs755632223 Chr.19: 48211929-48211929 on Build GRCh38
rs776993799 Chr.19: 48211935-48211935 on Build GRCh38
rs141801097 Chr.19: 48211948-48211948 on Build GRCh38
rs573785078 Chr.19: 48211893-48211893 on Build GRCh38
rs138779699 Chr.19: 48211875-48211875 on Build GRCh38
rs368359327 Chr.19: 48211803-48211803 on Build GRCh38
rs8112588 Chr.19: 48211779-48211779 on Build GRCh38
rs746082189 Chr.19: 48211801-48211801 on Build GRCh38
rs939770324 Chr.19: 48211979-48211979 on Build GRCh38
rs35920934 Chr.19: 48211775-48211775 on Build GRCh38
rs61755273 Chr.19: 48211908-48211908 on Build GRCh38
rs996190980 Chr.19: 48212159-48212159 on Build GRCh38
rs945544090 Chr.19: 48212020-48212020 on Build GRCh38
rs34477929 Chr.19: 48212124-48212124 on Build GRCh38
rs764776725 Chr.19: 48211752-48211752 on Build GRCh38
rs781123298 Chr.19: 48211882-48211882 on Build GRCh38
rs371795173 Chr.19: 48211927-48211927 on Build GRCh38
rs1032355863 Chr.19: 48212136-48212136 on Build GRCh38
rs1032324679 Chr.19: 48211913-48211913 on Build GRCh38
rs770291118 Chr.19: 48211947-48211947 on Build GRCh38
rs756075950 Chr.19: 48211841-48211842 on Build GRCh38
rs749260047 Chr.19: 48211784-48211784 on Build GRCh38
rs749211389 Chr.19: 48211972-48211972 on Build GRCh38
rs558188524 Chr.19: 48211814-48211814 on Build GRCh38
rs1024589650 Chr.19: 48211906-48211906 on Build GRCh38
rs146199265 Chr.19: 48211859-48211859 on Build GRCh38
rs767222046 Chr.19: 48211969-48211969 on Build GRCh38
rs190997806 Chr.19: 48211749-48211749 on Build GRCh38
rs764409113 Chr.19: 48211831-48211831 on Build GRCh38
rs766186900 Chr.19: 48211862-48211862 on Build GRCh38
rs1013223816 Chr.19: 48212161-48212161 on Build GRCh38
rs375457501 Chr.19: 48211981-48211981 on Build GRCh38
rs753686618 Chr.19: 48212008-48212008 on Build GRCh38
rs571589529 Chr.19: 48212200-48212200 on Build GRCh38
rs748050357 Chr.19: 48211890-48211890 on Build GRCh38
rs529527612 Chr.19: 48212123-48212123 on Build GRCh38
rs780293112 Chr.19: 48211914-48211914 on Build GRCh38
rs531890099 Chr.19: 48212164-48212164 on Build GRCh38
rs995434554 Chr.19: 48211811-48211811 on Build GRCh38
rs551898633 Chr.19: 48212198-48212198 on Build GRCh38
rs749876926 Chr.19: 48211856-48211856 on Build GRCh38
rs747242060 Chr.19: 48212011-48212011 on Build GRCh38
rs749851325 Chr.19: 48211796-48211797 on Build GRCh38
rs1013863362 Chr.19: 48212047-48212047 on Build GRCh38
rs999862762 Chr.19: 48211837-48211837 on Build GRCh38
rs778506041 Chr.19: 48212001-48212001 on Build GRCh38
rs758539025 Chr.19: 48212010-48212010 on Build GRCh38
rs898139743 Chr.19: 48211808-48211808 on Build GRCh38
rs549609094 Chr.19: 48212134-48212134 on Build GRCh38
rs764663963 Chr.19: 48211848-48211848 on Build GRCh38
rs774431953 Chr.19: 48211787-48211787 on Build GRCh38
rs549570428 Chr.19: 48211768-48211768 on Build GRCh38
rs34632751 Chr.19: 48211939-48211939 on Build GRCh38
rs2008527 Chr.19: 48212160-48212160 on Build GRCh38
rs770975518 Chr.19: 48211785-48211785 on Build GRCh38
rs769063453 Chr.19: 48211934-48211934 on Build GRCh38
rs150504639 Chr.19: 48211949-48211949 on Build GRCh38
rs750097954 Chr.19: 48211755-48211755 on Build GRCh38
rs2008521 Chr.19: 48212012-48212012 on Build GRCh38
rs781437480 Chr.19: 48212014-48212014 on Build GRCh38
rs751229617 Chr.19: 48211863-48211863 on Build GRCh38
rs188716648 Chr.19: 48211965-48211965 on Build GRCh38
rs374276938 Chr.19: 48212177-48212177 on Build GRCh38
rs146657505 Chr.19: 48211841-48211841 on Build GRCh38
rs954971163 Chr.19: 48212166-48212166 on Build GRCh38
rs11669386 Chr.19: 48212089-48212089 on Build GRCh38
rs987648875 Chr.19: 48211964-48211964 on Build GRCh38
rs775684490 Chr.19: 48212031-48212031 on Build GRCh38

View Full Product Details