Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48221547-48222094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
UniGene
Hs.446146
Gene Chromosome Location
Chr.19: 48203148-48256269 on Build GRCh38
Transcripts
NM_001184900.1 NM_001184901.1 NM_001184902.1 NM_001184903.1 NM_014959.3 NR_033678.1 NR_033679.1 NR_033680.1 XM_006723091.3 XM_006723092.3 XM_006723093.3 XM_006723094.3 XM_006723095.3 XM_006723096.3 XM_006723097.3 XM_006723098.3 XM_006723099.3 XM_006723101.2 XM_006723102.3 XM_006723104.3 XM_006723106.3 XM_006723107.3 XM_006723109.3 XM_011526641.2 XM_011526642.2 XM_011526643.2 XM_011526644.2 XM_011526646.2 XM_011526647.2 XM_011526648.2 XM_011526650.2 XM_011526653.2 XM_017026482.1 XM_017026483.1 XM_017026484.1 XM_017026485.1 XM_017026486.1 XM_017026487.1 XM_017026488.1 XM_017026489.1 XM_017026490.1 XM_017026491.1 XM_017026492.1 XM_017026493.1 XM_017026494.1 XM_017026495.1 XM_017026496.1 XM_017026497.1 XM_017026498.1 XM_017026499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199579 c.1046A>G p.D349G Chr.19: 48221845-48221845 on Build GRCh38
3107044 c.740G>A p.R247H Chr.19: 48221833-48221833 on Build GRCh38
5856307 c.1136C>T p.S379F Chr.19: 48221755-48221755 on Build GRCh38
5724516 c.719C>A p.A240E Chr.19: 48221854-48221854 on Build GRCh38
5856305 c.1149A>T p.K383N Chr.19: 48221742-48221742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013889254 Chr.19: 48221801-48221801 on Build GRCh38
rs1014071865 Chr.19: 48221965-48221965 on Build GRCh38
rs76090926 Chr.19: 48221629-48221629 on Build GRCh38
rs759504038 Chr.19: 48221884-48221884 on Build GRCh38
rs769398386 Chr.19: 48221863-48221863 on Build GRCh38
rs374000674 Chr.19: 48221732-48221732 on Build GRCh38
rs577706628 Chr.19: 48221871-48221871 on Build GRCh38
rs559636832 Chr.19: 48222062-48222062 on Build GRCh38
rs901370223 Chr.19: 48221735-48221735 on Build GRCh38
rs780893730 Chr.19: 48221777-48221777 on Build GRCh38
rs768128637 Chr.19: 48221769-48221769 on Build GRCh38
rs149003172 Chr.19: 48221797-48221797 on Build GRCh38
rs151036070 Chr.19: 48221825-48221825 on Build GRCh38
rs374214594 Chr.19: 48221831-48221831 on Build GRCh38
rs1034263962 Chr.19: 48221895-48221895 on Build GRCh38
rs776290879 Chr.19: 48221917-48221918 on Build GRCh38
rs1004816247 Chr.19: 48222012-48222012 on Build GRCh38
rs764486100 Chr.19: 48221614-48221614 on Build GRCh38
rs143813106 Chr.19: 48221734-48221734 on Build GRCh38
rs754081264 Chr.19: 48221717-48221717 on Build GRCh38
rs758762341 Chr.19: 48221742-48221742 on Build GRCh38
rs868210323 Chr.19: 48221950-48221950 on Build GRCh38
rs776784381 Chr.19: 48221889-48221889 on Build GRCh38
rs899006101 Chr.19: 48222032-48222032 on Build GRCh38
rs993356846 Chr.19: 48221586-48221586 on Build GRCh38
rs775658778 Chr.19: 48221842-48221844 on Build GRCh38
rs376143416 Chr.19: 48221701-48221701 on Build GRCh38
rs925980133 Chr.19: 48222009-48222009 on Build GRCh38
rs978809206 Chr.19: 48222000-48222000 on Build GRCh38
rs13343747 Chr.19: 48221991-48221991 on Build GRCh38
rs557625355 Chr.19: 48221870-48221870 on Build GRCh38
rs747789818 Chr.19: 48221780-48221780 on Build GRCh38
rs768868556 Chr.19: 48221886-48221886 on Build GRCh38
rs541969712 Chr.19: 48221729-48221729 on Build GRCh38
rs528840021 Chr.19: 48221974-48221974 on Build GRCh38
rs555715998 Chr.19: 48221819-48221819 on Build GRCh38
rs934212757 Chr.19: 48222050-48222050 on Build GRCh38
rs752681583 Chr.19: 48221687-48221687 on Build GRCh38
rs781067304 Chr.19: 48222024-48222024 on Build GRCh38
rs757762638 Chr.19: 48221721-48221721 on Build GRCh38
rs140600610 Chr.19: 48221798-48221798 on Build GRCh38
rs16981828 Chr.19: 48221805-48221805 on Build GRCh38
rs922859193 Chr.19: 48222034-48222034 on Build GRCh38
rs146809917 Chr.19: 48222068-48222068 on Build GRCh38
rs559743935 Chr.19: 48221963-48221963 on Build GRCh38
rs751266811 Chr.19: 48221997-48221997 on Build GRCh38
rs753412730 Chr.19: 48221634-48221634 on Build GRCh38
rs909337038 Chr.19: 48221594-48221594 on Build GRCh38
rs908682292 Chr.19: 48222058-48222058 on Build GRCh38
rs926139745 Chr.19: 48221934-48221934 on Build GRCh38
rs767640791 Chr.19: 48221679-48221679 on Build GRCh38
rs754534514 Chr.19: 48222019-48222019 on Build GRCh38
rs886385827 Chr.19: 48222011-48222011 on Build GRCh38
rs762067591 Chr.19: 48221820-48221820 on Build GRCh38
rs752846364 Chr.19: 48221711-48221711 on Build GRCh38
rs367887359 Chr.19: 48221764-48221764 on Build GRCh38
rs765700609 Chr.19: 48221823-48221823 on Build GRCh38
rs761549453 Chr.19: 48221880-48221881 on Build GRCh38
rs927935747 Chr.19: 48221779-48221779 on Build GRCh38
rs540915752 Chr.19: 48221833-48221833 on Build GRCh38
rs779644048 Chr.19: 48221747-48221747 on Build GRCh38
rs978868419 Chr.19: 48221922-48221922 on Build GRCh38
rs147714341 Chr.19: 48222057-48222057 on Build GRCh38
rs772927054 Chr.19: 48221791-48221791 on Build GRCh38
rs963428493 Chr.19: 48221827-48221827 on Build GRCh38
rs752071016 Chr.19: 48221832-48221832 on Build GRCh38
rs946205009 Chr.19: 48221913-48221913 on Build GRCh38
rs762153721 Chr.19: 48221896-48221896 on Build GRCh38
rs1001446432 Chr.19: 48221867-48221867 on Build GRCh38
rs764460685 Chr.19: 48221712-48221712 on Build GRCh38
rs754170971 Chr.19: 48221813-48221813 on Build GRCh38
rs764264265 Chr.19: 48221806-48221806 on Build GRCh38
rs61755274 Chr.19: 48221781-48221781 on Build GRCh38
rs142575238 Chr.19: 48222061-48222061 on Build GRCh38
rs774221552 Chr.19: 48221880-48221880 on Build GRCh38
rs747806231 Chr.19: 48221853-48221853 on Build GRCh38
rs777647245 Chr.19: 48221869-48221869 on Build GRCh38
rs73572976 Chr.19: 48221571-48221571 on Build GRCh38
rs539936351 Chr.19: 48221879-48221879 on Build GRCh38
rs758963965 Chr.19: 48221829-48221829 on Build GRCh38
rs140937765 Chr.19: 48221834-48221834 on Build GRCh38
rs934161281 Chr.19: 48221775-48221775 on Build GRCh38
rs913393970 Chr.19: 48221708-48221711 on Build GRCh38
rs978778071 Chr.19: 48221674-48221674 on Build GRCh38
rs113490923 Chr.19: 48221630-48221630 on Build GRCh38
rs555170984 Chr.19: 48221605-48221605 on Build GRCh38

View Full Product Details