Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97724324-97724824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
UniGene
Hs.234569
Gene Chromosome Location
Chr.2: 97712030-97744327 on Build GRCh38
Transcripts
NM_001079.3 XM_017004867.1 XM_017004868.1 XM_017004869.1 XM_017004870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6534528 c.291C>G p.N97K Chr.2: 97724327-97724327 on Build GRCh38
4618272 c.370C>T p.R124C Chr.2: 97724406-97724406 on Build GRCh38
5748611 c.335C>T p.P112L Chr.2: 97724371-97724371 on Build GRCh38
3840327 c.303G>A p.P101P Chr.2: 97724339-97724339 on Build GRCh38
5508801 c.360C>T p.D120D Chr.2: 97724396-97724396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371153389 Chr.2: 97724471-97724471 on Build GRCh38
rs767294957 Chr.2: 97724482-97724482 on Build GRCh38
rs919324397 Chr.2: 97724547-97724547 on Build GRCh38
rs560210342 Chr.2: 97724527-97724527 on Build GRCh38
rs774210535 Chr.2: 97724617-97724617 on Build GRCh38
rs200107228 Chr.2: 97724441-97724441 on Build GRCh38
rs866378859 Chr.2: 97724667-97724667 on Build GRCh38
rs925144320 Chr.2: 97724463-97724463 on Build GRCh38
rs781033785 Chr.2: 97724353-97724353 on Build GRCh38
rs552541836 Chr.2: 97724675-97724675 on Build GRCh38
rs1002377206 Chr.2: 97724616-97724616 on Build GRCh38
rs754511997 Chr.2: 97724722-97724722 on Build GRCh38
rs868316208 Chr.2: 97724572-97724572 on Build GRCh38
rs768492740 Chr.2: 97724396-97724396 on Build GRCh38
rs55679020 Chr.2: 97724347-97724347 on Build GRCh38
rs1000618778 Chr.2: 97724684-97724684 on Build GRCh38
rs542973270 Chr.2: 97724351-97724351 on Build GRCh38
rs902260642 Chr.2: 97724670-97724670 on Build GRCh38
rs559342309 Chr.2: 97724361-97724361 on Build GRCh38
rs768332464 Chr.2: 97724486-97724486 on Build GRCh38
rs1049218289 Chr.2: 97724568-97724568 on Build GRCh38
rs867787314 Chr.2: 97724506-97724506 on Build GRCh38
rs55824472 Chr.2: 97724465-97724465 on Build GRCh38
rs565708035 Chr.2: 97724736-97724736 on Build GRCh38
rs967386356 Chr.2: 97724639-97724639 on Build GRCh38
rs13006704 Chr.2: 97724657-97724657 on Build GRCh38
rs200514384 Chr.2: 97724413-97724413 on Build GRCh38
rs750345154 Chr.2: 97724487-97724487 on Build GRCh38
rs747793121 Chr.2: 97724729-97724729 on Build GRCh38
rs770900178 Chr.2: 97724496-97724496 on Build GRCh38
rs751420623 Chr.2: 97724346-97724346 on Build GRCh38
rs929429105 Chr.2: 97724562-97724562 on Build GRCh38
rs757033990 Chr.2: 97724481-97724481 on Build GRCh38
rs1010883080 Chr.2: 97724601-97724601 on Build GRCh38
rs111771234 Chr.2: 97724379-97724379 on Build GRCh38
rs765313500 Chr.2: 97724444-97724444 on Build GRCh38
rs76085176 Chr.2: 97724725-97724725 on Build GRCh38
rs1021105226 Chr.2: 97724366-97724366 on Build GRCh38
rs896757271 Chr.2: 97724638-97724638 on Build GRCh38
rs748956300 Chr.2: 97724390-97724390 on Build GRCh38
rs887950494 Chr.2: 97724584-97724584 on Build GRCh38
rs944650476 Chr.2: 97724790-97724790 on Build GRCh38
rs55845489 Chr.2: 97724345-97724345 on Build GRCh38
rs959527712 Chr.2: 97724664-97724664 on Build GRCh38
rs982370975 Chr.2: 97724651-97724651 on Build GRCh38
rs201551373 Chr.2: 97724405-97724405 on Build GRCh38
rs545129970 Chr.2: 97724451-97724451 on Build GRCh38
rs779657149 Chr.2: 97724375-97724375 on Build GRCh38
rs199996419 Chr.2: 97724408-97724408 on Build GRCh38
rs774255492 Chr.2: 97724539-97724539 on Build GRCh38
rs1040401534 Chr.2: 97724503-97724503 on Build GRCh38
rs55668444 Chr.2: 97724376-97724376 on Build GRCh38
rs996968760 Chr.2: 97724540-97724540 on Build GRCh38
rs777031626 Chr.2: 97724426-97724426 on Build GRCh38
rs1055799226 Chr.2: 97724620-97724620 on Build GRCh38
rs780138156 Chr.2: 97724665-97724665 on Build GRCh38
rs990534155 Chr.2: 97724705-97724705 on Build GRCh38
rs56264206 Chr.2: 97724344-97724344 on Build GRCh38
rs201910784 Chr.2: 97724468-97724468 on Build GRCh38
rs755602828 Chr.2: 97724360-97724360 on Build GRCh38
rs958031543 Chr.2: 97724598-97724598 on Build GRCh38
rs549714930 Chr.2: 97724566-97724566 on Build GRCh38
rs985369366 Chr.2: 97724735-97724735 on Build GRCh38
rs747379748 Chr.2: 97724407-97724407 on Build GRCh38
rs774131317 Chr.2: 97724406-97724406 on Build GRCh38
rs1001713813 Chr.2: 97724749-97724749 on Build GRCh38
rs745778680 Chr.2: 97724355-97724355 on Build GRCh38
rs1008750552 Chr.2: 97724378-97724378 on Build GRCh38
rs748868410 Chr.2: 97724679-97724679 on Build GRCh38
rs2289919 Chr.2: 97724732-97724732 on Build GRCh38

View Full Product Details