Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42708579-42709077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
UniGene
Hs.604182
Gene Chromosome Location
Chr.3: 42704118-42773253 on Build GRCh38
Transcripts
NM_144719.3 XM_005264898.3 XM_011533418.2 XM_011533419.2 XM_017005781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190138 c.2080G>T p.E694* Chr.3: 42709048-42709048 on Build GRCh38
1199821 c.2133G>T p.K711N Chr.3: 42708995-42708995 on Build GRCh38
6621434 c.2125C>T p.Q709* Chr.3: 42709003-42709003 on Build GRCh38
7359740 c.2137G>T p.G713C Chr.3: 42708991-42708991 on Build GRCh38
3066251 c.2123G>A p.R708Q Chr.3: 42709005-42709005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567654483 Chr.3: 42708794-42708794 on Build GRCh38
rs372226961 Chr.3: 42708894-42708894 on Build GRCh38
rs141942240 Chr.3: 42709006-42709006 on Build GRCh38
rs923515954 Chr.3: 42708691-42708691 on Build GRCh38
rs746071139 Chr.3: 42708995-42708995 on Build GRCh38
rs200174039 Chr.3: 42709024-42709024 on Build GRCh38
rs374039046 Chr.3: 42708962-42708962 on Build GRCh38
rs200473600 Chr.3: 42708928-42708928 on Build GRCh38
rs999069235 Chr.3: 42708762-42708762 on Build GRCh38
rs768052445 Chr.3: 42708939-42708939 on Build GRCh38
rs751305770 Chr.3: 42709050-42709050 on Build GRCh38
rs761219422 Chr.3: 42708930-42708930 on Build GRCh38
rs1002338420 Chr.3: 42708886-42708886 on Build GRCh38
rs532465318 Chr.3: 42708699-42708699 on Build GRCh38
rs938663524 Chr.3: 42708764-42708764 on Build GRCh38
rs754878917 Chr.3: 42708982-42708982 on Build GRCh38
rs560474255 Chr.3: 42708650-42708650 on Build GRCh38
rs562708393 Chr.3: 42708746-42708746 on Build GRCh38
rs141541158 Chr.3: 42709009-42709009 on Build GRCh38
rs566499550 Chr.3: 42708893-42708893 on Build GRCh38
rs201648184 Chr.3: 42708999-42708999 on Build GRCh38
rs766191990 Chr.3: 42709046-42709046 on Build GRCh38
rs772864738 Chr.3: 42709026-42709026 on Build GRCh38
rs183257754 Chr.3: 42708751-42708751 on Build GRCh38
rs747544106 Chr.3: 42709000-42709000 on Build GRCh38
rs943477566 Chr.3: 42709015-42709015 on Build GRCh38
rs972013117 Chr.3: 42708918-42708918 on Build GRCh38
rs2240861 Chr.3: 42708832-42708832 on Build GRCh38
rs777730260 Chr.3: 42708986-42708986 on Build GRCh38
rs971947515 Chr.3: 42708617-42708617 on Build GRCh38
rs759228351 Chr.3: 42709051-42709051 on Build GRCh38
rs746348067 Chr.3: 42709009-42709009 on Build GRCh38
rs776146571 Chr.3: 42708951-42708951 on Build GRCh38
rs893346192 Chr.3: 42708782-42708782 on Build GRCh38
rs368060583 Chr.3: 42708953-42708953 on Build GRCh38
rs1021846014 Chr.3: 42708864-42708864 on Build GRCh38
rs868590399 Chr.3: 42708778-42708778 on Build GRCh38
rs984623217 Chr.3: 42709003-42709003 on Build GRCh38
rs773063970 Chr.3: 42709032-42709032 on Build GRCh38
rs1057189888 Chr.3: 42708785-42708785 on Build GRCh38
rs930459472 Chr.3: 42708655-42708655 on Build GRCh38
rs367554932 Chr.3: 42709035-42709035 on Build GRCh38
rs969147171 Chr.3: 42708870-42708870 on Build GRCh38
rs1022494798 Chr.3: 42708847-42708847 on Build GRCh38
rs1049645600 Chr.3: 42708693-42708693 on Build GRCh38
rs552473685 Chr.3: 42708743-42708743 on Build GRCh38
rs766113750 Chr.3: 42708963-42708963 on Build GRCh38
rs765174389 Chr.3: 42708895-42708895 on Build GRCh38
rs769257896 Chr.3: 42709005-42709005 on Build GRCh38
rs889192399 Chr.3: 42708714-42708714 on Build GRCh38
rs569475549 Chr.3: 42708996-42708996 on Build GRCh38
rs754010521 Chr.3: 42708991-42708991 on Build GRCh38
rs546602763 Chr.3: 42708885-42708885 on Build GRCh38
rs751529017 Chr.3: 42708979-42708979 on Build GRCh38
rs986665240 Chr.3: 42708733-42708733 on Build GRCh38
rs757283058 Chr.3: 42708994-42708994 on Build GRCh38
rs548041884 Chr.3: 42708760-42708760 on Build GRCh38
rs531366769 Chr.3: 42708749-42708749 on Build GRCh38
rs2240860 Chr.3: 42708954-42708954 on Build GRCh38
rs138479423 Chr.3: 42709007-42709007 on Build GRCh38
rs953735338 Chr.3: 42708921-42708921 on Build GRCh38
rs776760869 Chr.3: 42708774-42708774 on Build GRCh38

View Full Product Details