Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122421789-122422287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
UniGene
Hs.161008
Gene Chromosome Location
Chr.3: 122421901-122514939 on Build GRCh38
Transcripts
NM_002264.3 NR_026698.1 NR_125405.1

Gene Symbol
LOC102723582
Entrez Gene ID
102723582
Gene Name
uncharacterized LOC102723582
Gene Aliases
-
UniGene
Hs.675056
Gene Chromosome Location
Chr.3: 122416206-122443247 on Build GRCh38
Transcripts
NM_002264.3 NR_026698.1 NR_125405.1

SNP Detail

SNP ID Chromosome Location
rs897291019 Chr.3: 122422242-122422242 on Build GRCh38
rs755106493 Chr.3: 122422034-122422034 on Build GRCh38
rs1019360290 Chr.3: 122421980-122421980 on Build GRCh38
rs987848892 Chr.3: 122421917-122421917 on Build GRCh38
rs770377288 Chr.3: 122422160-122422160 on Build GRCh38
rs532204432 Chr.3: 122422125-122422125 on Build GRCh38
rs115874925 Chr.3: 122421894-122421894 on Build GRCh38
rs903609741 Chr.3: 122422263-122422263 on Build GRCh38
rs866238574 Chr.3: 122422101-122422101 on Build GRCh38
rs749602224 Chr.3: 122422243-122422243 on Build GRCh38
rs996722151 Chr.3: 122421892-122421892 on Build GRCh38
rs575190129 Chr.3: 122421913-122421913 on Build GRCh38
rs544899033 Chr.3: 122421815-122421815 on Build GRCh38
rs1037190280 Chr.3: 122421818-122421818 on Build GRCh38
rs184890366 Chr.3: 122422001-122422001 on Build GRCh38
rs925280739 Chr.3: 122422113-122422113 on Build GRCh38
rs1020094556 Chr.3: 122422029-122422029 on Build GRCh38
rs552523539 Chr.3: 122422259-122422259 on Build GRCh38
rs866755149 Chr.3: 122422148-122422148 on Build GRCh38
rs955326208 Chr.3: 122421900-122421900 on Build GRCh38
rs540187087 Chr.3: 122422090-122422090 on Build GRCh38
rs747255506 Chr.3: 122422027-122422027 on Build GRCh38
rs972111542 Chr.3: 122421984-122421984 on Build GRCh38
rs564319691 Chr.3: 122422121-122422121 on Build GRCh38
rs576565427 Chr.3: 122422212-122422212 on Build GRCh38
rs980905675 Chr.3: 122422033-122422033 on Build GRCh38
rs556037852 Chr.3: 122421996-122421996 on Build GRCh38
rs536425747 Chr.3: 122422233-122422233 on Build GRCh38
rs988629104 Chr.3: 122422021-122422021 on Build GRCh38
rs758128745 Chr.3: 122421993-122421993 on Build GRCh38
rs995747297 Chr.3: 122421875-122421875 on Build GRCh38
rs990812823 Chr.3: 122422138-122422138 on Build GRCh38
rs1006596691 Chr.3: 122421976-122421976 on Build GRCh38
rs372934551 Chr.3: 122422120-122422121 on Build GRCh38
rs367998569 Chr.3: 122421872-122421872 on Build GRCh38
rs1018967728 Chr.3: 122421924-122421924 on Build GRCh38
rs945052894 Chr.3: 122422174-122422174 on Build GRCh38
rs55781608 Chr.3: 122422072-122422072 on Build GRCh38
rs529644964 Chr.3: 122422084-122422084 on Build GRCh38
rs1009337681 Chr.3: 122421862-122421862 on Build GRCh38
rs773623475 Chr.3: 122422231-122422231 on Build GRCh38
rs1056511191 Chr.3: 122422123-122422123 on Build GRCh38
rs949428232 Chr.3: 122422179-122422179 on Build GRCh38
rs918037245 Chr.3: 122422140-122422140 on Build GRCh38

View Full Product Details