Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55087523-55088028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDR
Entrez Gene ID
3791
Gene Name
kinase insert domain receptor
Gene Aliases
CD309, FLK1, VEGFR, VEGFR2
UniGene
Hs.479756
Gene Chromosome Location
Chr.4: 55078259-55125595 on Build GRCh38
Transcripts
NM_002253.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3917934 c.3628C>T p.P1210S Chr.4: 55087641-55087641 on Build GRCh38
6437969 c.3511-7G>A p.? Chr.4: 55087765-55087765 on Build GRCh38
4755721 c.3511-1G>C p.? Chr.4: 55087759-55087759 on Build GRCh38
6938959 c.3546G>T p.E1182D Chr.4: 55087723-55087723 on Build GRCh38
6437967 c.3511G>A p.D1171N Chr.4: 55087758-55087758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768576998 Chr.4: 55087717-55087717 on Build GRCh38
rs908693616 Chr.4: 55087893-55087893 on Build GRCh38
rs762411907 Chr.4: 55087572-55087572 on Build GRCh38
rs11540507 Chr.4: 55087641-55087641 on Build GRCh38
rs950287107 Chr.4: 55087553-55087553 on Build GRCh38
rs139212484 Chr.4: 55087852-55087852 on Build GRCh38
rs375416763 Chr.4: 55087595-55087595 on Build GRCh38
rs143459233 Chr.4: 55087731-55087731 on Build GRCh38
rs1011564548 Chr.4: 55087660-55087660 on Build GRCh38
rs768199268 Chr.4: 55087702-55087702 on Build GRCh38
rs191767790 Chr.4: 55087581-55087581 on Build GRCh38
rs183234342 Chr.4: 55087925-55087925 on Build GRCh38
rs760114131 Chr.4: 55087730-55087730 on Build GRCh38
rs762732518 Chr.4: 55087652-55087654 on Build GRCh38
rs757239496 Chr.4: 55087784-55087784 on Build GRCh38
rs538723560 Chr.4: 55087662-55087662 on Build GRCh38
rs551498433 Chr.4: 55087546-55087546 on Build GRCh38
rs111398067 Chr.4: 55087831-55087831 on Build GRCh38
rs869107800 Chr.4: 55087581-55087582 on Build GRCh38
rs974262082 Chr.4: 55087931-55087931 on Build GRCh38
rs565460125 Chr.4: 55087547-55087547 on Build GRCh38
rs369296279 Chr.4: 55087584-55087584 on Build GRCh38
rs777625915 Chr.4: 55087648-55087648 on Build GRCh38
rs774791565 Chr.4: 55087706-55087706 on Build GRCh38
rs1020785732 Chr.4: 55087775-55087775 on Build GRCh38
rs900931519 Chr.4: 55087577-55087577 on Build GRCh38
rs869289403 Chr.4: 55087586-55087587 on Build GRCh38
rs746962241 Chr.4: 55087657-55087657 on Build GRCh38
rs200996447 Chr.4: 55087622-55087622 on Build GRCh38
rs574342836 Chr.4: 55087825-55087825 on Build GRCh38
rs868172841 Chr.4: 55087664-55087664 on Build GRCh38
rs770086421 Chr.4: 55087738-55087740 on Build GRCh38
rs779617216 Chr.4: 55087671-55087671 on Build GRCh38
rs971875379 Chr.4: 55087865-55087865 on Build GRCh38
rs7682740 Chr.4: 55087881-55087881 on Build GRCh38
rs146738218 Chr.4: 55087756-55087756 on Build GRCh38
rs745611872 Chr.4: 55087666-55087666 on Build GRCh38
rs764514760 Chr.4: 55087573-55087573 on Build GRCh38
rs755024874 Chr.4: 55087635-55087635 on Build GRCh38
rs7682773 Chr.4: 55087936-55087936 on Build GRCh38
rs867819358 Chr.4: 55087632-55087632 on Build GRCh38
rs755585942 Chr.4: 55087576-55087576 on Build GRCh38
rs771605584 Chr.4: 55087575-55087576 on Build GRCh38
rs145493938 Chr.4: 55087665-55087665 on Build GRCh38
rs976418206 Chr.4: 55087962-55087962 on Build GRCh38
rs201171220 Chr.4: 55087585-55087585 on Build GRCh38
rs932400268 Chr.4: 55087741-55087741 on Build GRCh38
rs961619426 Chr.4: 55087908-55087908 on Build GRCh38
rs200251218 Chr.4: 55087807-55087807 on Build GRCh38
rs902490777 Chr.4: 55087699-55087699 on Build GRCh38
rs137873945 Chr.4: 55087732-55087732 on Build GRCh38
rs551175926 Chr.4: 55087558-55087558 on Build GRCh38
rs991898537 Chr.4: 55087851-55087851 on Build GRCh38
rs754095453 Chr.4: 55087737-55087737 on Build GRCh38
rs902453458 Chr.4: 55087780-55087780 on Build GRCh38
rs376242865 Chr.4: 55087588-55087588 on Build GRCh38
rs1033021479 Chr.4: 55087794-55087794 on Build GRCh38
rs890866273 Chr.4: 55087758-55087758 on Build GRCh38
rs769469885 Chr.4: 55087668-55087668 on Build GRCh38
rs1000757723 Chr.4: 55087790-55087790 on Build GRCh38
rs752218796 Chr.4: 55087636-55087636 on Build GRCh38
rs767112123 Chr.4: 55087603-55087603 on Build GRCh38
rs1012322904 Chr.4: 55087808-55087808 on Build GRCh38
rs756385325 Chr.4: 55087590-55087590 on Build GRCh38
rs769341200 Chr.4: 55087559-55087559 on Build GRCh38
rs531038897 Chr.4: 55087554-55087554 on Build GRCh38
rs1022745467 Chr.4: 55087825-55087825 on Build GRCh38
rs932372961 Chr.4: 55087953-55087953 on Build GRCh38
rs748810441 Chr.4: 55087685-55087685 on Build GRCh38
rs1042670223 Chr.4: 55087663-55087663 on Build GRCh38
rs940258117 Chr.4: 55087647-55087647 on Build GRCh38
rs567828738 Chr.4: 55087563-55087563 on Build GRCh38
rs537611846 Chr.4: 55087769-55087769 on Build GRCh38
rs894012405 Chr.4: 55087652-55087652 on Build GRCh38
rs750203024 Chr.4: 55087971-55087971 on Build GRCh38
rs568957983 Chr.4: 55087765-55087765 on Build GRCh38

View Full Product Details