Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122451354-122451864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2
Entrez Gene ID
3558
Gene Name
interleukin 2
Gene Aliases
IL-2, TCGF, lymphokine
UniGene
Hs.89679
Gene Chromosome Location
Chr.4: 122449479-122456495 on Build GRCh38
Transcripts
NM_000586.3 XM_017008177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694947 c.352G>A p.G118R Chr.4: 122451862-122451862 on Build GRCh38
3599779 c.429C>T p.T143T Chr.4: 122451785-122451785 on Build GRCh38
402509 c.409T>A p.F137I Chr.4: 122451805-122451805 on Build GRCh38
6098950 c.392C>A p.T131K Chr.4: 122451822-122451822 on Build GRCh38
5612591 c.447C>T p.I149I Chr.4: 122451767-122451767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529865285 Chr.4: 122451760-122451760 on Build GRCh38
rs953534267 Chr.4: 122451453-122451453 on Build GRCh38
rs200621841 Chr.4: 122451725-122451725 on Build GRCh38
rs565206790 Chr.4: 122451537-122451537 on Build GRCh38
rs1016159924 Chr.4: 122451809-122451809 on Build GRCh38
rs909714041 Chr.4: 122451592-122451592 on Build GRCh38
rs750932763 Chr.4: 122451744-122451744 on Build GRCh38
rs1055790067 Chr.4: 122451647-122451647 on Build GRCh38
rs903182581 Chr.4: 122451788-122451788 on Build GRCh38
rs997719159 Chr.4: 122451615-122451615 on Build GRCh38
rs369663857 Chr.4: 122451515-122451515 on Build GRCh38
rs867145253 Chr.4: 122451694-122451694 on Build GRCh38
rs765716210 Chr.4: 122451736-122451736 on Build GRCh38
rs937217405 Chr.4: 122451626-122451626 on Build GRCh38
rs1051753 Chr.4: 122451758-122451758 on Build GRCh38
rs965023593 Chr.4: 122451528-122451528 on Build GRCh38
rs777464920 Chr.4: 122451810-122451810 on Build GRCh38
rs766471427 Chr.4: 122451654-122451654 on Build GRCh38
rs901601673 Chr.4: 122451553-122451553 on Build GRCh38
rs764240155 Chr.4: 122451718-122451718 on Build GRCh38
rs543296992 Chr.4: 122451796-122451796 on Build GRCh38
rs76000366 Chr.4: 122451445-122451445 on Build GRCh38
rs760829674 Chr.4: 122451704-122451704 on Build GRCh38
rs758867724 Chr.4: 122451748-122451748 on Build GRCh38
rs371057839 Chr.4: 122451726-122451726 on Build GRCh38
rs911561997 Chr.4: 122451765-122451765 on Build GRCh38
rs770442564 Chr.4: 122451833-122451833 on Build GRCh38
rs1018074398 Chr.4: 122451524-122451524 on Build GRCh38
rs572211926 Chr.4: 122451540-122451540 on Build GRCh38
rs540787352 Chr.4: 122451556-122451556 on Build GRCh38
rs1033140216 Chr.4: 122451388-122451388 on Build GRCh38
rs1054473361 Chr.4: 122451423-122451423 on Build GRCh38
rs1006636965 Chr.4: 122451509-122451509 on Build GRCh38
rs976417238 Chr.4: 122451534-122451534 on Build GRCh38
rs865810716 Chr.4: 122451819-122451819 on Build GRCh38
rs984453861 Chr.4: 122451591-122451591 on Build GRCh38
rs377374894 Chr.4: 122451829-122451829 on Build GRCh38
rs374465594 Chr.4: 122451789-122451789 on Build GRCh38

View Full Product Details