Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122451719-122452219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2
Entrez Gene ID
3558
Gene Name
interleukin 2
Gene Aliases
IL-2, TCGF, lymphokine
UniGene
Hs.89679
Gene Chromosome Location
Chr.4: 122449479-122456495 on Build GRCh38
Transcripts
NM_000586.3 XM_017008177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694947 c.352G>A p.G118R Chr.4: 122451862-122451862 on Build GRCh38
402509 c.409T>A p.F137I Chr.4: 122451805-122451805 on Build GRCh38
6098950 c.392C>A p.T131K Chr.4: 122451822-122451822 on Build GRCh38
6217343 c.352-8T>C p.? Chr.4: 122451870-122451870 on Build GRCh38
4442507 c.440G>A p.S147N Chr.4: 122451774-122451774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529865285 Chr.4: 122451760-122451760 on Build GRCh38
rs924626364 Chr.4: 122452197-122452197 on Build GRCh38
rs991251959 Chr.4: 122452109-122452109 on Build GRCh38
rs528183004 Chr.4: 122452051-122452051 on Build GRCh38
rs909828782 Chr.4: 122452068-122452068 on Build GRCh38
rs1016159924 Chr.4: 122451809-122451809 on Build GRCh38
rs55752906 Chr.4: 122451901-122451901 on Build GRCh38
rs572734070 Chr.4: 122452100-122452101 on Build GRCh38
rs2069771 Chr.4: 122452137-122452137 on Build GRCh38
rs924119601 Chr.4: 122452003-122452003 on Build GRCh38
rs1054037140 Chr.4: 122452075-122452075 on Build GRCh38
rs917096324 Chr.4: 122452123-122452123 on Build GRCh38
rs554577223 Chr.4: 122451940-122451940 on Build GRCh38
rs769929741 Chr.4: 122452050-122452050 on Build GRCh38
rs951157630 Chr.4: 122452041-122452041 on Build GRCh38
rs777464920 Chr.4: 122451810-122451810 on Build GRCh38
rs2069772 Chr.4: 122451978-122451978 on Build GRCh38
rs1042199702 Chr.4: 122452179-122452179 on Build GRCh38
rs936857617 Chr.4: 122452004-122452004 on Build GRCh38
rs911561997 Chr.4: 122451765-122451765 on Build GRCh38
rs745754932 Chr.4: 122451865-122451865 on Build GRCh38
rs778851120 Chr.4: 122451852-122451852 on Build GRCh38
rs917958922 Chr.4: 122451950-122451950 on Build GRCh38
rs377374894 Chr.4: 122451829-122451829 on Build GRCh38
rs34501152 Chr.4: 122452025-122452025 on Build GRCh38
rs760459373 Chr.4: 122451887-122451887 on Build GRCh38
rs984054148 Chr.4: 122452045-122452045 on Build GRCh38
rs552151597 Chr.4: 122451918-122451918 on Build GRCh38
rs768819078 Chr.4: 122451895-122451895 on Build GRCh38
rs771869973 Chr.4: 122451868-122451868 on Build GRCh38
rs41304870 Chr.4: 122451929-122451929 on Build GRCh38
rs750932763 Chr.4: 122451744-122451744 on Build GRCh38
rs1055149665 Chr.4: 122451862-122451862 on Build GRCh38
rs903182581 Chr.4: 122451788-122451788 on Build GRCh38
rs2069773 Chr.4: 122451878-122451878 on Build GRCh38
rs1051753 Chr.4: 122451758-122451758 on Build GRCh38
rs879349345 Chr.4: 122452034-122452034 on Build GRCh38
rs997290275 Chr.4: 122452177-122452177 on Build GRCh38
rs543296992 Chr.4: 122451796-122451796 on Build GRCh38
rs958520735 Chr.4: 122452088-122452088 on Build GRCh38
rs758867724 Chr.4: 122451748-122451748 on Build GRCh38
rs770442564 Chr.4: 122451833-122451833 on Build GRCh38
rs548350343 Chr.4: 122452101-122452101 on Build GRCh38
rs781412836 Chr.4: 122451928-122451928 on Build GRCh38
rs56247996 Chr.4: 122451977-122451977 on Build GRCh38
rs865810716 Chr.4: 122451819-122451819 on Build GRCh38
rs1000418138 Chr.4: 122451843-122451843 on Build GRCh38
rs998347437 Chr.4: 122451965-122451965 on Build GRCh38
rs374465594 Chr.4: 122451789-122451789 on Build GRCh38

View Full Product Details