Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63572299-63572800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A1
Entrez Gene ID
7803
Gene Name
protein tyrosine phosphatase type IVA, member 1
Gene Aliases
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
UniGene
Hs.227777
Gene Chromosome Location
Chr.6: 63528021-63583588 on Build GRCh38
Transcripts
NM_003463.4 XM_011536112.1 XM_017011270.1 XM_017011271.1 XM_017011272.1

SNP Detail

SNP ID Chromosome Location
rs893061627 Chr.6: 63572439-63572439 on Build GRCh38
rs964912160 Chr.6: 63572733-63572733 on Build GRCh38
rs899551526 Chr.6: 63572355-63572355 on Build GRCh38
rs570981614 Chr.6: 63572501-63572501 on Build GRCh38
rs1050896905 Chr.6: 63572527-63572527 on Build GRCh38
rs939767081 Chr.6: 63572767-63572767 on Build GRCh38
rs946345798 Chr.6: 63572336-63572336 on Build GRCh38
rs1039388469 Chr.6: 63572353-63572353 on Build GRCh38
rs547056272 Chr.6: 63572537-63572537 on Build GRCh38
rs528563105 Chr.6: 63572522-63572522 on Build GRCh38
rs1029889563 Chr.6: 63572364-63572364 on Build GRCh38
rs1016037515 Chr.6: 63572750-63572750 on Build GRCh38
rs1010933126 Chr.6: 63572600-63572625 on Build GRCh38
rs997974980 Chr.6: 63572362-63572362 on Build GRCh38
rs539979797 Chr.6: 63572728-63572728 on Build GRCh38
rs530290302 Chr.6: 63572447-63572447 on Build GRCh38
rs962624368 Chr.6: 63572406-63572406 on Build GRCh38
rs894181391 Chr.6: 63572738-63572738 on Build GRCh38
rs886977536 Chr.6: 63572374-63572374 on Build GRCh38
rs868699109 Chr.6: 63572596-63572596 on Build GRCh38
rs971070460 Chr.6: 63572447-63572447 on Build GRCh38
rs1006383643 Chr.6: 63572739-63572739 on Build GRCh38
rs572488466 Chr.6: 63572717-63572717 on Build GRCh38
rs752370818 Chr.6: 63572507-63572507 on Build GRCh38
rs760839669 Chr.6: 63572335-63572335 on Build GRCh38
rs556984320 Chr.6: 63572631-63572631 on Build GRCh38
rs1012383829 Chr.6: 63572559-63572560 on Build GRCh38
rs1054696712 Chr.6: 63572520-63572520 on Build GRCh38
rs1022980803 Chr.6: 63572433-63572433 on Build GRCh38
rs908269691 Chr.6: 63572761-63572761 on Build GRCh38
rs918294365 Chr.6: 63572373-63572373 on Build GRCh38
rs563965951 Chr.6: 63572460-63572460 on Build GRCh38
rs557295555 Chr.6: 63572674-63572674 on Build GRCh38
rs962951011 Chr.6: 63572577-63572577 on Build GRCh38
rs991019618 Chr.6: 63572347-63572347 on Build GRCh38
rs548645381 Chr.6: 63572459-63572459 on Build GRCh38
rs981139514 Chr.6: 63572467-63572467 on Build GRCh38
rs1041659350 Chr.6: 63572527-63572527 on Build GRCh38
rs911269574 Chr.6: 63572601-63572601 on Build GRCh38
rs2758257 Chr.6: 63572523-63572523 on Build GRCh38
rs559729253 Chr.6: 63572426-63572426 on Build GRCh38
rs1010561642 Chr.6: 63572524-63572524 on Build GRCh38
rs973374969 Chr.6: 63572582-63572582 on Build GRCh38
rs568763934 Chr.6: 63572647-63572647 on Build GRCh38
rs1038200215 Chr.6: 63572768-63572768 on Build GRCh38
rs933730330 Chr.6: 63572512-63572512 on Build GRCh38
rs910098165 Chr.6: 63572484-63572484 on Build GRCh38
rs892038961 Chr.6: 63572573-63572590 on Build GRCh38
rs914770746 Chr.6: 63572695-63572695 on Build GRCh38
rs959503123 Chr.6: 63572475-63572475 on Build GRCh38
rs992581178 Chr.6: 63572361-63572361 on Build GRCh38
rs1031722268 Chr.6: 63572627-63572627 on Build GRCh38
rs943637330 Chr.6: 63572490-63572490 on Build GRCh38
rs982209945 Chr.6: 63572339-63572339 on Build GRCh38
rs1004122928 Chr.6: 63572379-63572379 on Build GRCh38
rs530940270 Chr.6: 63572473-63572473 on Build GRCh38
rs1016824504 Chr.6: 63572400-63572400 on Build GRCh38
rs1012866929 Chr.6: 63572581-63572581 on Build GRCh38
rs201190738 Chr.6: 63572646-63572646 on Build GRCh38
rs924772430 Chr.6: 63572465-63572465 on Build GRCh38
rs971462394 Chr.6: 63572595-63572595 on Build GRCh38
rs990247276 Chr.6: 63572650-63572650 on Build GRCh38
rs539066760 Chr.6: 63572649-63572649 on Build GRCh38
rs1054675505 Chr.6: 63572330-63572330 on Build GRCh38
rs1012900614 Chr.6: 63572416-63572416 on Build GRCh38
rs1037444991 Chr.6: 63572382-63572382 on Build GRCh38
rs914801756 Chr.6: 63572482-63572482 on Build GRCh38
rs918578456 Chr.6: 63572502-63572502 on Build GRCh38
rs934056809 Chr.6: 63572403-63572403 on Build GRCh38
rs11547431 Chr.6: 63572642-63572642 on Build GRCh38
rs971876921 Chr.6: 63572437-63572437 on Build GRCh38
rs552560956 Chr.6: 63572474-63572474 on Build GRCh38
rs955327684 Chr.6: 63572589-63572589 on Build GRCh38
rs2734852 Chr.6: 63572411-63572411 on Build GRCh38
rs938206162 Chr.6: 63572346-63572346 on Build GRCh38
rs1041997089 Chr.6: 63572638-63572638 on Build GRCh38
rs535635354 Chr.6: 63572545-63572545 on Build GRCh38
rs941349940 Chr.6: 63572361-63572362 on Build GRCh38
rs1003362269 Chr.6: 63572615-63572615 on Build GRCh38
rs1020377388 Chr.6: 63572441-63572441 on Build GRCh38
rs1038229503 Chr.6: 63572543-63572543 on Build GRCh38
rs942486267 Chr.6: 63572517-63572517 on Build GRCh38
rs1006754729 Chr.6: 63572421-63572421 on Build GRCh38
rs764702798 Chr.6: 63572755-63572755 on Build GRCh38
rs928135057 Chr.6: 63572344-63572344 on Build GRCh38
rs942786234 Chr.6: 63572603-63572603 on Build GRCh38
rs1020008147 Chr.6: 63572327-63572327 on Build GRCh38
rs765095848 Chr.6: 63572407-63572407 on Build GRCh38
rs555434646 Chr.6: 63572743-63572743 on Build GRCh38
rs907148810 Chr.6: 63572676-63572676 on Build GRCh38
rs963187761 Chr.6: 63572763-63572763 on Build GRCh38
rs981573213 Chr.6: 63572440-63572440 on Build GRCh38
rs1004011585 Chr.6: 63572542-63572542 on Build GRCh38
rs920918708 Chr.6: 63572747-63572747 on Build GRCh38
rs1023369609 Chr.6: 63572587-63572587 on Build GRCh38
rs957046345 Chr.6: 63572323-63572323 on Build GRCh38
rs559134641 Chr.6: 63572539-63572539 on Build GRCh38
rs956243110 Chr.6: 63572470-63572470 on Build GRCh38
rs889885573 Chr.6: 63572514-63572514 on Build GRCh38
rs750397156 Chr.6: 63572756-63572756 on Build GRCh38
rs1054579579 Chr.6: 63572612-63572612 on Build GRCh38
rs1051219056 Chr.6: 63572413-63572413 on Build GRCh38
rs941617683 Chr.6: 63572495-63572495 on Build GRCh38
rs889604376 Chr.6: 63572420-63572420 on Build GRCh38

View Full Product Details