Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63581895-63582396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A1
Entrez Gene ID
7803
Gene Name
protein tyrosine phosphatase type IVA, member 1
Gene Aliases
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
UniGene
Hs.227777
Gene Chromosome Location
Chr.6: 63528021-63583588 on Build GRCh38
Transcripts
NM_003463.4 XM_011536111.1 XM_011536112.1 XM_017011270.1 XM_017011271.1

SNP Detail

SNP ID Chromosome Location
rs377511267 Chr.6: 63582348-63582348 on Build GRCh38
rs917628598 Chr.6: 63582261-63582261 on Build GRCh38
rs1052150297 Chr.6: 63582050-63582050 on Build GRCh38
rs953441387 Chr.6: 63582275-63582275 on Build GRCh38
rs905600159 Chr.6: 63582122-63582122 on Build GRCh38
rs369141968 Chr.6: 63582187-63582187 on Build GRCh38
rs901916695 Chr.6: 63582152-63582152 on Build GRCh38
rs544280994 Chr.6: 63581985-63581985 on Build GRCh38
rs865990966 Chr.6: 63582343-63582343 on Build GRCh38
rs1062193 Chr.6: 63582248-63582248 on Build GRCh38
rs144571773 Chr.6: 63581936-63581936 on Build GRCh38
rs979291546 Chr.6: 63582196-63582196 on Build GRCh38
rs575033403 Chr.6: 63581989-63581989 on Build GRCh38
rs930950145 Chr.6: 63582075-63582075 on Build GRCh38
rs935008153 Chr.6: 63582049-63582049 on Build GRCh38
rs953471896 Chr.6: 63581954-63581954 on Build GRCh38
rs888034721 Chr.6: 63582304-63582305 on Build GRCh38
rs1014976785 Chr.6: 63582134-63582134 on Build GRCh38
rs763561170 Chr.6: 63582271-63582272 on Build GRCh38
rs185421858 Chr.6: 63582137-63582137 on Build GRCh38
rs777648977 Chr.6: 63582185-63582185 on Build GRCh38
rs191348222 Chr.6: 63582056-63582056 on Build GRCh38
rs1029048282 Chr.6: 63582358-63582358 on Build GRCh38
rs869233616 Chr.6: 63582227-63582228 on Build GRCh38
rs370018421 Chr.6: 63582308-63582308 on Build GRCh38
rs975896808 Chr.6: 63581947-63581947 on Build GRCh38
rs901868935 Chr.6: 63582339-63582339 on Build GRCh38
rs760012553 Chr.6: 63582060-63582062 on Build GRCh38
rs917664095 Chr.6: 63582062-63582062 on Build GRCh38
rs1036771033 Chr.6: 63582327-63582327 on Build GRCh38
rs1032675771 Chr.6: 63582202-63582202 on Build GRCh38
rs561884512 Chr.6: 63582192-63582193 on Build GRCh38
rs563779284 Chr.6: 63582356-63582358 on Build GRCh38
rs927415510 Chr.6: 63582281-63582281 on Build GRCh38
rs997598121 Chr.6: 63582162-63582162 on Build GRCh38
rs940934433 Chr.6: 63582039-63582039 on Build GRCh38
rs1020177948 Chr.6: 63582241-63582241 on Build GRCh38
rs1019457111 Chr.6: 63582297-63582297 on Build GRCh38
rs988202142 Chr.6: 63582251-63582251 on Build GRCh38
rs560149022 Chr.6: 63582155-63582155 on Build GRCh38
rs1000185087 Chr.6: 63582266-63582266 on Build GRCh38
rs921983051 Chr.6: 63581925-63581925 on Build GRCh38
rs953243740 Chr.6: 63582194-63582194 on Build GRCh38
rs965867728 Chr.6: 63581946-63581947 on Build GRCh38
rs1036575192 Chr.6: 63582067-63582067 on Build GRCh38
rs953295725 Chr.6: 63581965-63581970 on Build GRCh38
rs979343828 Chr.6: 63582010-63582010 on Build GRCh38
rs145585575 Chr.6: 63582304-63582307 on Build GRCh38
rs937512306 Chr.6: 63582282-63582283 on Build GRCh38
rs756944897 Chr.6: 63582274-63582274 on Build GRCh38
rs186956654 Chr.6: 63581960-63581960 on Build GRCh38
rs181201842 Chr.6: 63582106-63582106 on Build GRCh38
rs987590011 Chr.6: 63582296-63582296 on Build GRCh38
rs1001278203 Chr.6: 63582130-63582130 on Build GRCh38
rs920861477 Chr.6: 63582072-63582072 on Build GRCh38
rs567385578 Chr.6: 63582342-63582342 on Build GRCh38
rs962210602 Chr.6: 63582340-63582340 on Build GRCh38
rs906054274 Chr.6: 63582084-63582084 on Build GRCh38
rs1056886237 Chr.6: 63582132-63582132 on Build GRCh38
rs146224762 Chr.6: 63582225-63582226 on Build GRCh38
rs924961611 Chr.6: 63582045-63582045 on Build GRCh38
rs937562993 Chr.6: 63582089-63582090 on Build GRCh38
rs778219091 Chr.6: 63582102-63582103 on Build GRCh38

View Full Product Details