Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156815660-156816159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTRK1
Entrez Gene ID
4914
Gene Name
neurotrophic receptor tyrosine kinase 1
Gene Aliases
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA
UniGene
Hs.406293
Gene Chromosome Location
Chr.1: 156815750-156881850 on Build GRCh38
Transcripts
NM_001007792.1 NM_001161441.1 NM_001161442.1 NM_001161443.1 NM_001161444.1 NM_003975.3 XM_006711615.2 XM_017002762.1 XM_017002763.1 XM_017002764.1 XM_017002765.1 XM_017002766.1 XM_017002767.1

Gene Symbol
SH2D2A
Entrez Gene ID
9047
Gene Name
SH2 domain containing 2A
Gene Aliases
F2771, SCAP, TSAD, VRAP
UniGene
Hs.103527
Gene Chromosome Location
Chr.1: 156806238-156816853 on Build GRCh38
Transcripts
NM_001007792.1 NM_001161441.1 NM_001161442.1 NM_001161443.1 NM_001161444.1 NM_003975.3 XM_006711615.2 XM_017002762.1 XM_017002763.1 XM_017002764.1 XM_017002765.1 XM_017002766.1 XM_017002767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3965886 c.86T>C p.M29T Chr.1: 156816043-156816043 on Build GRCh38
1984337 c.68C>T p.T23I Chr.1: 156816061-156816061 on Build GRCh38
1151866 c.42C>T p.H14H Chr.1: 156816087-156816087 on Build GRCh38
1984336 c.68C>T p.T23I Chr.1: 156816061-156816061 on Build GRCh38
1225547 c.61T>G p.F21V Chr.1: 156816068-156816068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564227571 Chr.1: 156815891-156815891 on Build GRCh38
rs200011053 Chr.1: 156815752-156815752 on Build GRCh38
rs903106341 Chr.1: 156815743-156815743 on Build GRCh38
rs771165291 Chr.1: 156815974-156815974 on Build GRCh38
rs759998500 Chr.1: 156815978-156815978 on Build GRCh38
rs892921029 Chr.1: 156816110-156816110 on Build GRCh38
rs751380885 Chr.1: 156815856-156815857 on Build GRCh38
rs766829057 Chr.1: 156815808-156815808 on Build GRCh38
rs1006353272 Chr.1: 156815765-156815765 on Build GRCh38
rs755696857 Chr.1: 156815847-156815847 on Build GRCh38
rs765107804 Chr.1: 156815994-156815994 on Build GRCh38
rs753901156 Chr.1: 156816115-156816115 on Build GRCh38
rs938603449 Chr.1: 156815914-156815914 on Build GRCh38
rs779097484 Chr.1: 156815725-156815725 on Build GRCh38
rs201892904 Chr.1: 156816101-156816101 on Build GRCh38
rs780133657 Chr.1: 156815779-156815779 on Build GRCh38
rs771167339 Chr.1: 156815879-156815879 on Build GRCh38
rs767306706 Chr.1: 156815827-156815827 on Build GRCh38
rs1031016446 Chr.1: 156816044-156816044 on Build GRCh38
rs938307871 Chr.1: 156815744-156815744 on Build GRCh38
rs898019076 Chr.1: 156816034-156816034 on Build GRCh38
rs775270596 Chr.1: 156816031-156816031 on Build GRCh38
rs997762437 Chr.1: 156816035-156816035 on Build GRCh38
rs1020105887 Chr.1: 156816125-156816125 on Build GRCh38
rs369809596 Chr.1: 156816119-156816119 on Build GRCh38
rs1015554660 Chr.1: 156815766-156815766 on Build GRCh38
rs758191370 Chr.1: 156816006-156816006 on Build GRCh38
rs779560691 Chr.1: 156816118-156816118 on Build GRCh38
rs575068011 Chr.1: 156815760-156815760 on Build GRCh38
rs372843554 Chr.1: 156815933-156815933 on Build GRCh38
rs1056796351 Chr.1: 156815745-156815745 on Build GRCh38
rs542067267 Chr.1: 156815789-156815789 on Build GRCh38
rs114004093 Chr.1: 156815954-156815954 on Build GRCh38
rs146503193 Chr.1: 156816045-156816045 on Build GRCh38
rs752881064 Chr.1: 156816070-156816070 on Build GRCh38
rs1045378275 Chr.1: 156815911-156815911 on Build GRCh38
rs372317622 Chr.1: 156815878-156815878 on Build GRCh38
rs761241472 Chr.1: 156816041-156816041 on Build GRCh38
rs377668169 Chr.1: 156816037-156816037 on Build GRCh38
rs563084655 Chr.1: 156815747-156815747 on Build GRCh38
rs769930586 Chr.1: 156815960-156815960 on Build GRCh38
rs369870153 Chr.1: 156816038-156816038 on Build GRCh38
rs374760070 Chr.1: 156815781-156815781 on Build GRCh38
rs777944501 Chr.1: 156816016-156816016 on Build GRCh38
rs200656218 Chr.1: 156815854-156815854 on Build GRCh38
rs762359647 Chr.1: 156816117-156816117 on Build GRCh38
rs761047833 Chr.1: 156815970-156815971 on Build GRCh38
rs377396834 Chr.1: 156816120-156816120 on Build GRCh38
rs149790796 Chr.1: 156815986-156815986 on Build GRCh38
rs756324388 Chr.1: 156816086-156816086 on Build GRCh38
rs776157082 Chr.1: 156816040-156816040 on Build GRCh38
rs374374158 Chr.1: 156816128-156816128 on Build GRCh38
rs373057791 Chr.1: 156816030-156816030 on Build GRCh38
rs774733806 Chr.1: 156815975-156815975 on Build GRCh38
rs1800600 Chr.1: 156815987-156815987 on Build GRCh38
rs1015088087 Chr.1: 156816033-156816033 on Build GRCh38
rs986244724 Chr.1: 156815707-156815707 on Build GRCh38
rs750159727 Chr.1: 156816003-156816003 on Build GRCh38
rs368557134 Chr.1: 156815874-156815874 on Build GRCh38
rs756122528 Chr.1: 156816013-156816013 on Build GRCh38
rs375140058 Chr.1: 156816017-156816017 on Build GRCh38
rs369302487 Chr.1: 156815890-156815890 on Build GRCh38
rs377378555 Chr.1: 156816020-156816020 on Build GRCh38
rs146718843 Chr.1: 156816008-156816008 on Build GRCh38
rs374636761 Chr.1: 156816069-156816069 on Build GRCh38
rs775284911 Chr.1: 156815980-156815980 on Build GRCh38
rs949631277 Chr.1: 156816046-156816046 on Build GRCh38
rs763226014 Chr.1: 156815807-156815807 on Build GRCh38
rs867004596 Chr.1: 156815970-156815971 on Build GRCh38
rs914387978 Chr.1: 156815720-156815720 on Build GRCh38
rs139790457 Chr.1: 156815941-156815941 on Build GRCh38
rs371878969 Chr.1: 156815856-156815856 on Build GRCh38
rs369490353 Chr.1: 156816028-156816028 on Build GRCh38
rs367808960 Chr.1: 156815795-156815795 on Build GRCh38
rs11325907 Chr.1: 156815971-156815971 on Build GRCh38
rs760259992 Chr.1: 156815981-156815981 on Build GRCh38
rs986136491 Chr.1: 156816061-156816061 on Build GRCh38
rs374331077 Chr.1: 156815870-156815870 on Build GRCh38
rs762721468 Chr.1: 156816057-156816057 on Build GRCh38
rs531638430 Chr.1: 156815907-156815907 on Build GRCh38
rs774736434 Chr.1: 156816021-156816021 on Build GRCh38
rs1800601 Chr.1: 156815825-156815825 on Build GRCh38
rs766336211 Chr.1: 156815820-156815820 on Build GRCh38
rs894165799 Chr.1: 156815746-156815746 on Build GRCh38
rs7546838 Chr.1: 156815979-156815979 on Build GRCh38
rs529950735 Chr.1: 156815973-156815973 on Build GRCh38
rs1041993978 Chr.1: 156815733-156815733 on Build GRCh38
rs146775862 Chr.1: 156815753-156815753 on Build GRCh38
rs765419185 Chr.1: 156815755-156815755 on Build GRCh38
rs145574758 Chr.1: 156816018-156816018 on Build GRCh38
rs781385955 Chr.1: 156816007-156816007 on Build GRCh38
rs527803577 Chr.1: 156815853-156815853 on Build GRCh38
rs770230661 Chr.1: 156815793-156815793 on Build GRCh38
rs750425454 Chr.1: 156816051-156816051 on Build GRCh38
rs374934496 Chr.1: 156815735-156815735 on Build GRCh38
rs755294320 Chr.1: 156815855-156815856 on Build GRCh38
rs200646544 Chr.1: 156815821-156815821 on Build GRCh38

View Full Product Details