Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55152309-55152802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884833 c.739G>T p.D247Y Chr.7: 55152656-55152656 on Build GRCh38
4884832 c.739G>T p.D247Y Chr.7: 55152656-55152656 on Build GRCh38
4384116 c.727C>T p.P243S Chr.7: 55152644-55152644 on Build GRCh38
5779666 c.654G>A p.Q218Q Chr.7: 55152571-55152571 on Build GRCh38
6008958 c.745C>G p.L249V Chr.7: 55152662-55152662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554981236 Chr.7: 55152647-55152647 on Build GRCh38
rs373970245 Chr.7: 55152550-55152550 on Build GRCh38
rs558961840 Chr.7: 55152766-55152766 on Build GRCh38
rs771165011 Chr.7: 55152542-55152542 on Build GRCh38
rs1028913009 Chr.7: 55152483-55152483 on Build GRCh38
rs752616839 Chr.7: 55152600-55152600 on Build GRCh38
rs540647354 Chr.7: 55152711-55152711 on Build GRCh38
rs376868785 Chr.7: 55152670-55152670 on Build GRCh38
rs779156097 Chr.7: 55152549-55152549 on Build GRCh38
rs751133675 Chr.7: 55152498-55152498 on Build GRCh38
rs987086326 Chr.7: 55152490-55152490 on Build GRCh38
rs917270624 Chr.7: 55152503-55152503 on Build GRCh38
rs780001754 Chr.7: 55152656-55152656 on Build GRCh38
rs559096241 Chr.7: 55152505-55152505 on Build GRCh38
rs1001067573 Chr.7: 55152485-55152485 on Build GRCh38
rs750423932 Chr.7: 55152551-55152551 on Build GRCh38
rs370236572 Chr.7: 55152672-55152672 on Build GRCh38
rs774889652 Chr.7: 55152686-55152686 on Build GRCh38
rs759615551 Chr.7: 55152502-55152502 on Build GRCh38
rs879289031 Chr.7: 55152747-55152747 on Build GRCh38
rs566948029 Chr.7: 55152394-55152394 on Build GRCh38
rs754581841 Chr.7: 55152500-55152500 on Build GRCh38
rs974327404 Chr.7: 55152750-55152750 on Build GRCh38
rs750033984 Chr.7: 55152396-55152396 on Build GRCh38
rs776886714 Chr.7: 55152578-55152578 on Build GRCh38
rs1048814560 Chr.7: 55152353-55152353 on Build GRCh38
rs1027951918 Chr.7: 55152692-55152692 on Build GRCh38
rs751295137 Chr.7: 55152582-55152582 on Build GRCh38
rs374561933 Chr.7: 55152643-55152643 on Build GRCh38
rs777575581 Chr.7: 55152609-55152609 on Build GRCh38
rs1040130291 Chr.7: 55152621-55152621 on Build GRCh38
rs749472815 Chr.7: 55152539-55152539 on Build GRCh38
rs115681740 Chr.7: 55152481-55152481 on Build GRCh38
rs546253199 Chr.7: 55152428-55152428 on Build GRCh38
rs577274312 Chr.7: 55152778-55152778 on Build GRCh38
rs908480400 Chr.7: 55152719-55152719 on Build GRCh38
rs45621033 Chr.7: 55152577-55152577 on Build GRCh38
rs761098433 Chr.7: 55152575-55152575 on Build GRCh38
rs940465245 Chr.7: 55152619-55152619 on Build GRCh38
rs17289830 Chr.7: 55152677-55152677 on Build GRCh38
rs886150691 Chr.7: 55152386-55152386 on Build GRCh38
rs763962907 Chr.7: 55152616-55152616 on Build GRCh38
rs200664836 Chr.7: 55152648-55152648 on Build GRCh38
rs776050604 Chr.7: 55152571-55152571 on Build GRCh38
rs766104143 Chr.7: 55152580-55152580 on Build GRCh38
rs955073578 Chr.7: 55152714-55152714 on Build GRCh38
rs528156232 Chr.7: 55152401-55152401 on Build GRCh38
rs1004572133 Chr.7: 55152395-55152395 on Build GRCh38
rs866386005 Chr.7: 55152456-55152456 on Build GRCh38
rs548566111 Chr.7: 55152334-55152334 on Build GRCh38
rs1046438724 Chr.7: 55152597-55152597 on Build GRCh38
rs756040606 Chr.7: 55152608-55152608 on Build GRCh38
rs777438007 Chr.7: 55152532-55152532 on Build GRCh38
rs370744986 Chr.7: 55152639-55152639 on Build GRCh38
rs571277669 Chr.7: 55152472-55152472 on Build GRCh38
rs752638444 Chr.7: 55152513-55152513 on Build GRCh38
rs766952405 Chr.7: 55152596-55152596 on Build GRCh38
rs971365814 Chr.7: 55152508-55152508 on Build GRCh38
rs17289823 Chr.7: 55152676-55152676 on Build GRCh38
rs761095472 Chr.7: 55152370-55152370 on Build GRCh38
rs765027291 Chr.7: 55152715-55152715 on Build GRCh38
rs773450086 Chr.7: 55152457-55152457 on Build GRCh38
rs775903941 Chr.7: 55152347-55152347 on Build GRCh38
rs774374179 Chr.7: 55152492-55152492 on Build GRCh38
rs767013310 Chr.7: 55152681-55152681 on Build GRCh38
rs759106015 Chr.7: 55152588-55152588 on Build GRCh38
rs765943494 Chr.7: 55152496-55152496 on Build GRCh38
rs763909508 Chr.7: 55152710-55152710 on Build GRCh38
rs370904966 Chr.7: 55152522-55152522 on Build GRCh38
rs903744972 Chr.7: 55152749-55152749 on Build GRCh38
rs765893589 Chr.7: 55152663-55152663 on Build GRCh38
rs770121025 Chr.7: 55152364-55152364 on Build GRCh38
rs767991661 Chr.7: 55152339-55152339 on Build GRCh38
rs760286038 Chr.7: 55152707-55152707 on Build GRCh38
rs145506643 Chr.7: 55152592-55152592 on Build GRCh38
rs770346907 Chr.7: 55152673-55152673 on Build GRCh38
rs930797191 Chr.7: 55152337-55152337 on Build GRCh38
rs11506105 Chr.7: 55152484-55152484 on Build GRCh38
rs772130986 Chr.7: 55152655-55152655 on Build GRCh38

View Full Product Details