Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 118926414-118926916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF11B
Entrez Gene ID
4982
Gene Name
TNF receptor superfamily member 11b
Gene Aliases
OCIF, OPG, PDB5, TR1
UniGene
Hs.81791
Gene Chromosome Location
Chr.8: 118923557-118952144 on Build GRCh38
Transcripts
NM_002546.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1454481 c.700G>A p.A234T Chr.8: 118926611-118926611 on Build GRCh38
1229873 c.679T>G p.L227V Chr.8: 118926632-118926632 on Build GRCh38
3644579 c.621C>T p.F207F Chr.8: 118926690-118926690 on Build GRCh38
6804980 c.682C>T p.P228S Chr.8: 118926629-118926629 on Build GRCh38
3644577 c.623G>A p.R208K Chr.8: 118926688-118926688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767927201 Chr.8: 118926591-118926591 on Build GRCh38
rs977797503 Chr.8: 118926680-118926680 on Build GRCh38
rs773538970 Chr.8: 118926664-118926664 on Build GRCh38
rs757066664 Chr.8: 118926807-118926807 on Build GRCh38
rs201393730 Chr.8: 118926611-118926611 on Build GRCh38
rs547836403 Chr.8: 118926642-118926642 on Build GRCh38
rs751454703 Chr.8: 118926442-118926442 on Build GRCh38
rs770777429 Chr.8: 118926536-118926536 on Build GRCh38
rs776225644 Chr.8: 118926542-118926542 on Build GRCh38
rs530867123 Chr.8: 118926649-118926649 on Build GRCh38
rs757704068 Chr.8: 118926466-118926466 on Build GRCh38
rs747711082 Chr.8: 118926519-118926519 on Build GRCh38
rs763182383 Chr.8: 118926451-118926451 on Build GRCh38
rs901358389 Chr.8: 118926870-118926870 on Build GRCh38
rs933035461 Chr.8: 118926879-118926879 on Build GRCh38
rs199810725 Chr.8: 118926562-118926562 on Build GRCh38
rs897164960 Chr.8: 118926873-118926873 on Build GRCh38
rs1131380 Chr.8: 118926523-118926523 on Build GRCh38
rs768436617 Chr.8: 118926758-118926758 on Build GRCh38
rs367991519 Chr.8: 118926465-118926465 on Build GRCh38
rs767035135 Chr.8: 118926602-118926602 on Build GRCh38
rs1052175886 Chr.8: 118926445-118926445 on Build GRCh38
rs1043311885 Chr.8: 118926783-118926783 on Build GRCh38
rs7844539 Chr.8: 118926486-118926486 on Build GRCh38
rs766836970 Chr.8: 118926688-118926688 on Build GRCh38
rs754392133 Chr.8: 118926603-118926603 on Build GRCh38
rs1033591517 Chr.8: 118926606-118926606 on Build GRCh38
rs374594601 Chr.8: 118926582-118926582 on Build GRCh38
rs755957559 Chr.8: 118926735-118926735 on Build GRCh38
rs749228162 Chr.8: 118926751-118926751 on Build GRCh38
rs934906746 Chr.8: 118926811-118926811 on Build GRCh38
rs780028766 Chr.8: 118926627-118926627 on Build GRCh38
rs756823120 Chr.8: 118926617-118926617 on Build GRCh38
rs936839707 Chr.8: 118926711-118926711 on Build GRCh38
rs750987558 Chr.8: 118926708-118926708 on Build GRCh38
rs11573929 Chr.8: 118926835-118926835 on Build GRCh38
rs763057283 Chr.8: 118926579-118926579 on Build GRCh38
rs926232843 Chr.8: 118926810-118926810 on Build GRCh38
rs150457771 Chr.8: 118926612-118926612 on Build GRCh38
rs61761330 Chr.8: 118926528-118926528 on Build GRCh38
rs750069796 Chr.8: 118926734-118926734 on Build GRCh38
rs11573930 Chr.8: 118926597-118926597 on Build GRCh38
rs774691067 Chr.8: 118926587-118926587 on Build GRCh38
rs761190753 Chr.8: 118926671-118926671 on Build GRCh38
rs753695242 Chr.8: 118926789-118926789 on Build GRCh38
rs546127798 Chr.8: 118926657-118926657 on Build GRCh38
rs767759522 Chr.8: 118926468-118926468 on Build GRCh38
rs936869456 Chr.8: 118926444-118926444 on Build GRCh38
rs566199412 Chr.8: 118926746-118926746 on Build GRCh38
rs769545395 Chr.8: 118926621-118926621 on Build GRCh38
rs567977879 Chr.8: 118926728-118926728 on Build GRCh38
rs528308716 Chr.8: 118926713-118926713 on Build GRCh38
rs759897062 Chr.8: 118926692-118926692 on Build GRCh38
rs371614996 Chr.8: 118926663-118926663 on Build GRCh38
rs761883178 Chr.8: 118926589-118926589 on Build GRCh38
rs760714285 Chr.8: 118926636-118926636 on Build GRCh38
rs764345890 Chr.8: 118926459-118926459 on Build GRCh38
rs757952532 Chr.8: 118926521-118926521 on Build GRCh38
rs781066936 Chr.8: 118926619-118926619 on Build GRCh38
rs755120990 Chr.8: 118926822-118926822 on Build GRCh38
rs2228568 Chr.8: 118926543-118926543 on Build GRCh38
rs369130157 Chr.8: 118926729-118926729 on Build GRCh38
rs756526392 Chr.8: 118926598-118926598 on Build GRCh38
rs915869445 Chr.8: 118926712-118926712 on Build GRCh38
rs756620500 Chr.8: 118926498-118926498 on Build GRCh38
rs376708982 Chr.8: 118926690-118926690 on Build GRCh38
rs779765236 Chr.8: 118926742-118926742 on Build GRCh38
rs763694902 Chr.8: 118926704-118926704 on Build GRCh38
rs368397401 Chr.8: 118926545-118926545 on Build GRCh38
rs558927039 Chr.8: 118926499-118926499 on Build GRCh38

View Full Product Details