Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47637953-47638446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4 XM_017029339.1

SNP Detail

SNP ID Chromosome Location
rs1024226320 Chr.X: 47638413-47638413 on Build GRCh38
rs770479468 Chr.X: 47638145-47638145 on Build GRCh38
rs376822462 Chr.X: 47638112-47638112 on Build GRCh38
rs773489927 Chr.X: 47637990-47637990 on Build GRCh38
rs2229088 Chr.X: 47638165-47638165 on Build GRCh38
rs781594313 Chr.X: 47638323-47638323 on Build GRCh38
rs760441334 Chr.X: 47638220-47638220 on Build GRCh38
rs751772833 Chr.X: 47638001-47638001 on Build GRCh38
rs752423073 Chr.X: 47638027-47638027 on Build GRCh38
rs371259584 Chr.X: 47638113-47638113 on Build GRCh38
rs774244991 Chr.X: 47638167-47638167 on Build GRCh38
rs994111784 Chr.X: 47638325-47638325 on Build GRCh38
rs200141730 Chr.X: 47638124-47638124 on Build GRCh38
rs1012252639 Chr.X: 47638412-47638412 on Build GRCh38
rs564143194 Chr.X: 47638407-47638407 on Build GRCh38
rs763164568 Chr.X: 47638159-47638159 on Build GRCh38
rs765768273 Chr.X: 47637975-47637975 on Build GRCh38
rs944642204 Chr.X: 47638259-47638259 on Build GRCh38
rs762419349 Chr.X: 47637973-47637973 on Build GRCh38
rs201698802 Chr.X: 47638057-47638057 on Build GRCh38
rs755114172 Chr.X: 47638007-47638007 on Build GRCh38
rs200172068 Chr.X: 47638009-47638009 on Build GRCh38
rs778250617 Chr.X: 47638110-47638110 on Build GRCh38
rs774412399 Chr.X: 47638338-47638338 on Build GRCh38
rs762117199 Chr.X: 47638386-47638386 on Build GRCh38
rs187486290 Chr.X: 47638168-47638168 on Build GRCh38
rs756731775 Chr.X: 47638102-47638102 on Build GRCh38
rs897178212 Chr.X: 47638266-47638266 on Build GRCh38
rs766556938 Chr.X: 47637995-47637995 on Build GRCh38
rs376525319 Chr.X: 47637993-47637993 on Build GRCh38
rs890258474 Chr.X: 47638258-47638258 on Build GRCh38
rs376202455 Chr.X: 47638348-47638348 on Build GRCh38
rs773831559 Chr.X: 47638153-47638153 on Build GRCh38
rs1036254890 Chr.X: 47638260-47638260 on Build GRCh38
rs766899855 Chr.X: 47638123-47638123 on Build GRCh38
rs756113016 Chr.X: 47638053-47638053 on Build GRCh38
rs888523184 Chr.X: 47638339-47638339 on Build GRCh38
rs752547814 Chr.X: 47638197-47638197 on Build GRCh38
rs767628955 Chr.X: 47638177-47638177 on Build GRCh38
rs373267605 Chr.X: 47637974-47637974 on Build GRCh38

View Full Product Details