Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64269749-64270257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAD
Entrez Gene ID
572
Gene Name
BCL2 associated agonist of cell death
Gene Aliases
BBC2, BCL2L8
UniGene
Hs.370254
Gene Chromosome Location
Chr.11: 64269828-64284704 on Build GRCh38
Transcripts
NM_004322.3 NM_032989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4656837 c.481A>G p.R161G Chr.11: 64270235-64270235 on Build GRCh38
4035084 c.499T>C p.S167P Chr.11: 64270217-64270217 on Build GRCh38
6483750 c.496C>T p.P166S Chr.11: 64270220-64270220 on Build GRCh38
4035085 c.488G>A p.S163N Chr.11: 64270228-64270228 on Build GRCh38
2163284 c.483G>T p.R161S Chr.11: 64270233-64270233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762874651 Chr.11: 64270125-64270125 on Build GRCh38
rs971837272 Chr.11: 64269818-64269818 on Build GRCh38
rs777377835 Chr.11: 64270173-64270173 on Build GRCh38
rs758348141 Chr.11: 64270105-64270105 on Build GRCh38
rs963285257 Chr.11: 64270090-64270090 on Build GRCh38
rs922975141 Chr.11: 64269832-64269832 on Build GRCh38
rs576392138 Chr.11: 64269918-64269918 on Build GRCh38
rs970587335 Chr.11: 64269924-64269924 on Build GRCh38
rs769286107 Chr.11: 64270147-64270147 on Build GRCh38
rs1054745529 Chr.11: 64269875-64269875 on Build GRCh38
rs572091521 Chr.11: 64270035-64270035 on Build GRCh38
rs779622078 Chr.11: 64269834-64269834 on Build GRCh38
rs10556 Chr.11: 64269935-64269935 on Build GRCh38
rs779785883 Chr.11: 64269954-64269954 on Build GRCh38
rs796510497 Chr.11: 64269949-64269949 on Build GRCh38
rs144873026 Chr.11: 64270130-64270130 on Build GRCh38
rs892170792 Chr.11: 64270022-64270022 on Build GRCh38
rs892117142 Chr.11: 64270220-64270220 on Build GRCh38
rs190892320 Chr.11: 64269915-64269915 on Build GRCh38
rs911384299 Chr.11: 64270028-64270028 on Build GRCh38
rs565081290 Chr.11: 64269962-64269962 on Build GRCh38
rs552180755 Chr.11: 64270097-64270097 on Build GRCh38
rs759247628 Chr.11: 64270155-64270155 on Build GRCh38
rs984697573 Chr.11: 64269776-64269776 on Build GRCh38
rs922976481 Chr.11: 64269996-64269996 on Build GRCh38
rs372512554 Chr.11: 64270198-64270198 on Build GRCh38
rs749547055 Chr.11: 64270190-64270190 on Build GRCh38
rs745732316 Chr.11: 64270183-64270183 on Build GRCh38
rs765045279 Chr.11: 64270165-64270165 on Build GRCh38
rs908871391 Chr.11: 64269785-64269785 on Build GRCh38
rs972107393 Chr.11: 64269986-64269986 on Build GRCh38
rs574648805 Chr.11: 64269889-64269889 on Build GRCh38
rs751927323 Chr.11: 64269926-64269926 on Build GRCh38
rs907226582 Chr.11: 64270106-64270106 on Build GRCh38
rs763492858 Chr.11: 64270222-64270222 on Build GRCh38
rs773042731 Chr.11: 64270038-64270038 on Build GRCh38
rs1024752195 Chr.11: 64270032-64270032 on Build GRCh38
rs1032437055 Chr.11: 64269887-64269887 on Build GRCh38
rs186286987 Chr.11: 64270041-64270041 on Build GRCh38
rs527760717 Chr.11: 64269972-64269972 on Build GRCh38
rs765325772 Chr.11: 64269908-64269908 on Build GRCh38
rs745316042 Chr.11: 64269873-64269873 on Build GRCh38
rs764499854 Chr.11: 64269923-64269923 on Build GRCh38
rs1030396906 Chr.11: 64270203-64270203 on Build GRCh38
rs751423549 Chr.11: 64270175-64270175 on Build GRCh38
rs541265239 Chr.11: 64269984-64269984 on Build GRCh38
rs543721700 Chr.11: 64269891-64269891 on Build GRCh38
rs759787241 Chr.11: 64270048-64270048 on Build GRCh38
rs1054630287 Chr.11: 64270036-64270036 on Build GRCh38
rs943574245 Chr.11: 64270001-64270001 on Build GRCh38
rs753742453 Chr.11: 64270138-64270138 on Build GRCh38
rs770676243 Chr.11: 64270117-64270117 on Build GRCh38
rs766875705 Chr.11: 64270208-64270208 on Build GRCh38
rs374193764 Chr.11: 64270148-64270148 on Build GRCh38
rs750374497 Chr.11: 64270237-64270237 on Build GRCh38
rs541333380 Chr.11: 64270098-64270098 on Build GRCh38
rs780082386 Chr.11: 64270186-64270186 on Build GRCh38
rs776454584 Chr.11: 64270119-64270119 on Build GRCh38
rs1000873868 Chr.11: 64270016-64270016 on Build GRCh38
rs1017895674 Chr.11: 64269867-64269867 on Build GRCh38
rs950759190 Chr.11: 64270112-64270113 on Build GRCh38
rs752256324 Chr.11: 64270213-64270213 on Build GRCh38
rs940398418 Chr.11: 64269805-64269805 on Build GRCh38
rs765272640 Chr.11: 64270061-64270061 on Build GRCh38
rs1033946069 Chr.11: 64270080-64270080 on Build GRCh38
rs565814097 Chr.11: 64270099-64270099 on Build GRCh38
rs769570355 Chr.11: 64269894-64269894 on Build GRCh38
rs764576103 Chr.11: 64270086-64270086 on Build GRCh38
rs530207685 Chr.11: 64270018-64270018 on Build GRCh38
rs534841154 Chr.11: 64269863-64269863 on Build GRCh38
rs768575949 Chr.11: 64270191-64270191 on Build GRCh38
rs961938960 Chr.11: 64269970-64269970 on Build GRCh38
rs775239803 Chr.11: 64270100-64270100 on Build GRCh38
rs1016177531 Chr.11: 64269963-64269963 on Build GRCh38
rs772535203 Chr.11: 64269916-64269916 on Build GRCh38
rs1002976022 Chr.11: 64269938-64269938 on Build GRCh38
rs894312172 Chr.11: 64270142-64270142 on Build GRCh38
rs59283629 Chr.11: 64270207-64270207 on Build GRCh38
rs1050029 Chr.11: 64269939-64269939 on Build GRCh38
rs1051672377 Chr.11: 64269838-64269838 on Build GRCh38
rs754486155 Chr.11: 64270003-64270003 on Build GRCh38
rs1008219417 Chr.11: 64269845-64269845 on Build GRCh38
rs79723489 Chr.11: 64270230-64270230 on Build GRCh38
rs778541753 Chr.11: 64270101-64270101 on Build GRCh38
rs911415367 Chr.11: 64269852-64269852 on Build GRCh38
rs747666044 Chr.11: 64270102-64270102 on Build GRCh38
rs370353523 Chr.11: 64270184-64270184 on Build GRCh38
rs981076803 Chr.11: 64269930-64269930 on Build GRCh38
rs748571587 Chr.11: 64269865-64269865 on Build GRCh38
rs5792317 Chr.11: 64270049-64270050 on Build GRCh38
rs773702754 Chr.11: 64270204-64270204 on Build GRCh38
rs752110012 Chr.11: 64270093-64270093 on Build GRCh38
rs780953269 Chr.11: 64270180-64270180 on Build GRCh38
rs775620494 Chr.11: 64270227-64270227 on Build GRCh38
rs762411683 Chr.11: 64270143-64270143 on Build GRCh38
rs534722899 Chr.11: 64270120-64270120 on Build GRCh38
rs572443195 Chr.11: 64269993-64269993 on Build GRCh38
rs746993262 Chr.11: 64270109-64270109 on Build GRCh38
rs1053414502 Chr.11: 64270019-64270019 on Build GRCh38
rs774529140 Chr.11: 64270192-64270192 on Build GRCh38
rs890634575 Chr.11: 64269793-64269793 on Build GRCh38
rs553378700 Chr.11: 64269871-64269871 on Build GRCh38
rs3741400 Chr.11: 64269886-64269886 on Build GRCh38
rs532175074 Chr.11: 64270084-64270084 on Build GRCh38
rs73500186 Chr.11: 64270009-64270009 on Build GRCh38
rs958563024 Chr.11: 64269947-64269947 on Build GRCh38
rs752395879 Chr.11: 64270170-64270170 on Build GRCh38
rs775543241 Chr.11: 64270124-64270124 on Build GRCh38
rs550453760 Chr.11: 64270029-64270029 on Build GRCh38
rs199759687 Chr.11: 64270228-64270228 on Build GRCh38
rs1006089579 Chr.11: 64269953-64269953 on Build GRCh38
rs777517740 Chr.11: 64270107-64270107 on Build GRCh38
rs762709817 Chr.11: 64270218-64270218 on Build GRCh38
rs753165178 Chr.11: 64269911-64269911 on Build GRCh38
rs770500791 Chr.11: 64270044-64270044 on Build GRCh38
rs933093234 Chr.11: 64269835-64269835 on Build GRCh38
rs757801724 Chr.11: 64270172-64270172 on Build GRCh38
rs565771216 Chr.11: 64269787-64269787 on Build GRCh38
rs985926419 Chr.11: 64270017-64270017 on Build GRCh38
rs770061234 Chr.11: 64270122-64270122 on Build GRCh38
rs1024805422 Chr.11: 64269919-64269919 on Build GRCh38
rs775596557 Chr.11: 64270063-64270063 on Build GRCh38
rs568246472 Chr.11: 64270179-64270179 on Build GRCh38

View Full Product Details