Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35871715-35872259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL5
Entrez Gene ID
6352
Gene Name
C-C motif chemokine ligand 5
Gene Aliases
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
UniGene
Hs.514821
Gene Chromosome Location
Chr.17: 35871491-35880373 on Build GRCh38
Transcripts
NM_001278736.1 NM_002985.2

SNP Detail

SNP ID Chromosome Location
rs925028081 Chr.17: 35872121-35872121 on Build GRCh38
rs199967645 Chr.17: 35872047-35872047 on Build GRCh38
rs1006346649 Chr.17: 35872073-35872073 on Build GRCh38
rs931758421 Chr.17: 35871956-35871956 on Build GRCh38
rs867090903 Chr.17: 35872198-35872198 on Build GRCh38
rs910842914 Chr.17: 35871866-35871866 on Build GRCh38
rs1142895 Chr.17: 35872109-35872109 on Build GRCh38
rs867719219 Chr.17: 35872008-35872008 on Build GRCh38
rs887942487 Chr.17: 35872070-35872070 on Build GRCh38
rs551118547 Chr.17: 35872143-35872143 on Build GRCh38
rs541327993 Chr.17: 35871829-35871829 on Build GRCh38
rs776640911 Chr.17: 35872231-35872232 on Build GRCh38
rs561886350 Chr.17: 35871821-35871821 on Build GRCh38
rs964690127 Chr.17: 35871867-35871867 on Build GRCh38
rs867271934 Chr.17: 35872203-35872203 on Build GRCh38
rs564783592 Chr.17: 35871844-35871844 on Build GRCh38
rs372014447 Chr.17: 35871916-35871916 on Build GRCh38
rs1010663185 Chr.17: 35872108-35872108 on Build GRCh38
rs544588366 Chr.17: 35871914-35871916 on Build GRCh38
rs962014743 Chr.17: 35871884-35871884 on Build GRCh38
rs775374492 Chr.17: 35872227-35872227 on Build GRCh38
rs563578870 Chr.17: 35871967-35871967 on Build GRCh38
rs920329967 Chr.17: 35871950-35871950 on Build GRCh38
rs1010297635 Chr.17: 35871745-35871746 on Build GRCh38
rs960214132 Chr.17: 35872214-35872214 on Build GRCh38
rs528774647 Chr.17: 35871960-35871960 on Build GRCh38
rs534629656 Chr.17: 35871961-35871961 on Build GRCh38
rs948703249 Chr.17: 35872052-35872052 on Build GRCh38
rs968825843 Chr.17: 35871753-35871753 on Build GRCh38
rs1050232586 Chr.17: 35871991-35871991 on Build GRCh38
rs1045748930 Chr.17: 35872053-35872053 on Build GRCh38
rs1001601648 Chr.17: 35872163-35872163 on Build GRCh38
rs966805960 Chr.17: 35872110-35872110 on Build GRCh38
rs570894226 Chr.17: 35872186-35872186 on Build GRCh38
rs922983257 Chr.17: 35871898-35871898 on Build GRCh38
rs1029977397 Chr.17: 35872088-35872088 on Build GRCh38
rs528911373 Chr.17: 35872014-35872014 on Build GRCh38
rs904615193 Chr.17: 35872129-35872129 on Build GRCh38
rs1046774581 Chr.17: 35872199-35872199 on Build GRCh38
rs546847772 Chr.17: 35872055-35872055 on Build GRCh38
rs973142042 Chr.17: 35871900-35871900 on Build GRCh38
rs41446448 Chr.17: 35872123-35872123 on Build GRCh38
rs1047764015 Chr.17: 35872223-35872223 on Build GRCh38
rs796760673 Chr.17: 35872082-35872082 on Build GRCh38
rs534598836 Chr.17: 35872142-35872142 on Build GRCh38
rs936529938 Chr.17: 35872044-35872044 on Build GRCh38
rs1001652338 Chr.17: 35871817-35871817 on Build GRCh38
rs41370246 Chr.17: 35871870-35871870 on Build GRCh38
rs565507539 Chr.17: 35872078-35872078 on Build GRCh38
rs3817656 Chr.17: 35872158-35872158 on Build GRCh38
rs55633465 Chr.17: 35871939-35871939 on Build GRCh38
rs1142896 Chr.17: 35872107-35872107 on Build GRCh38
rs929347368 Chr.17: 35872215-35872215 on Build GRCh38
rs925093453 Chr.17: 35872022-35872022 on Build GRCh38
rs977826406 Chr.17: 35872112-35872112 on Build GRCh38
rs11554344 Chr.17: 35872192-35872192 on Build GRCh38
rs368468281 Chr.17: 35872046-35872046 on Build GRCh38
rs949314256 Chr.17: 35872161-35872161 on Build GRCh38
rs1022133385 Chr.17: 35871746-35871748 on Build GRCh38
rs965002900 Chr.17: 35872081-35872081 on Build GRCh38
rs9915517 Chr.17: 35871945-35871945 on Build GRCh38
rs58404351 Chr.17: 35871808-35871808 on Build GRCh38
rs572426234 Chr.17: 35871824-35871824 on Build GRCh38
rs149302576 Chr.17: 35871740-35871740 on Build GRCh38
rs1026392114 Chr.17: 35872067-35872067 on Build GRCh38
rs993530953 Chr.17: 35872066-35872066 on Build GRCh38
rs911758676 Chr.17: 35872036-35872036 on Build GRCh38
rs144272853 Chr.17: 35871783-35871783 on Build GRCh38
rs756983092 Chr.17: 35871853-35871853 on Build GRCh38

View Full Product Details