Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111929438-111929947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MERTK
Entrez Gene ID
10461
Gene Name
MER proto-oncogene, tyrosine kinase
Gene Aliases
MER, RP38, Tyro12, c-Eyk, c-mer
UniGene
Hs.306178
Gene Chromosome Location
Chr.2: 111898479-112039946 on Build GRCh38
Transcripts
NM_006343.2 XM_005263565.4 XM_005263568.4 XM_011510490.2 XM_017003164.1 XM_017003165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6087406 c.452A>T p.D151V Chr.2: 111929510-111929510 on Build GRCh38
5574006 c.459_460insA p.T154fs*23 Chr.2: 111929517-111929518 on Build GRCh38
5047748 c.451G>A p.D151N Chr.2: 111929509-111929509 on Build GRCh38
48560 c.476C>T p.S159F Chr.2: 111929534-111929534 on Build GRCh38
4496500 c.476C>T p.S159F Chr.2: 111929534-111929534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752543013 Chr.2: 111929467-111929467 on Build GRCh38
rs774132699 Chr.2: 111929651-111929651 on Build GRCh38
rs779030836 Chr.2: 111929507-111929507 on Build GRCh38
rs759200969 Chr.2: 111929569-111929569 on Build GRCh38
rs56097129 Chr.2: 111929662-111929662 on Build GRCh38
rs974517624 Chr.2: 111929816-111929816 on Build GRCh38
rs536211802 Chr.2: 111929710-111929710 on Build GRCh38
rs373445297 Chr.2: 111929521-111929521 on Build GRCh38
rs1054477188 Chr.2: 111929869-111929869 on Build GRCh38
rs987365522 Chr.2: 111929852-111929852 on Build GRCh38
rs796423115 Chr.2: 111929579-111929579 on Build GRCh38
rs747428653 Chr.2: 111929531-111929531 on Build GRCh38
rs768990836 Chr.2: 111929533-111929533 on Build GRCh38
rs370201392 Chr.2: 111929475-111929475 on Build GRCh38
rs757295748 Chr.2: 111929485-111929485 on Build GRCh38
rs1042769916 Chr.2: 111929682-111929682 on Build GRCh38
rs144054471 Chr.2: 111929764-111929764 on Build GRCh38
rs777169813 Chr.2: 111929538-111929538 on Build GRCh38
rs1007845219 Chr.2: 111929714-111929714 on Build GRCh38
rs764113900 Chr.2: 111929478-111929478 on Build GRCh38
rs947515641 Chr.2: 111929843-111929843 on Build GRCh38
rs746074841 Chr.2: 111929514-111929514 on Build GRCh38
rs891178290 Chr.2: 111929773-111929773 on Build GRCh38
rs571163398 Chr.2: 111929553-111929553 on Build GRCh38
rs374645167 Chr.2: 111929539-111929539 on Build GRCh38
rs545576536 Chr.2: 111929713-111929713 on Build GRCh38
rs758631997 Chr.2: 111929529-111929529 on Build GRCh38
rs992098360 Chr.2: 111929820-111929820 on Build GRCh38
rs565578860 Chr.2: 111929731-111929731 on Build GRCh38
rs545808644 Chr.2: 111929827-111929828 on Build GRCh38
rs903429313 Chr.2: 111929637-111929637 on Build GRCh38
rs753014895 Chr.2: 111929826-111929827 on Build GRCh38
rs72180817 Chr.2: 111929754-111929762 on Build GRCh38
rs56348540 Chr.2: 111929696-111929696 on Build GRCh38
rs1007360892 Chr.2: 111929543-111929543 on Build GRCh38
rs553341078 Chr.2: 111929727-111929727 on Build GRCh38
rs753768681 Chr.2: 111929481-111929481 on Build GRCh38
rs1017429696 Chr.2: 111929728-111929728 on Build GRCh38
rs780214104 Chr.2: 111929530-111929530 on Build GRCh38
rs561159228 Chr.2: 111929837-111929837 on Build GRCh38
rs915682643 Chr.2: 111929672-111929672 on Build GRCh38
rs869042589 Chr.2: 111929599-111929600 on Build GRCh38
rs938855306 Chr.2: 111929657-111929657 on Build GRCh38
rs184783214 Chr.2: 111929601-111929601 on Build GRCh38
rs1008736791 Chr.2: 111929775-111929775 on Build GRCh38
rs934132390 Chr.2: 111929587-111929587 on Build GRCh38
rs983456710 Chr.2: 111929648-111929648 on Build GRCh38
rs138909702 Chr.2: 111929580-111929581 on Build GRCh38
rs998494327 Chr.2: 111929701-111929701 on Build GRCh38
rs192364027 Chr.2: 111929595-111929595 on Build GRCh38
rs573568476 Chr.2: 111929650-111929650 on Build GRCh38
rs982768184 Chr.2: 111929772-111929772 on Build GRCh38
rs70962966 Chr.2: 111929598-111929599 on Build GRCh38
rs934690033 Chr.2: 111929645-111929645 on Build GRCh38
rs56374282 Chr.2: 111929853-111929853 on Build GRCh38
rs200162501 Chr.2: 111929599-111929599 on Build GRCh38
rs527958949 Chr.2: 111929738-111929738 on Build GRCh38
rs934885792 Chr.2: 111929859-111929859 on Build GRCh38
rs924325155 Chr.2: 111929867-111929867 on Build GRCh38
rs111891621 Chr.2: 111929812-111929812 on Build GRCh38
rs138226978 Chr.2: 111929564-111929564 on Build GRCh38
rs140020217 Chr.2: 111929766-111929766 on Build GRCh38
rs947109821 Chr.2: 111929679-111929679 on Build GRCh38
rs372330161 Chr.2: 111929793-111929793 on Build GRCh38

View Full Product Details