Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56579104-56579634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP3
Entrez Gene ID
8613
Gene Name
phospholipid phosphatase 3
Gene Aliases
Dri42, LPP3, PAP2B, PPAP2B, VCIP
UniGene
Hs.405156
Gene Chromosome Location
Chr.1: 56494747-56579584 on Build GRCh38
Transcripts
NM_003713.4

SNP Detail

SNP ID Chromosome Location
rs1055163040 Chr.1: 56579543-56579543 on Build GRCh38
rs902761077 Chr.1: 56579377-56579377 on Build GRCh38
rs940051104 Chr.1: 56579308-56579308 on Build GRCh38
rs977104927 Chr.1: 56579146-56579146 on Build GRCh38
rs1009521749 Chr.1: 56579386-56579386 on Build GRCh38
rs1033289110 Chr.1: 56579283-56579283 on Build GRCh38
rs146636536 Chr.1: 56579125-56579126 on Build GRCh38
rs200376806 Chr.1: 56579493-56579493 on Build GRCh38
rs879190422 Chr.1: 56579133-56579133 on Build GRCh38
rs937749203 Chr.1: 56579488-56579488 on Build GRCh38
rs939905150 Chr.1: 56579466-56579468 on Build GRCh38
rs931284900 Chr.1: 56579431-56579431 on Build GRCh38
rs1011844091 Chr.1: 56579309-56579309 on Build GRCh38
rs975402139 Chr.1: 56579164-56579164 on Build GRCh38
rs1019743351 Chr.1: 56579352-56579352 on Build GRCh38
rs1019819574 Chr.1: 56579149-56579149 on Build GRCh38
rs61772628 Chr.1: 56579261-56579261 on Build GRCh38
rs531022735 Chr.1: 56579437-56579437 on Build GRCh38
rs557063426 Chr.1: 56579217-56579217 on Build GRCh38
rs903424618 Chr.1: 56579604-56579604 on Build GRCh38
rs111267228 Chr.1: 56579348-56579348 on Build GRCh38
rs866731742 Chr.1: 56579346-56579346 on Build GRCh38
rs983963696 Chr.1: 56579418-56579418 on Build GRCh38
rs1011013039 Chr.1: 56579364-56579364 on Build GRCh38
rs540133056 Chr.1: 56579218-56579218 on Build GRCh38
rs965125516 Chr.1: 56579193-56579193 on Build GRCh38
rs908495204 Chr.1: 56579446-56579446 on Build GRCh38
rs982406013 Chr.1: 56579502-56579502 on Build GRCh38
rs1055108326 Chr.1: 56579336-56579336 on Build GRCh38
rs546963639 Chr.1: 56579566-56579566 on Build GRCh38
rs1001865300 Chr.1: 56579282-56579282 on Build GRCh38
rs952553352 Chr.1: 56579413-56579413 on Build GRCh38
rs1046214181 Chr.1: 56579466-56579474 on Build GRCh38
rs181305627 Chr.1: 56579489-56579489 on Build GRCh38
rs927984594 Chr.1: 56579534-56579534 on Build GRCh38
rs878864442 Chr.1: 56579129-56579129 on Build GRCh38
rs534837578 Chr.1: 56579465-56579466 on Build GRCh38
rs151280937 Chr.1: 56579609-56579610 on Build GRCh38
rs1022270875 Chr.1: 56579312-56579312 on Build GRCh38
rs1019284186 Chr.1: 56579390-56579390 on Build GRCh38
rs908422958 Chr.1: 56579464-56579465 on Build GRCh38
rs1021657601 Chr.1: 56579372-56579372 on Build GRCh38
rs537494271 Chr.1: 56579130-56579130 on Build GRCh38
rs189375844 Chr.1: 56579398-56579398 on Build GRCh38
rs921219788 Chr.1: 56579190-56579190 on Build GRCh38
rs938107394 Chr.1: 56579334-56579334 on Build GRCh38
rs937991206 Chr.1: 56579539-56579539 on Build GRCh38
rs893882669 Chr.1: 56579556-56579556 on Build GRCh38
rs1028056420 Chr.1: 56579407-56579407 on Build GRCh38
rs374584663 Chr.1: 56579323-56579323 on Build GRCh38
rs984060556 Chr.1: 56579453-56579453 on Build GRCh38
rs372391578 Chr.1: 56579257-56579257 on Build GRCh38
rs912610843 Chr.1: 56579126-56579126 on Build GRCh38
rs975433070 Chr.1: 56579388-56579388 on Build GRCh38
rs1019774370 Chr.1: 56579608-56579609 on Build GRCh38
rs931225530 Chr.1: 56579197-56579197 on Build GRCh38
rs143555908 Chr.1: 56579254-56579254 on Build GRCh38
rs561143421 Chr.1: 56579457-56579457 on Build GRCh38
rs1043320788 Chr.1: 56579577-56579577 on Build GRCh38
rs377579078 Chr.1: 56579605-56579605 on Build GRCh38
rs544444716 Chr.1: 56579455-56579455 on Build GRCh38
rs545507815 Chr.1: 56579269-56579269 on Build GRCh38
rs975177431 Chr.1: 56579402-56579402 on Build GRCh38
rs1046391655 Chr.1: 56579313-56579313 on Build GRCh38
rs939955424 Chr.1: 56579468-56579468 on Build GRCh38
rs566483335 Chr.1: 56579608-56579608 on Build GRCh38

View Full Product Details