Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31576683-31577182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
UniGene
Hs.241570
Gene Chromosome Location
Chr.6: 31575567-31578336 on Build GRCh38
Transcripts
NM_000594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734842 c.320G>A p.R107H Chr.6: 31577155-31577155 on Build GRCh38
3394076 c.347G>A p.G116D Chr.6: 31577182-31577182 on Build GRCh38
3253322 c.323G>A p.R108Q Chr.6: 31577158-31577158 on Build GRCh38
5996851 c.317A>G p.N106S Chr.6: 31577152-31577152 on Build GRCh38
3624581 c.233-1G>A p.? Chr.6: 31576766-31576766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1800619 Chr.6: 31576729-31576729 on Build GRCh38
rs918715293 Chr.6: 31577138-31577138 on Build GRCh38
rs941582023 Chr.6: 31577045-31577045 on Build GRCh38
rs13306710 Chr.6: 31576918-31576918 on Build GRCh38
rs755360319 Chr.6: 31577103-31577103 on Build GRCh38
rs956575648 Chr.6: 31576951-31576951 on Build GRCh38
rs565005379 Chr.6: 31576840-31576840 on Build GRCh38
rs772270241 Chr.6: 31576779-31576779 on Build GRCh38
rs1007655911 Chr.6: 31576900-31576900 on Build GRCh38
rs888924952 Chr.6: 31576877-31576877 on Build GRCh38
rs369783162 Chr.6: 31576772-31576772 on Build GRCh38
rs892418402 Chr.6: 31576789-31576789 on Build GRCh38
rs1026305650 Chr.6: 31577000-31577000 on Build GRCh38
rs755342875 Chr.6: 31576746-31576746 on Build GRCh38
rs369268421 Chr.6: 31576984-31576984 on Build GRCh38
rs281865419 Chr.6: 31577157-31577157 on Build GRCh38
rs542219684 Chr.6: 31577022-31577022 on Build GRCh38
rs780350382 Chr.6: 31576731-31576731 on Build GRCh38
rs547381707 Chr.6: 31576925-31576925 on Build GRCh38
rs768383794 Chr.6: 31576855-31576855 on Build GRCh38
rs1800618 Chr.6: 31577141-31577141 on Build GRCh38
rs771174703 Chr.6: 31577162-31577162 on Build GRCh38
rs766958740 Chr.6: 31576866-31576866 on Build GRCh38
rs750865758 Chr.6: 31576725-31576725 on Build GRCh38
rs1038617709 Chr.6: 31577071-31577071 on Build GRCh38
rs1800620 Chr.6: 31576814-31576814 on Build GRCh38
rs1049796997 Chr.6: 31576722-31576722 on Build GRCh38
rs990535649 Chr.6: 31576908-31576909 on Build GRCh38
rs761134052 Chr.6: 31577080-31577080 on Build GRCh38
rs897338372 Chr.6: 31576782-31576782 on Build GRCh38
rs762644987 Chr.6: 31576979-31576979 on Build GRCh38
rs565722472 Chr.6: 31577028-31577028 on Build GRCh38
rs745738344 Chr.6: 31576786-31576786 on Build GRCh38
rs758704433 Chr.6: 31577155-31577155 on Build GRCh38
rs976353858 Chr.6: 31576973-31576973 on Build GRCh38
rs967910752 Chr.6: 31576883-31576883 on Build GRCh38
rs911236926 Chr.6: 31577023-31577023 on Build GRCh38
rs1056210947 Chr.6: 31576828-31576828 on Build GRCh38
rs1000808024 Chr.6: 31576897-31576897 on Build GRCh38
rs941620360 Chr.6: 31576751-31576751 on Build GRCh38
rs966073125 Chr.6: 31576930-31576930 on Build GRCh38
rs530014933 Chr.6: 31577102-31577102 on Build GRCh38
rs766880428 Chr.6: 31577092-31577092 on Build GRCh38
rs756628184 Chr.6: 31576730-31576730 on Build GRCh38
rs28646872 Chr.6: 31576994-31576994 on Build GRCh38
rs922678266 Chr.6: 31576982-31576982 on Build GRCh38
rs4645843 Chr.6: 31576785-31576785 on Build GRCh38
rs947804964 Chr.6: 31576710-31576710 on Build GRCh38
rs754263939 Chr.6: 31576732-31576732 on Build GRCh38
rs745792555 Chr.6: 31576748-31576748 on Build GRCh38
rs4645844 Chr.6: 31576875-31576875 on Build GRCh38
rs3093664 Chr.6: 31576865-31576865 on Build GRCh38
rs371089170 Chr.6: 31577044-31577044 on Build GRCh38
rs779209412 Chr.6: 31577114-31577114 on Build GRCh38
rs775106258 Chr.6: 31576822-31576822 on Build GRCh38
rs749959168 Chr.6: 31577073-31577073 on Build GRCh38
rs769637264 Chr.6: 31576822-31576823 on Build GRCh38
rs779177870 Chr.6: 31576759-31576759 on Build GRCh38
rs770957521 Chr.6: 31576989-31576989 on Build GRCh38
rs747115001 Chr.6: 31577158-31577158 on Build GRCh38
rs375472217 Chr.6: 31577127-31577127 on Build GRCh38
rs764984097 Chr.6: 31576864-31576864 on Build GRCh38
rs377532354 Chr.6: 31576953-31576953 on Build GRCh38
rs190788828 Chr.6: 31576794-31576794 on Build GRCh38

View Full Product Details