Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48230664-48231083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
UniGene
Hs.446146
Gene Chromosome Location
Chr.19: 48203148-48256269 on Build GRCh38
Transcripts
NM_001184900.1 NM_001184901.1 NM_001184902.1 NM_001184903.1 NM_014959.3 NR_033678.1 NR_033679.1 NR_033680.1 XM_006723091.3 XM_006723092.3 XM_006723093.3 XM_006723094.3 XM_006723095.3 XM_006723096.3 XM_006723097.3 XM_006723098.3 XM_006723099.3 XM_006723101.2 XM_006723102.3 XM_006723104.3 XM_006723106.3 XM_006723107.3 XM_006723109.3 XM_011526641.2 XM_011526642.2 XM_011526643.2 XM_011526644.2 XM_011526646.2 XM_011526647.2 XM_011526648.2 XM_011526650.2 XM_011526653.2 XM_017026482.1 XM_017026483.1 XM_017026484.1 XM_017026485.1 XM_017026486.1 XM_017026487.1 XM_017026488.1 XM_017026489.1 XM_017026490.1 XM_017026491.1 XM_017026492.1 XM_017026493.1 XM_017026494.1 XM_017026495.1 XM_017026496.1 XM_017026497.1 XM_017026498.1 XM_017026499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5812944 c.775G>A p.G259S Chr.19: 48230698-48230698 on Build GRCh38
4154286 c.773-7C>G p.? Chr.19: 48230707-48230707 on Build GRCh38
712107 c.409G>C p.E137Q Chr.19: 48230825-48230825 on Build GRCh38
3823531 c.453C>G p.L151L Chr.19: 48230781-48230781 on Build GRCh38
5697861 c.391G>A p.D131N Chr.19: 48230843-48230843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755425697 Chr.19: 48230745-48230745 on Build GRCh38
rs375996935 Chr.19: 48231032-48231032 on Build GRCh38
rs376991226 Chr.19: 48230769-48230769 on Build GRCh38
rs749800607 Chr.19: 48230836-48230836 on Build GRCh38
rs563466972 Chr.19: 48230751-48230751 on Build GRCh38
rs752967190 Chr.19: 48230897-48230897 on Build GRCh38
rs146362031 Chr.19: 48230806-48230806 on Build GRCh38
rs757899979 Chr.19: 48231045-48231045 on Build GRCh38
rs377425345 Chr.19: 48230734-48230734 on Build GRCh38
rs150049816 Chr.19: 48230838-48230838 on Build GRCh38
rs148708617 Chr.19: 48230886-48230886 on Build GRCh38
rs758537107 Chr.19: 48230799-48230799 on Build GRCh38
rs554659325 Chr.19: 48231000-48231000 on Build GRCh38
rs761405892 Chr.19: 48230868-48230868 on Build GRCh38
rs541040361 Chr.19: 48230690-48230690 on Build GRCh38
rs549990776 Chr.19: 48230874-48230874 on Build GRCh38
rs767205602 Chr.19: 48230729-48230729 on Build GRCh38
rs905396709 Chr.19: 48230921-48230921 on Build GRCh38
rs922661751 Chr.19: 48230695-48230695 on Build GRCh38
rs370075376 Chr.19: 48230730-48230730 on Build GRCh38
rs185151716 Chr.19: 48231059-48231059 on Build GRCh38
rs768431244 Chr.19: 48230859-48230859 on Build GRCh38
rs59878320 Chr.19: 48230938-48230938 on Build GRCh38
rs34646986 Chr.19: 48230779-48230779 on Build GRCh38
rs58650907 Chr.19: 48230750-48230750 on Build GRCh38
rs964088585 Chr.19: 48230817-48230817 on Build GRCh38
rs535028752 Chr.19: 48230959-48230959 on Build GRCh38
rs778794766 Chr.19: 48230818-48230818 on Build GRCh38
rs758449330 Chr.19: 48230736-48230736 on Build GRCh38
rs747188833 Chr.19: 48230744-48230744 on Build GRCh38
rs150036211 Chr.19: 48230957-48230957 on Build GRCh38
rs761677875 Chr.19: 48230867-48230867 on Build GRCh38
rs199629022 Chr.19: 48230860-48230860 on Build GRCh38
rs773848177 Chr.19: 48230716-48230716 on Build GRCh38
rs988000898 Chr.19: 48230861-48230861 on Build GRCh38
rs779328099 Chr.19: 48231035-48231035 on Build GRCh38
rs370395701 Chr.19: 48230855-48230855 on Build GRCh38
rs779478328 Chr.19: 48231054-48231054 on Build GRCh38
rs376578612 Chr.19: 48230826-48230826 on Build GRCh38
rs1005454978 Chr.19: 48231022-48231022 on Build GRCh38
rs757618336 Chr.19: 48231034-48231034 on Build GRCh38
rs750936952 Chr.19: 48231039-48231039 on Build GRCh38
rs766187048 Chr.19: 48230712-48230712 on Build GRCh38
rs201348979 Chr.19: 48231006-48231006 on Build GRCh38
rs144642248 Chr.19: 48230919-48230919 on Build GRCh38
rs773837448 Chr.19: 48230755-48230755 on Build GRCh38
rs753780239 Chr.19: 48230710-48230710 on Build GRCh38
rs145413035 Chr.19: 48230856-48230856 on Build GRCh38
rs762719288 Chr.19: 48230711-48230711 on Build GRCh38
rs781639798 Chr.19: 48230811-48230811 on Build GRCh38
rs774111311 Chr.19: 48230764-48230764 on Build GRCh38
rs879043118 Chr.19: 48230948-48230948 on Build GRCh38
rs777257654 Chr.19: 48230687-48230687 on Build GRCh38
rs139343522 Chr.19: 48230926-48230926 on Build GRCh38
rs975563099 Chr.19: 48230824-48230824 on Build GRCh38
rs748512582 Chr.19: 48230812-48230812 on Build GRCh38
rs547827116 Chr.19: 48230813-48230813 on Build GRCh38
rs752399327 Chr.19: 48230853-48230854 on Build GRCh38
rs759641064 Chr.19: 48230875-48230875 on Build GRCh38
rs369665290 Chr.19: 48230760-48230760 on Build GRCh38
rs756605952 Chr.19: 48230900-48230900 on Build GRCh38
rs745564007 Chr.19: 48230854-48230854 on Build GRCh38
rs753446469 Chr.19: 48230796-48230796 on Build GRCh38
rs142485811 Chr.19: 48230794-48230794 on Build GRCh38
rs771578974 Chr.19: 48230950-48230950 on Build GRCh38
rs769671624 Chr.19: 48230994-48230994 on Build GRCh38
rs371457675 Chr.19: 48231028-48231028 on Build GRCh38
rs545897168 Chr.19: 48230771-48230771 on Build GRCh38
rs759442795 Chr.19: 48230720-48230720 on Build GRCh38
rs754852892 Chr.19: 48230768-48230770 on Build GRCh38
rs372985137 Chr.19: 48230804-48230804 on Build GRCh38
rs543716168 Chr.19: 48231016-48231016 on Build GRCh38
rs1001301549 Chr.19: 48230772-48230772 on Build GRCh38
rs979470099 Chr.19: 48230920-48230920 on Build GRCh38
rs761767678 Chr.19: 48230990-48230990 on Build GRCh38
rs370242064 Chr.19: 48230789-48230789 on Build GRCh38
rs768449522 Chr.19: 48230965-48230965 on Build GRCh38
rs778221625 Chr.19: 48230850-48230850 on Build GRCh38
rs758184482 Chr.19: 48230976-48230976 on Build GRCh38
rs868409433 Chr.19: 48230787-48230787 on Build GRCh38
rs377045399 Chr.19: 48230709-48230709 on Build GRCh38
rs780290989 Chr.19: 48230739-48230739 on Build GRCh38
rs1043742422 Chr.19: 48230746-48230746 on Build GRCh38
rs779639405 Chr.19: 48230952-48230952 on Build GRCh38
rs2304134 Chr.19: 48230761-48230761 on Build GRCh38
rs745416902 Chr.19: 48230944-48230944 on Build GRCh38
rs752981551 Chr.19: 48231015-48231015 on Build GRCh38
rs139383364 Chr.19: 48230814-48230814 on Build GRCh38
rs17854917 Chr.19: 48230791-48230791 on Build GRCh38
rs572420336 Chr.19: 48230686-48230686 on Build GRCh38
rs368761208 Chr.19: 48231027-48231027 on Build GRCh38
rs574606828 Chr.19: 48230735-48230735 on Build GRCh38
rs778494684 Chr.19: 48230816-48230816 on Build GRCh38
rs150982505 Chr.19: 48231005-48231005 on Build GRCh38
rs764067338 Chr.19: 48230795-48230795 on Build GRCh38
rs1040075465 Chr.19: 48230691-48230691 on Build GRCh38
rs754402344 Chr.19: 48230924-48230924 on Build GRCh38
rs1047926244 Chr.19: 48230958-48230958 on Build GRCh38
rs766481667 Chr.19: 48230869-48230869 on Build GRCh38
rs375071739 Chr.19: 48230962-48230962 on Build GRCh38
rs542641676 Chr.19: 48230810-48230810 on Build GRCh38
rs769438877 Chr.19: 48230696-48230696 on Build GRCh38
rs772812237 Chr.19: 48230707-48230707 on Build GRCh38
rs776337527 Chr.19: 48230968-48230968 on Build GRCh38
rs943649078 Chr.19: 48230754-48230754 on Build GRCh38

View Full Product Details