Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56495172-56495726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP3
Entrez Gene ID
8613
Gene Name
phospholipid phosphatase 3
Gene Aliases
Dri42, LPP3, PAP2B, PPAP2B, VCIP
UniGene
Hs.405156
Gene Chromosome Location
Chr.1: 56494747-56579584 on Build GRCh38
Transcripts
NM_003713.4

SNP Detail

SNP ID Chromosome Location
rs559380579 Chr.1: 56495214-56495214 on Build GRCh38
rs962491355 Chr.1: 56495247-56495247 on Build GRCh38
rs1001022161 Chr.1: 56495664-56495664 on Build GRCh38
rs1013808178 Chr.1: 56495646-56495646 on Build GRCh38
rs960917952 Chr.1: 56495384-56495384 on Build GRCh38
rs1050189370 Chr.1: 56495227-56495227 on Build GRCh38
rs545630606 Chr.1: 56495337-56495337 on Build GRCh38
rs190751782 Chr.1: 56495648-56495648 on Build GRCh38
rs573361350 Chr.1: 56495246-56495246 on Build GRCh38
rs933216251 Chr.1: 56495514-56495514 on Build GRCh38
rs1023326278 Chr.1: 56495615-56495615 on Build GRCh38
rs1037381887 Chr.1: 56495272-56495272 on Build GRCh38
rs773002165 Chr.1: 56495537-56495537 on Build GRCh38
rs867858950 Chr.1: 56495453-56495453 on Build GRCh38
rs888889633 Chr.1: 56495242-56495242 on Build GRCh38
rs956605417 Chr.1: 56495672-56495672 on Build GRCh38
rs1032425090 Chr.1: 56495465-56495465 on Build GRCh38
rs985985190 Chr.1: 56495546-56495546 on Build GRCh38
rs113082878 Chr.1: 56495542-56495542 on Build GRCh38
rs897522582 Chr.1: 56495340-56495340 on Build GRCh38
rs765389535 Chr.1: 56495392-56495392 on Build GRCh38
rs186450936 Chr.1: 56495639-56495639 on Build GRCh38
rs1023880600 Chr.1: 56495647-56495647 on Build GRCh38
rs7519755 Chr.1: 56495379-56495379 on Build GRCh38
rs941868477 Chr.1: 56495557-56495557 on Build GRCh38
rs10888976 Chr.1: 56495682-56495682 on Build GRCh38
rs972350764 Chr.1: 56495251-56495251 on Build GRCh38
rs537773664 Chr.1: 56495629-56495629 on Build GRCh38
rs1036311540 Chr.1: 56495305-56495305 on Build GRCh38
rs935127729 Chr.1: 56495429-56495429 on Build GRCh38
rs565326908 Chr.1: 56495350-56495350 on Build GRCh38
rs551273463 Chr.1: 56495396-56495396 on Build GRCh38
rs928967136 Chr.1: 56495608-56495608 on Build GRCh38
rs1013778491 Chr.1: 56495391-56495391 on Build GRCh38
rs969015202 Chr.1: 56495633-56495633 on Build GRCh38
rs889091662 Chr.1: 56495469-56495469 on Build GRCh38
rs774531895 Chr.1: 56495592-56495592 on Build GRCh38
rs923205046 Chr.1: 56495488-56495488 on Build GRCh38
rs752038782 Chr.1: 56495703-56495703 on Build GRCh38
rs181442104 Chr.1: 56495372-56495372 on Build GRCh38
rs923190168 Chr.1: 56495442-56495442 on Build GRCh38
rs993162732 Chr.1: 56495365-56495365 on Build GRCh38
rs546724110 Chr.1: 56495444-56495444 on Build GRCh38
rs1035978551 Chr.1: 56495552-56495552 on Build GRCh38
rs925154847 Chr.1: 56495414-56495414 on Build GRCh38
rs988088256 Chr.1: 56495476-56495476 on Build GRCh38
rs979569957 Chr.1: 56495401-56495401 on Build GRCh38
rs183248028 Chr.1: 56495565-56495565 on Build GRCh38
rs988121152 Chr.1: 56495702-56495702 on Build GRCh38
rs544449804 Chr.1: 56495258-56495259 on Build GRCh38
rs1016450586 Chr.1: 56495238-56495238 on Build GRCh38
rs954635905 Chr.1: 56495705-56495705 on Build GRCh38
rs185927668 Chr.1: 56495404-56495404 on Build GRCh38
rs1050374032 Chr.1: 56495462-56495462 on Build GRCh38
rs529676634 Chr.1: 56495433-56495433 on Build GRCh38
rs568940469 Chr.1: 56495575-56495575 on Build GRCh38
rs897556754 Chr.1: 56495606-56495606 on Build GRCh38
rs750446893 Chr.1: 56495593-56495593 on Build GRCh38
rs190739990 Chr.1: 56495470-56495470 on Build GRCh38
rs960399396 Chr.1: 56495349-56495349 on Build GRCh38
rs956760517 Chr.1: 56495691-56495691 on Build GRCh38
rs140667727 Chr.1: 56495547-56495547 on Build GRCh38
rs1038156422 Chr.1: 56495474-56495474 on Build GRCh38

View Full Product Details