Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56524255-56524629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP3
Entrez Gene ID
8613
Gene Name
phospholipid phosphatase 3
Gene Aliases
Dri42, LPP3, PAP2B, PPAP2B, VCIP
UniGene
Hs.405156
Gene Chromosome Location
Chr.1: 56494747-56579584 on Build GRCh38
Transcripts
NM_003713.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4755909 c.350C>T p.T117M Chr.1: 56524502-56524502 on Build GRCh38
4755910 c.305C>T p.T102M Chr.1: 56524547-56524547 on Build GRCh38
458979 c.454G>T p.G152W Chr.1: 56524398-56524398 on Build GRCh38
4824334 c.531G>C p.Q177H Chr.1: 56524321-56524321 on Build GRCh38
7131576 c.306G>T p.T102T Chr.1: 56524546-56524546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370936833 Chr.1: 56524363-56524363 on Build GRCh38
rs753280597 Chr.1: 56524509-56524509 on Build GRCh38
rs982341573 Chr.1: 56524400-56524400 on Build GRCh38
rs776490806 Chr.1: 56524594-56524594 on Build GRCh38
rs779974600 Chr.1: 56524300-56524300 on Build GRCh38
rs777523216 Chr.1: 56524534-56524534 on Build GRCh38
rs758626117 Chr.1: 56524481-56524481 on Build GRCh38
rs371910856 Chr.1: 56524546-56524546 on Build GRCh38
rs756131844 Chr.1: 56524294-56524294 on Build GRCh38
rs775715678 Chr.1: 56524370-56524370 on Build GRCh38
rs375354068 Chr.1: 56524423-56524423 on Build GRCh38
rs754749952 Chr.1: 56524312-56524312 on Build GRCh38
rs767309067 Chr.1: 56524505-56524505 on Build GRCh38
rs370490743 Chr.1: 56524486-56524486 on Build GRCh38
rs200887319 Chr.1: 56524318-56524318 on Build GRCh38
rs202227786 Chr.1: 56524452-56524452 on Build GRCh38
rs931768539 Chr.1: 56524605-56524605 on Build GRCh38
rs749957963 Chr.1: 56524395-56524395 on Build GRCh38
rs753858581 Chr.1: 56524436-56524436 on Build GRCh38
rs776542639 Chr.1: 56524489-56524489 on Build GRCh38
rs201168992 Chr.1: 56524501-56524501 on Build GRCh38
rs747883707 Chr.1: 56524479-56524479 on Build GRCh38
rs771463266 Chr.1: 56524350-56524350 on Build GRCh38
rs778831660 Chr.1: 56524460-56524460 on Build GRCh38
rs193118962 Chr.1: 56524559-56524559 on Build GRCh38
rs367613181 Chr.1: 56524474-56524474 on Build GRCh38
rs749523087 Chr.1: 56524493-56524493 on Build GRCh38
rs200079726 Chr.1: 56524360-56524360 on Build GRCh38
rs761724959 Chr.1: 56524605-56524605 on Build GRCh38
rs774820793 Chr.1: 56524602-56524602 on Build GRCh38
rs749937717 Chr.1: 56524293-56524293 on Build GRCh38
rs766245772 Chr.1: 56524402-56524402 on Build GRCh38
rs751819679 Chr.1: 56524535-56524535 on Build GRCh38
rs749068177 Chr.1: 56524306-56524306 on Build GRCh38
rs928116304 Chr.1: 56524457-56524457 on Build GRCh38
rs777936496 Chr.1: 56524482-56524482 on Build GRCh38
rs747029264 Chr.1: 56524338-56524338 on Build GRCh38
rs151304097 Chr.1: 56524506-56524506 on Build GRCh38
rs749692108 Chr.1: 56524579-56524579 on Build GRCh38
rs974898389 Chr.1: 56524504-56524504 on Build GRCh38
rs148766697 Chr.1: 56524495-56524495 on Build GRCh38
rs769189187 Chr.1: 56524587-56524587 on Build GRCh38
rs941431841 Chr.1: 56524337-56524337 on Build GRCh38
rs761614114 Chr.1: 56524383-56524383 on Build GRCh38
rs763340277 Chr.1: 56524502-56524502 on Build GRCh38
rs200383278 Chr.1: 56524508-56524508 on Build GRCh38
rs201684971 Chr.1: 56524533-56524533 on Build GRCh38
rs754998338 Chr.1: 56524510-56524510 on Build GRCh38
rs140476566 Chr.1: 56524511-56524511 on Build GRCh38
rs778042742 Chr.1: 56524394-56524394 on Build GRCh38
rs773296707 Chr.1: 56524336-56524336 on Build GRCh38
rs755645709 Chr.1: 56524396-56524396 on Build GRCh38
rs780667335 Chr.1: 56524576-56524576 on Build GRCh38
rs778546686 Chr.1: 56524314-56524314 on Build GRCh38
rs752524983 Chr.1: 56524522-56524522 on Build GRCh38
rs754837219 Chr.1: 56524459-56524459 on Build GRCh38
rs757418849 Chr.1: 56524540-56524540 on Build GRCh38
rs142369156 Chr.1: 56524547-56524547 on Build GRCh38
rs765676648 Chr.1: 56524364-56524364 on Build GRCh38
rs747117364 Chr.1: 56524485-56524485 on Build GRCh38

View Full Product Details