Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 118932765-118933282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF11B
Entrez Gene ID
4982
Gene Name
TNF receptor superfamily member 11b
Gene Aliases
OCIF, OPG, PDB5, TR1
UniGene
Hs.81791
Gene Chromosome Location
Chr.8: 118923557-118952144 on Build GRCh38
Transcripts
NM_002546.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6250166 c.276C>A p.Y92* Chr.8: 118933055-118933055 on Build GRCh38
6179654 c.105C>G p.T35T Chr.8: 118933226-118933226 on Build GRCh38
1240468 c.277G>A p.V93I Chr.8: 118933054-118933054 on Build GRCh38
4903060 c.94G>A p.D32N Chr.8: 118933237-118933237 on Build GRCh38
6112154 c.255C>G p.P85P Chr.8: 118933076-118933076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756316405 Chr.8: 118932933-118932933 on Build GRCh38
rs770097032 Chr.8: 118933089-118933092 on Build GRCh38
rs1564858 Chr.8: 118932927-118932927 on Build GRCh38
rs1030137776 Chr.8: 118932846-118932846 on Build GRCh38
rs776644606 Chr.8: 118933256-118933256 on Build GRCh38
rs748687558 Chr.8: 118933052-118933052 on Build GRCh38
rs755350563 Chr.8: 118933045-118933045 on Build GRCh38
rs1007358655 Chr.8: 118933193-118933193 on Build GRCh38
rs758662133 Chr.8: 118933167-118933167 on Build GRCh38
rs749701904 Chr.8: 118932943-118932943 on Build GRCh38
rs746791412 Chr.8: 118933015-118933015 on Build GRCh38
rs104894092 Chr.8: 118932982-118932982 on Build GRCh38
rs986205746 Chr.8: 118932866-118932866 on Build GRCh38
rs769186979 Chr.8: 118933076-118933076 on Build GRCh38
rs1057298539 Chr.8: 118932952-118932952 on Build GRCh38
rs774979470 Chr.8: 118933001-118933001 on Build GRCh38
rs755659288 Chr.8: 118933214-118933214 on Build GRCh38
rs748597515 Chr.8: 118932989-118932989 on Build GRCh38
rs1023694993 Chr.8: 118932791-118932791 on Build GRCh38
rs1001173056 Chr.8: 118932802-118932802 on Build GRCh38
rs781271789 Chr.8: 118932923-118932923 on Build GRCh38
rs753111952 Chr.8: 118933166-118933166 on Build GRCh38
rs757467378 Chr.8: 118932916-118932916 on Build GRCh38
rs199830002 Chr.8: 118933079-118933079 on Build GRCh38
rs146450643 Chr.8: 118933262-118933262 on Build GRCh38
rs116321565 Chr.8: 118933144-118933144 on Build GRCh38
rs1018837640 Chr.8: 118932855-118932855 on Build GRCh38
rs372860364 Chr.8: 118932926-118932926 on Build GRCh38
rs372009662 Chr.8: 118933022-118933022 on Build GRCh38
rs768977312 Chr.8: 118932954-118932954 on Build GRCh38
rs200629343 Chr.8: 118932999-118932999 on Build GRCh38
rs371566584 Chr.8: 118933075-118933075 on Build GRCh38
rs757189572 Chr.8: 118932906-118932906 on Build GRCh38
rs770435026 Chr.8: 118932997-118932997 on Build GRCh38
rs765861529 Chr.8: 118932882-118932882 on Build GRCh38
rs756124545 Chr.8: 118933024-118933024 on Build GRCh38
rs767538379 Chr.8: 118933127-118933127 on Build GRCh38
rs201167408 Chr.8: 118933000-118933000 on Build GRCh38
rs188556114 Chr.8: 118932893-118932893 on Build GRCh38
rs786205594 Chr.8: 118933137-118933137 on Build GRCh38
rs772995755 Chr.8: 118932967-118932967 on Build GRCh38
rs200071478 Chr.8: 118933105-118933105 on Build GRCh38
rs754063545 Chr.8: 118933150-118933150 on Build GRCh38
rs550357283 Chr.8: 118932787-118932787 on Build GRCh38
rs759163971 Chr.8: 118932891-118932891 on Build GRCh38
rs370759767 Chr.8: 118932969-118932969 on Build GRCh38
rs752491803 Chr.8: 118932896-118932896 on Build GRCh38
rs771971014 Chr.8: 118932965-118932965 on Build GRCh38
rs11573906 Chr.8: 118933021-118933021 on Build GRCh38
rs975856053 Chr.8: 118932823-118932823 on Build GRCh38
rs529894410 Chr.8: 118933035-118933035 on Build GRCh38
rs762633243 Chr.8: 118933080-118933080 on Build GRCh38
rs546710840 Chr.8: 118933055-118933055 on Build GRCh38
rs780302337 Chr.8: 118932942-118932942 on Build GRCh38
rs144654126 Chr.8: 118933097-118933097 on Build GRCh38
rs768572122 Chr.8: 118933090-118933090 on Build GRCh38
rs150777320 Chr.8: 118933227-118933227 on Build GRCh38
rs775116631 Chr.8: 118933046-118933046 on Build GRCh38
rs202090603 Chr.8: 118933234-118933234 on Build GRCh38
rs903548041 Chr.8: 118932792-118932792 on Build GRCh38
rs4876870 Chr.8: 118933235-118933235 on Build GRCh38
rs751552719 Chr.8: 118933016-118933016 on Build GRCh38
rs369646136 Chr.8: 118933054-118933054 on Build GRCh38
rs762822313 Chr.8: 118932898-118932898 on Build GRCh38
rs375274060 Chr.8: 118933023-118933023 on Build GRCh38
rs956740221 Chr.8: 118932819-118932819 on Build GRCh38
rs778414770 Chr.8: 118933183-118933183 on Build GRCh38
rs755154434 Chr.8: 118933151-118933151 on Build GRCh38
rs104894091 Chr.8: 118933071-118933071 on Build GRCh38
rs953235402 Chr.8: 118933141-118933141 on Build GRCh38
rs766512148 Chr.8: 118933253-118933253 on Build GRCh38
rs531660121 Chr.8: 118933136-118933136 on Build GRCh38
rs747461694 Chr.8: 118933197-118933197 on Build GRCh38
rs745793701 Chr.8: 118932929-118932929 on Build GRCh38
rs760463291 Chr.8: 118933252-118933252 on Build GRCh38
rs1030106642 Chr.8: 118933104-118933104 on Build GRCh38
rs1030960174 Chr.8: 118932809-118932809 on Build GRCh38
rs780363937 Chr.8: 118933028-118933028 on Build GRCh38
rs776161597 Chr.8: 118932886-118932886 on Build GRCh38
rs184799075 Chr.8: 118932865-118932865 on Build GRCh38
rs149349985 Chr.8: 118932955-118932955 on Build GRCh38
rs762911895 Chr.8: 118933003-118933003 on Build GRCh38
rs751542395 Chr.8: 118932915-118932915 on Build GRCh38
rs142201380 Chr.8: 118933145-118933145 on Build GRCh38
rs953432652 Chr.8: 118932858-118932858 on Build GRCh38
rs144062067 Chr.8: 118933245-118933245 on Build GRCh38
rs774246126 Chr.8: 118933096-118933096 on Build GRCh38

View Full Product Details