Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175074525-175075079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF2
Entrez Gene ID
1386
Gene Name
activating transcription factor 2
Gene Aliases
CRE-BP1, CREB-2, CREB2, HB16, TREB7
UniGene
Hs.592510
Gene Chromosome Location
Chr.2: 175072250-175168206 on Build GRCh38
Transcripts
NM_001256090.1 NM_001256091.1 NM_001256092.1 NM_001256093.1 NM_001880.3 NR_045768.1 NR_045769.1 NR_045770.1 NR_045771.1 NR_045772.1 NR_045773.1 NR_045774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391136 c.1309A>G p.S437G Chr.2: 175074818-175074818 on Build GRCh38
2894204 c.1332G>A p.P444P Chr.2: 175074795-175074795 on Build GRCh38
3379865 c.1331C>A p.P444Q Chr.2: 175074796-175074796 on Build GRCh38
271115 c.1398C>A p.T466T Chr.2: 175074729-175074729 on Build GRCh38
1733616 c.1477G>A p.V493I Chr.2: 175074650-175074650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763190197 Chr.2: 175074633-175074633 on Build GRCh38
rs753144395 Chr.2: 175074656-175074656 on Build GRCh38
rs376796632 Chr.2: 175074875-175074875 on Build GRCh38
rs772072332 Chr.2: 175074756-175074756 on Build GRCh38
rs749514665 Chr.2: 175074739-175074739 on Build GRCh38
rs551197382 Chr.2: 175074965-175074965 on Build GRCh38
rs55961757 Chr.2: 175074583-175074583 on Build GRCh38
rs749822996 Chr.2: 175074653-175074653 on Build GRCh38
rs778434807 Chr.2: 175074573-175074573 on Build GRCh38
rs1013328885 Chr.2: 175075033-175075033 on Build GRCh38
rs776915363 Chr.2: 175074889-175074889 on Build GRCh38
rs541449241 Chr.2: 175074639-175074639 on Build GRCh38
rs1057193337 Chr.2: 175074569-175074569 on Build GRCh38
rs746816475 Chr.2: 175074587-175074587 on Build GRCh38
rs188875772 Chr.2: 175074642-175074642 on Build GRCh38
rs998443906 Chr.2: 175075051-175075051 on Build GRCh38
rs762676390 Chr.2: 175074905-175074905 on Build GRCh38
rs201852265 Chr.2: 175074577-175074577 on Build GRCh38
rs371171989 Chr.2: 175074886-175074886 on Build GRCh38
rs764967442 Chr.2: 175074803-175074803 on Build GRCh38
rs36047041 Chr.2: 175074807-175074807 on Build GRCh38
rs900583670 Chr.2: 175074669-175074669 on Build GRCh38
rs79907460 Chr.2: 175075034-175075034 on Build GRCh38
rs141304902 Chr.2: 175074705-175074705 on Build GRCh38
rs775524817 Chr.2: 175074873-175074873 on Build GRCh38
rs888007558 Chr.2: 175074976-175074976 on Build GRCh38
rs1033319539 Chr.2: 175074611-175074611 on Build GRCh38
rs894852040 Chr.2: 175075024-175075024 on Build GRCh38
rs773341486 Chr.2: 175074624-175074624 on Build GRCh38
rs781022245 Chr.2: 175074730-175074730 on Build GRCh38
rs754913663 Chr.2: 175074825-175074825 on Build GRCh38
rs2302663 Chr.2: 175074876-175074876 on Build GRCh38
rs774384167 Chr.2: 175074646-175074646 on Build GRCh38
rs772160617 Chr.2: 175074869-175074869 on Build GRCh38
rs771180346 Chr.2: 175074742-175074742 on Build GRCh38
rs768363052 Chr.2: 175074728-175074728 on Build GRCh38
rs761194042 Chr.2: 175074773-175074773 on Build GRCh38
rs747963598 Chr.2: 175074835-175074835 on Build GRCh38
rs765943326 Chr.2: 175074820-175074820 on Build GRCh38
rs937145008 Chr.2: 175074554-175074554 on Build GRCh38
rs555768175 Chr.2: 175074745-175074745 on Build GRCh38
rs761437442 Chr.2: 175074798-175074798 on Build GRCh38
rs770965410 Chr.2: 175074637-175074637 on Build GRCh38
rs538343616 Chr.2: 175074900-175074900 on Build GRCh38
rs537479900 Chr.2: 175074727-175074727 on Build GRCh38
rs762248247 Chr.2: 175074897-175074897 on Build GRCh38
rs147478928 Chr.2: 175074760-175074760 on Build GRCh38
rs373544668 Chr.2: 175074566-175074566 on Build GRCh38
rs762946975 Chr.2: 175074600-175074600 on Build GRCh38
rs557063934 Chr.2: 175074957-175074957 on Build GRCh38
rs759627533 Chr.2: 175074752-175074752 on Build GRCh38
rs72927392 Chr.2: 175074987-175074987 on Build GRCh38
rs777018630 Chr.2: 175074796-175074796 on Build GRCh38
rs780915529 Chr.2: 175074828-175074828 on Build GRCh38
rs746048259 Chr.2: 175074861-175074861 on Build GRCh38
rs773732331 Chr.2: 175074580-175074580 on Build GRCh38
rs755824068 Chr.2: 175074565-175074565 on Build GRCh38
rs760104049 Chr.2: 175074650-175074650 on Build GRCh38
rs755847095 Chr.2: 175074839-175074839 on Build GRCh38
rs760950559 Chr.2: 175074679-175074679 on Build GRCh38
rs143157308 Chr.2: 175074613-175074613 on Build GRCh38
rs139006296 Chr.2: 175074744-175074744 on Build GRCh38
rs992616431 Chr.2: 175074858-175074858 on Build GRCh38
rs954668428 Chr.2: 175074717-175074717 on Build GRCh38
rs556817471 Chr.2: 175074702-175074702 on Build GRCh38
rs2302664 Chr.2: 175074989-175074989 on Build GRCh38
rs1041594096 Chr.2: 175074588-175074588 on Build GRCh38
rs764479396 Chr.2: 175074684-175074684 on Build GRCh38
rs770839889 Chr.2: 175074887-175074891 on Build GRCh38
rs756749428 Chr.2: 175074571-175074571 on Build GRCh38
rs771117385 Chr.2: 175074723-175074723 on Build GRCh38
rs142443599 Chr.2: 175074815-175074815 on Build GRCh38
rs201072854 Chr.2: 175074629-175074629 on Build GRCh38
rs756908057 Chr.2: 175074689-175074689 on Build GRCh38
rs749945884 Chr.2: 175074713-175074713 on Build GRCh38
rs372589188 Chr.2: 175074614-175074614 on Build GRCh38
rs753058454 Chr.2: 175074561-175074561 on Build GRCh38
rs766030518 Chr.2: 175074906-175074906 on Build GRCh38
rs567924476 Chr.2: 175074795-175074795 on Build GRCh38
rs2193004 Chr.2: 175074868-175074868 on Build GRCh38
rs753454204 Chr.2: 175074685-175074685 on Build GRCh38
rs751077492 Chr.2: 175074912-175074912 on Build GRCh38
rs751503459 Chr.2: 175074823-175074823 on Build GRCh38
rs959081624 Chr.2: 175074635-175074635 on Build GRCh38
rs369362331 Chr.2: 175074651-175074651 on Build GRCh38
rs753463114 Chr.2: 175074570-175074570 on Build GRCh38

View Full Product Details