Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 164771359-164771858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100505795
Entrez Gene ID
100505795
Gene Name
uncharacterized LOC100505795
Gene Aliases
-
UniGene
Hs.661039
Gene Chromosome Location
Chr.1: 164769116-164774641 on Build GRCh38
Transcripts
NR_038072.1 NM_001204961.1 NM_001204963.1 NM_002585.3 XM_005245228.3 XM_005245229.3 XM_011509590.1 XM_011509591.2 XM_011509592.1 XM_017001395.1 XM_017001396.1

Gene Symbol
PBX1
Entrez Gene ID
5087
Gene Name
PBX homeobox 1
Gene Aliases
-
UniGene
Hs.557097
Gene Chromosome Location
Chr.1: 164559360-164886047 on Build GRCh38
Transcripts
NR_038072.1 NM_001204961.1 NM_001204963.1 NM_002585.3 XM_005245228.3 XM_005245229.3 XM_011509590.1 XM_011509591.2 XM_011509592.1 XM_017001395.1 XM_017001396.1

SNP Detail

SNP ID Chromosome Location
rs12098218 Chr.1: 164771654-164771654 on Build GRCh38
rs548553241 Chr.1: 164771608-164771608 on Build GRCh38
rs144730101 Chr.1: 164771583-164771583 on Build GRCh38
rs942104440 Chr.1: 164771733-164771733 on Build GRCh38
rs1004104711 Chr.1: 164771714-164771714 on Build GRCh38
rs572303883 Chr.1: 164771455-164771455 on Build GRCh38
rs919261244 Chr.1: 164771790-164771790 on Build GRCh38
rs527696241 Chr.1: 164771621-164771621 on Build GRCh38
rs982283544 Chr.1: 164771534-164771534 on Build GRCh38
rs547739564 Chr.1: 164771639-164771639 on Build GRCh38
rs185995972 Chr.1: 164771477-164771477 on Build GRCh38
rs906690206 Chr.1: 164771490-164771490 on Build GRCh38
rs749046669 Chr.1: 164771405-164771405 on Build GRCh38
rs897849313 Chr.1: 164771459-164771459 on Build GRCh38
rs958462552 Chr.1: 164771592-164771592 on Build GRCh38
rs985943981 Chr.1: 164771691-164771691 on Build GRCh38
rs766390910 Chr.1: 164771761-164771761 on Build GRCh38
rs1014895045 Chr.1: 164771425-164771425 on Build GRCh38
rs967474707 Chr.1: 164771645-164771645 on Build GRCh38
rs942009807 Chr.1: 164771430-164771430 on Build GRCh38
rs951317478 Chr.1: 164771400-164771400 on Build GRCh38
rs562945325 Chr.1: 164771514-164771514 on Build GRCh38
rs3215011 Chr.1: 164771581-164771581 on Build GRCh38
rs1017284288 Chr.1: 164771801-164771801 on Build GRCh38
rs1034061701 Chr.1: 164771562-164771562 on Build GRCh38
rs973918811 Chr.1: 164771758-164771758 on Build GRCh38
rs1002382784 Chr.1: 164771550-164771550 on Build GRCh38
rs74121225 Chr.1: 164771487-164771487 on Build GRCh38
rs963394106 Chr.1: 164771802-164771802 on Build GRCh38
rs946396197 Chr.1: 164771641-164771641 on Build GRCh38
rs190342625 Chr.1: 164771613-164771613 on Build GRCh38
rs982700960 Chr.1: 164771422-164771422 on Build GRCh38
rs910636986 Chr.1: 164771395-164771395 on Build GRCh38
rs928138379 Chr.1: 164771578-164771578 on Build GRCh38
rs149782092 Chr.1: 164771478-164771478 on Build GRCh38
rs879071197 Chr.1: 164771694-164771694 on Build GRCh38
rs973489253 Chr.1: 164771431-164771431 on Build GRCh38
rs139695397 Chr.1: 164771426-164771426 on Build GRCh38
rs910605802 Chr.1: 164771729-164771729 on Build GRCh38
rs1030699618 Chr.1: 164771674-164771674 on Build GRCh38
rs540919689 Chr.1: 164771461-164771461 on Build GRCh38
rs535415212 Chr.1: 164771416-164771416 on Build GRCh38
rs886951321 Chr.1: 164771694-164771694 on Build GRCh38
rs1037718203 Chr.1: 164771456-164771456 on Build GRCh38
rs954504944 Chr.1: 164771685-164771685 on Build GRCh38
rs1029579370 Chr.1: 164771389-164771389 on Build GRCh38

View Full Product Details