Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48257340-48257840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD8-AS1
Entrez Gene ID
100505812
Gene Name
CARD8 antisense RNA 1
Gene Aliases
-
UniGene
Hs.614136
Gene Chromosome Location
Chr.19: 48255675-48258194 on Build GRCh38
Transcripts
NR_040599.1

SNP Detail

SNP ID Chromosome Location
rs1009333494 Chr.19: 48257545-48257545 on Build GRCh38
rs983572963 Chr.19: 48257677-48257677 on Build GRCh38
rs887861371 Chr.19: 48257694-48257694 on Build GRCh38
rs1009715117 Chr.19: 48257804-48257804 on Build GRCh38
rs192031621 Chr.19: 48257548-48257548 on Build GRCh38
rs959197009 Chr.19: 48257615-48257615 on Build GRCh38
rs142546434 Chr.19: 48257688-48257688 on Build GRCh38
rs929351987 Chr.19: 48257647-48257647 on Build GRCh38
rs537749264 Chr.19: 48257536-48257536 on Build GRCh38
rs1021018511 Chr.19: 48257546-48257546 on Build GRCh38
rs386810022 Chr.19: 48257414-48257419 on Build GRCh38
rs942071271 Chr.19: 48257695-48257695 on Build GRCh38
rs10610566 Chr.19: 48257416-48257419 on Build GRCh38
rs891989019 Chr.19: 48257777-48257777 on Build GRCh38
rs10412163 Chr.19: 48257633-48257633 on Build GRCh38
rs573663990 Chr.19: 48257778-48257778 on Build GRCh38
rs1033591312 Chr.19: 48257606-48257607 on Build GRCh38
rs900792224 Chr.19: 48257719-48257719 on Build GRCh38
rs956187640 Chr.19: 48257450-48257450 on Build GRCh38
rs7248766 Chr.19: 48257514-48257514 on Build GRCh38
rs970733995 Chr.19: 48257387-48257387 on Build GRCh38
rs796419842 Chr.19: 48257762-48257763 on Build GRCh38
rs997716696 Chr.19: 48257494-48257494 on Build GRCh38
rs140890214 Chr.19: 48257411-48257414 on Build GRCh38
rs767821565 Chr.19: 48257738-48257738 on Build GRCh38
rs766502358 Chr.19: 48257410-48257411 on Build GRCh38
rs774566119 Chr.19: 48257661-48257661 on Build GRCh38
rs759831558 Chr.19: 48257698-48257698 on Build GRCh38
rs993327860 Chr.19: 48257652-48257652 on Build GRCh38
rs10412609 Chr.19: 48257808-48257808 on Build GRCh38
rs369301577 Chr.19: 48257622-48257622 on Build GRCh38
rs7248752 Chr.19: 48257487-48257487 on Build GRCh38
rs1023400484 Chr.19: 48257720-48257720 on Build GRCh38
rs979747705 Chr.19: 48257584-48257584 on Build GRCh38
rs1022926452 Chr.19: 48257383-48257383 on Build GRCh38
rs1051906636 Chr.19: 48257750-48257750 on Build GRCh38
rs991915298 Chr.19: 48257627-48257627 on Build GRCh38
rs1029414149 Chr.19: 48257500-48257500 on Build GRCh38
rs987888707 Chr.19: 48257486-48257486 on Build GRCh38
rs751704580 Chr.19: 48257415-48257416 on Build GRCh38
rs943294920 Chr.19: 48257569-48257569 on Build GRCh38
rs770031128 Chr.19: 48257372-48257372 on Build GRCh38
rs577799125 Chr.19: 48257553-48257553 on Build GRCh38
rs75491588 Chr.19: 48257414-48257414 on Build GRCh38
rs910198384 Chr.19: 48257549-48257549 on Build GRCh38
rs7343088 Chr.19: 48257419-48257419 on Build GRCh38
rs71181664 Chr.19: 48257768-48257769 on Build GRCh38
rs74807407 Chr.19: 48257764-48257764 on Build GRCh38
rs981629264 Chr.19: 48257391-48257391 on Build GRCh38

View Full Product Details