Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88772955-88773520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36BP2
Entrez Gene ID
645784
Gene Name
ankyrin repeat domain 36B pseudogene 2
Gene Aliases
-
UniGene
Hs.105323
Gene Chromosome Location
Chr.2: 88765902-88806596 on Build GRCh38
Transcripts
NR_015424.1

SNP Detail

SNP ID Chromosome Location
rs79210011 Chr.2: 88773412-88773412 on Build GRCh38
rs569357744 Chr.2: 88773332-88773332 on Build GRCh38
rs56056406 Chr.2: 88773130-88773130 on Build GRCh38
rs977800780 Chr.2: 88773227-88773227 on Build GRCh38
rs77269344 Chr.2: 88773398-88773398 on Build GRCh38
rs62157814 Chr.2: 88773275-88773275 on Build GRCh38
rs113861252 Chr.2: 88773387-88773387 on Build GRCh38
rs1024839436 Chr.2: 88773151-88773151 on Build GRCh38
rs76573351 Chr.2: 88773232-88773232 on Build GRCh38
rs994203809 Chr.2: 88773346-88773346 on Build GRCh38
rs76012192 Chr.2: 88773216-88773216 on Build GRCh38
rs566162975 Chr.2: 88773467-88773467 on Build GRCh38
rs60997763 Chr.2: 88773122-88773122 on Build GRCh38
rs897215342 Chr.2: 88773135-88773135 on Build GRCh38
rs78018052 Chr.2: 88773147-88773147 on Build GRCh38
rs994272391 Chr.2: 88773149-88773149 on Build GRCh38
rs62157817 Chr.2: 88773370-88773370 on Build GRCh38
rs335120 Chr.2: 88773214-88773214 on Build GRCh38
rs970578747 Chr.2: 88773173-88773173 on Build GRCh38
rs78655653 Chr.2: 88773128-88773128 on Build GRCh38
rs539969765 Chr.2: 88773473-88773473 on Build GRCh38
rs200033415 Chr.2: 88773016-88773016 on Build GRCh38
rs369820751 Chr.2: 88772993-88772993 on Build GRCh38
rs143967945 Chr.2: 88773121-88773122 on Build GRCh38
rs75841812 Chr.2: 88773230-88773230 on Build GRCh38
rs889443441 Chr.2: 88773323-88773323 on Build GRCh38
rs77608973 Chr.2: 88773031-88773031 on Build GRCh38
rs62157815 Chr.2: 88773338-88773338 on Build GRCh38
rs959257401 Chr.2: 88773436-88773436 on Build GRCh38
rs1003824841 Chr.2: 88773175-88773175 on Build GRCh38
rs1038355945 Chr.2: 88773325-88773325 on Build GRCh38
rs78213287 Chr.2: 88772976-88772976 on Build GRCh38
rs62157816 Chr.2: 88773365-88773365 on Build GRCh38
rs934174495 Chr.2: 88773248-88773248 on Build GRCh38
rs959558661 Chr.2: 88773181-88773181 on Build GRCh38
rs534788774 Chr.2: 88773413-88773413 on Build GRCh38
rs867357079 Chr.2: 88773177-88773177 on Build GRCh38
rs544144445 Chr.2: 88773083-88773083 on Build GRCh38
rs907283450 Chr.2: 88773384-88773384 on Build GRCh38
rs562134935 Chr.2: 88773121-88773121 on Build GRCh38
rs745345394 Chr.2: 88773036-88773037 on Build GRCh38
rs549217344 Chr.2: 88773386-88773386 on Build GRCh38
rs869198697 Chr.2: 88773038-88773039 on Build GRCh38
rs541285049 Chr.2: 88773190-88773190 on Build GRCh38
rs922381624 Chr.2: 88773237-88773237 on Build GRCh38
rs941607290 Chr.2: 88773321-88773321 on Build GRCh38
rs142876015 Chr.2: 88773106-88773107 on Build GRCh38
rs576425832 Chr.2: 88773490-88773490 on Build GRCh38
rs769296957 Chr.2: 88772984-88772987 on Build GRCh38
rs76682228 Chr.2: 88773460-88773460 on Build GRCh38
rs368290073 Chr.2: 88773122-88773123 on Build GRCh38
rs911445419 Chr.2: 88773296-88773296 on Build GRCh38
rs553220251 Chr.2: 88773437-88773437 on Build GRCh38
rs1045812361 Chr.2: 88773347-88773347 on Build GRCh38
rs869296776 Chr.2: 88773107-88773108 on Build GRCh38
rs558987875 Chr.2: 88773198-88773198 on Build GRCh38
rs530704686 Chr.2: 88773369-88773369 on Build GRCh38
rs796990902 Chr.2: 88773040-88773040 on Build GRCh38
rs550939358 Chr.2: 88773249-88773249 on Build GRCh38
rs1033942555 Chr.2: 88773419-88773419 on Build GRCh38
rs894011551 Chr.2: 88773446-88773446 on Build GRCh38
rs200224593 Chr.2: 88773168-88773168 on Build GRCh38
rs890074551 Chr.2: 88773081-88773081 on Build GRCh38

View Full Product Details