Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88782752-88783316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36BP2
Entrez Gene ID
645784
Gene Name
ankyrin repeat domain 36B pseudogene 2
Gene Aliases
-
UniGene
Hs.105323
Gene Chromosome Location
Chr.2: 88765902-88806596 on Build GRCh38
Transcripts
NR_015424.1

SNP Detail

SNP ID Chromosome Location
rs74599148 Chr.2: 88782986-88782986 on Build GRCh38
rs879109435 Chr.2: 88782779-88782779 on Build GRCh38
rs978964144 Chr.2: 88783200-88783200 on Build GRCh38
rs142861467 Chr.2: 88782871-88782871 on Build GRCh38
rs540576533 Chr.2: 88783098-88783098 on Build GRCh38
rs888379400 Chr.2: 88783071-88783071 on Build GRCh38
rs80130259 Chr.2: 88783167-88783167 on Build GRCh38
rs868396956 Chr.2: 88782893-88782893 on Build GRCh38
rs202235353 Chr.2: 88783178-88783178 on Build GRCh38
rs1014416908 Chr.2: 88783182-88783182 on Build GRCh38
rs879572004 Chr.2: 88782899-88782899 on Build GRCh38
rs138526184 Chr.2: 88782798-88782798 on Build GRCh38
rs143191250 Chr.2: 88782862-88782862 on Build GRCh38
rs944728489 Chr.2: 88783282-88783282 on Build GRCh38
rs75619364 Chr.2: 88783185-88783185 on Build GRCh38
rs571708778 Chr.2: 88782987-88782987 on Build GRCh38
rs150412285 Chr.2: 88782823-88782826 on Build GRCh38
rs567361839 Chr.2: 88783156-88783156 on Build GRCh38
rs200826528 Chr.2: 88783089-88783089 on Build GRCh38
rs72930616 Chr.2: 88783213-88783213 on Build GRCh38
rs768425923 Chr.2: 88783264-88783264 on Build GRCh38
rs528476047 Chr.2: 88783173-88783173 on Build GRCh38
rs1039152537 Chr.2: 88783038-88783038 on Build GRCh38
rs570153164 Chr.2: 88782827-88782827 on Build GRCh38
rs959802304 Chr.2: 88782895-88782895 on Build GRCh38
rs149368521 Chr.2: 88783099-88783099 on Build GRCh38
rs78593876 Chr.2: 88782905-88782905 on Build GRCh38
rs553141545 Chr.2: 88782972-88782972 on Build GRCh38
rs530776025 Chr.2: 88782831-88782831 on Build GRCh38
rs557568242 Chr.2: 88783247-88783247 on Build GRCh38
rs140000456 Chr.2: 88782856-88782856 on Build GRCh38
rs567596685 Chr.2: 88782911-88782911 on Build GRCh38
rs374280222 Chr.2: 88782787-88782787 on Build GRCh38
rs532176470 Chr.2: 88783118-88783118 on Build GRCh38
rs369392603 Chr.2: 88782835-88782836 on Build GRCh38
rs546843414 Chr.2: 88783176-88783176 on Build GRCh38
rs544409434 Chr.2: 88783127-88783127 on Build GRCh38
rs143108186 Chr.2: 88783101-88783101 on Build GRCh38
rs926174290 Chr.2: 88783219-88783219 on Build GRCh38
rs140274492 Chr.2: 88782818-88782818 on Build GRCh38
rs575314892 Chr.2: 88783023-88783023 on Build GRCh38
rs956257100 Chr.2: 88783243-88783243 on Build GRCh38
rs535030403 Chr.2: 88782939-88782939 on Build GRCh38
rs899662501 Chr.2: 88783039-88783039 on Build GRCh38
rs979381396 Chr.2: 88782948-88782948 on Build GRCh38
rs1019847257 Chr.2: 88782788-88782788 on Build GRCh38
rs143182368 Chr.2: 88782820-88782820 on Build GRCh38
rs563613088 Chr.2: 88783133-88783133 on Build GRCh38
rs72930610 Chr.2: 88782954-88782954 on Build GRCh38
rs201059577 Chr.2: 88782887-88782887 on Build GRCh38
rs78717019 Chr.2: 88782978-88782978 on Build GRCh38
rs140822233 Chr.2: 88782864-88782864 on Build GRCh38
rs548960759 Chr.2: 88783136-88783136 on Build GRCh38
rs202032514 Chr.2: 88783228-88783228 on Build GRCh38
rs72930612 Chr.2: 88783003-88783003 on Build GRCh38
rs867865153 Chr.2: 88783022-88783022 on Build GRCh38
rs72490036 Chr.2: 88783048-88783048 on Build GRCh38
rs904793806 Chr.2: 88783119-88783119 on Build GRCh38
rs878879942 Chr.2: 88783032-88783032 on Build GRCh38
rs548999288 Chr.2: 88782881-88782881 on Build GRCh38
rs201698437 Chr.2: 88783019-88783019 on Build GRCh38
rs571931362 Chr.2: 88783192-88783192 on Build GRCh38
rs185073653 Chr.2: 88783056-88783056 on Build GRCh38
rs1003812781 Chr.2: 88783075-88783075 on Build GRCh38
rs916053993 Chr.2: 88782934-88782934 on Build GRCh38
rs539226876 Chr.2: 88782990-88782990 on Build GRCh38
rs934617488 Chr.2: 88782980-88782980 on Build GRCh38
rs573173412 Chr.2: 88783061-88783061 on Build GRCh38
rs114008584 Chr.2: 88782882-88782882 on Build GRCh38
rs542630789 Chr.2: 88783037-88783037 on Build GRCh38
rs370959214 Chr.2: 88782779-88782780 on Build GRCh38
rs568839069 Chr.2: 88783248-88783248 on Build GRCh38
rs879153455 Chr.2: 88783035-88783035 on Build GRCh38
rs952444429 Chr.2: 88782873-88782873 on Build GRCh38
rs530937865 Chr.2: 88783134-88783134 on Build GRCh38
rs538823774 Chr.2: 88783215-88783215 on Build GRCh38
rs76106688 Chr.2: 88783290-88783290 on Build GRCh38
rs556753472 Chr.2: 88783002-88783002 on Build GRCh38
rs1015270791 Chr.2: 88782883-88782883 on Build GRCh38

View Full Product Details