Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88791954-88792545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36BP2
Entrez Gene ID
645784
Gene Name
ankyrin repeat domain 36B pseudogene 2
Gene Aliases
-
UniGene
Hs.105323
Gene Chromosome Location
Chr.2: 88765902-88806596 on Build GRCh38
Transcripts
NR_015424.1

SNP Detail

SNP ID Chromosome Location
rs529853187 Chr.2: 88792299-88792299 on Build GRCh38
rs566424714 Chr.2: 88792464-88792464 on Build GRCh38
rs983544908 Chr.2: 88792509-88792509 on Build GRCh38
rs938042258 Chr.2: 88792106-88792106 on Build GRCh38
rs575369950 Chr.2: 88792027-88792027 on Build GRCh38
rs899316943 Chr.2: 88792266-88792266 on Build GRCh38
rs6719377 Chr.2: 88792143-88792143 on Build GRCh38
rs959260874 Chr.2: 88792248-88792248 on Build GRCh38
rs903084787 Chr.2: 88792191-88792191 on Build GRCh38
rs371904711 Chr.2: 88792087-88792087 on Build GRCh38
rs1047936129 Chr.2: 88792019-88792019 on Build GRCh38
rs544026756 Chr.2: 88791995-88791995 on Build GRCh38
rs865910060 Chr.2: 88792336-88792336 on Build GRCh38
rs951730865 Chr.2: 88792499-88792499 on Build GRCh38
rs563420106 Chr.2: 88791999-88791999 on Build GRCh38
rs966838086 Chr.2: 88792508-88792508 on Build GRCh38
rs1027559978 Chr.2: 88792498-88792498 on Build GRCh38
rs893150502 Chr.2: 88792112-88792112 on Build GRCh38
rs575642559 Chr.2: 88792421-88792421 on Build GRCh38
rs966562444 Chr.2: 88792201-88792201 on Build GRCh38
rs915335912 Chr.2: 88792493-88792493 on Build GRCh38
rs796437700 Chr.2: 88792061-88792061 on Build GRCh38
rs907643961 Chr.2: 88792292-88792292 on Build GRCh38
rs932187165 Chr.2: 88792300-88792300 on Build GRCh38
rs532723215 Chr.2: 88792262-88792262 on Build GRCh38
rs1054431300 Chr.2: 88792109-88792109 on Build GRCh38
rs893134482 Chr.2: 88792309-88792309 on Build GRCh38
rs1003816640 Chr.2: 88792218-88792218 on Build GRCh38
rs1020828743 Chr.2: 88792187-88792187 on Build GRCh38
rs959395514 Chr.2: 88792381-88792381 on Build GRCh38
rs771105264 Chr.2: 88792177-88792178 on Build GRCh38
rs201139265 Chr.2: 88792349-88792349 on Build GRCh38
rs944873146 Chr.2: 88792293-88792293 on Build GRCh38
rs755041207 Chr.2: 88792358-88792358 on Build GRCh38
rs181200718 Chr.2: 88791997-88791997 on Build GRCh38
rs914405019 Chr.2: 88792149-88792149 on Build GRCh38
rs1002979241 Chr.2: 88792360-88792360 on Build GRCh38
rs900777747 Chr.2: 88792056-88792056 on Build GRCh38
rs568956855 Chr.2: 88792278-88792278 on Build GRCh38
rs184591596 Chr.2: 88792268-88792268 on Build GRCh38
rs1034883700 Chr.2: 88792230-88792230 on Build GRCh38
rs1054715394 Chr.2: 88792303-88792303 on Build GRCh38
rs1028883513 Chr.2: 88792288-88792288 on Build GRCh38
rs1012113670 Chr.2: 88792475-88792475 on Build GRCh38
rs894992807 Chr.2: 88792449-88792449 on Build GRCh38
rs1010282816 Chr.2: 88792168-88792168 on Build GRCh38
rs1041792878 Chr.2: 88792348-88792348 on Build GRCh38
rs1040539943 Chr.2: 88792298-88792298 on Build GRCh38
rs1010687801 Chr.2: 88792329-88792329 on Build GRCh38
rs542602824 Chr.2: 88792034-88792034 on Build GRCh38
rs553863949 Chr.2: 88792060-88792061 on Build GRCh38
rs996448795 Chr.2: 88792064-88792064 on Build GRCh38
rs6719466 Chr.2: 88792158-88792158 on Build GRCh38
rs866166300 Chr.2: 88792020-88792020 on Build GRCh38
rs990713591 Chr.2: 88792251-88792251 on Build GRCh38
rs970939161 Chr.2: 88792345-88792345 on Build GRCh38
rs565454429 Chr.2: 88792240-88792240 on Build GRCh38
rs1034790317 Chr.2: 88792426-88792426 on Build GRCh38
rs1037888276 Chr.2: 88792258-88792258 on Build GRCh38
rs536790329 Chr.2: 88792269-88792269 on Build GRCh38
rs983211228 Chr.2: 88792283-88792283 on Build GRCh38

View Full Product Details