Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122456401-122456554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2
Entrez Gene ID
3558
Gene Name
interleukin 2
Gene Aliases
IL-2, TCGF, lymphokine
UniGene
Hs.89679
Gene Chromosome Location
Chr.4: 122449479-122456495 on Build GRCh38
Transcripts
NM_000586.3 XM_017008177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
342475 c.9G>C p.R3S Chr.4: 122456432-122456432 on Build GRCh38
7203338 c.38G>A p.S13N Chr.4: 122456403-122456403 on Build GRCh38
7191950 c.4T>C p.Y2H Chr.4: 122456437-122456437 on Build GRCh38
373664 c.31G>T p.A11S Chr.4: 122456410-122456410 on Build GRCh38
6545749 c.18C>A p.L6L Chr.4: 122456423-122456423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780645947 Chr.4: 122456452-122456452 on Build GRCh38
rs912291339 Chr.4: 122456511-122456511 on Build GRCh38
rs778990655 Chr.4: 122456439-122456439 on Build GRCh38
rs76833084 Chr.4: 122456450-122456450 on Build GRCh38
rs757619199 Chr.4: 122456430-122456430 on Build GRCh38
rs932272254 Chr.4: 122456494-122456494 on Build GRCh38
rs772015543 Chr.4: 122456451-122456451 on Build GRCh38
rs76468545 Chr.4: 122456480-122456480 on Build GRCh38
rs750727213 Chr.4: 122456446-122456446 on Build GRCh38
rs200067612 Chr.4: 122456467-122456467 on Build GRCh38
rs547165010 Chr.4: 122456509-122456509 on Build GRCh38
rs979661963 Chr.4: 122456470-122456470 on Build GRCh38
rs1049865826 Chr.4: 122456498-122456498 on Build GRCh38
rs763308059 Chr.4: 122456489-122456489 on Build GRCh38
rs372276938 Chr.4: 122456484-122456484 on Build GRCh38

View Full Product Details