Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55082430-55082602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDR
Entrez Gene ID
3791
Gene Name
kinase insert domain receptor
Gene Aliases
CD309, FLK1, VEGFR, VEGFR2
UniGene
Hs.479756
Gene Chromosome Location
Chr.4: 55078259-55125595 on Build GRCh38
Transcripts
NM_002253.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365817 c.3700G>T p.V1234L Chr.4: 55082598-55082598 on Build GRCh38
1579010 c.3762+9G>A p.? Chr.4: 55082527-55082527 on Build GRCh38
4605220 c.3707T>A p.V1236E Chr.4: 55082591-55082591 on Build GRCh38
108688 c.3728C>T p.P1243L Chr.4: 55082570-55082570 on Build GRCh38
3781167 c.3746T>A p.V1249E Chr.4: 55082552-55082552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142120845 Chr.4: 55082541-55082541 on Build GRCh38
rs772987575 Chr.4: 55082505-55082505 on Build GRCh38
rs765747186 Chr.4: 55082538-55082538 on Build GRCh38
rs894977242 Chr.4: 55082542-55082542 on Build GRCh38
rs781436645 Chr.4: 55082569-55082569 on Build GRCh38
rs111307973 Chr.4: 55082543-55082543 on Build GRCh38
rs751858962 Chr.4: 55082563-55082563 on Build GRCh38
rs150847930 Chr.4: 55082570-55082570 on Build GRCh38
rs757443823 Chr.4: 55082568-55082568 on Build GRCh38
rs547194966 Chr.4: 55082497-55082497 on Build GRCh38
rs530260334 Chr.4: 55082486-55082486 on Build GRCh38
rs150116423 Chr.4: 55082547-55082547 on Build GRCh38
rs78801899 Chr.4: 55082528-55082528 on Build GRCh38
rs778078301 Chr.4: 55082559-55082559 on Build GRCh38
rs760219983 Chr.4: 55082506-55082506 on Build GRCh38
rs902199058 Chr.4: 55082511-55082511 on Build GRCh38
rs745420505 Chr.4: 55082521-55082521 on Build GRCh38
rs1029908959 Chr.4: 55082460-55082460 on Build GRCh38
rs374982964 Chr.4: 55082527-55082527 on Build GRCh38

View Full Product Details