Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48410066-48410190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK8
Entrez Gene ID
5599
Gene Name
mitogen-activated protein kinase 8
Gene Aliases
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
UniGene
Hs.138211
Gene Chromosome Location
Chr.10: 48306639-48439360 on Build GRCh38
Transcripts
NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5733518 c.424C>T p.L142F Chr.10: 48410142-48410142 on Build GRCh38
684192 c.445C>T p.H149Y Chr.10: 48410163-48410163 on Build GRCh38
48880 c.358C>T p.Q120* Chr.10: 48410076-48410076 on Build GRCh38
6461174 c.428A>T p.H143L Chr.10: 48410146-48410146 on Build GRCh38
427597 c.448C>T p.R150W Chr.10: 48410166-48410166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969491000 Chr.10: 48410126-48410126 on Build GRCh38
rs771284001 Chr.10: 48410108-48410108 on Build GRCh38
rs779380684 Chr.10: 48410152-48410152 on Build GRCh38
rs749668518 Chr.10: 48410105-48410105 on Build GRCh38

View Full Product Details