Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48404903-48405028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK8
Entrez Gene ID
5599
Gene Name
mitogen-activated protein kinase 8
Gene Aliases
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
UniGene
Hs.138211
Gene Chromosome Location
Chr.10: 48306639-48439360 on Build GRCh38
Transcripts
NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7273664 c.239T>C p.V80A Chr.10: 48404968-48404968 on Build GRCh38
7273663 c.239T>C p.V80A Chr.10: 48404968-48404968 on Build GRCh38
1732626 c.192delG p.T65fs*10 Chr.10: 48404921-48404921 on Build GRCh38
1257320 c.205C>T p.R69W Chr.10: 48404934-48404934 on Build GRCh38
4994635 c.175C>T p.R59* Chr.10: 48404904-48404904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770119133 Chr.10: 48404992-48404992 on Build GRCh38
rs998000637 Chr.10: 48404985-48404985 on Build GRCh38
rs773584378 Chr.10: 48404990-48404990 on Build GRCh38
rs763551769 Chr.10: 48404991-48404991 on Build GRCh38

View Full Product Details