Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204425585-204425714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3C2B
Entrez Gene ID
5287
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta
Gene Aliases
C2-PI3K
UniGene
Hs.497487
Gene Chromosome Location
Chr.1: 204422630-204494815 on Build GRCh38
Transcripts
NM_002646.3 XM_005245257.2 XM_005245258.3 XM_011509630.2 XM_011509631.2 XM_017001473.1 XM_017001474.1 XM_017001475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775130 c.4685G>A p.R1562Q Chr.1: 204425644-204425644 on Build GRCh38
6746340 c.4649C>A p.P1550H Chr.1: 204425680-204425680 on Build GRCh38
4495911 c.4638C>T p.L1546L Chr.1: 204425691-204425691 on Build GRCh38
1689665 c.4640T>G p.L1547R Chr.1: 204425689-204425689 on Build GRCh38
902591 c.4632T>G p.I1544M Chr.1: 204425697-204425697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533581042 Chr.1: 204425623-204425623 on Build GRCh38
rs546949370 Chr.1: 204425681-204425681 on Build GRCh38
rs765527812 Chr.1: 204425649-204425649 on Build GRCh38
rs143735929 Chr.1: 204425632-204425632 on Build GRCh38
rs753176809 Chr.1: 204425665-204425665 on Build GRCh38
rs111728676 Chr.1: 204425669-204425669 on Build GRCh38
rs984827733 Chr.1: 204425670-204425670 on Build GRCh38
rs754138397 Chr.1: 204425644-204425644 on Build GRCh38
rs150308890 Chr.1: 204425628-204425628 on Build GRCh38
rs554907249 Chr.1: 204425658-204425658 on Build GRCh38
rs371494234 Chr.1: 204425645-204425645 on Build GRCh38

View Full Product Details