Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154542311-154542536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PD
Entrez Gene ID
2539
Gene Name
glucose-6-phosphate dehydrogenase
Gene Aliases
G6PD1
UniGene
Hs.461047
Gene Chromosome Location
Chr.X: 154531390-154547586 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2 NM_001099856.4 NM_001321396.1

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
UniGene
Hs.43505
Gene Chromosome Location
Chr.X: 154542240-154565046 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2 NM_001099856.4 NM_001321396.1

SNP Detail

SNP ID Chromosome Location
rs782789442 Chr.X: 154542343-154542343 on Build GRCh38
rs189237568 Chr.X: 154542411-154542411 on Build GRCh38
rs373355350 Chr.X: 154542477-154542477 on Build GRCh38
rs782457761 Chr.X: 154542501-154542501 on Build GRCh38
rs1040534131 Chr.X: 154542511-154542511 on Build GRCh38
rs782393436 Chr.X: 154542446-154542446 on Build GRCh38
rs368857323 Chr.X: 154542419-154542419 on Build GRCh38
rs782519851 Chr.X: 154542359-154542359 on Build GRCh38
rs782337273 Chr.X: 154542470-154542470 on Build GRCh38
rs781835485 Chr.X: 154542358-154542358 on Build GRCh38
rs930601658 Chr.X: 154542463-154542463 on Build GRCh38
rs371427128 Chr.X: 154542367-154542367 on Build GRCh38
rs782142125 Chr.X: 154542436-154542436 on Build GRCh38
rs782487928 Chr.X: 154542424-154542424 on Build GRCh38
rs11544439 Chr.X: 154542396-154542396 on Build GRCh38
rs782201526 Chr.X: 154542412-154542412 on Build GRCh38
rs782724801 Chr.X: 154542448-154542448 on Build GRCh38
rs782633369 Chr.X: 154542480-154542480 on Build GRCh38
rs781823051 Chr.X: 154542485-154542485 on Build GRCh38
rs782545516 Chr.X: 154542375-154542375 on Build GRCh38
rs782367664 Chr.X: 154542390-154542390 on Build GRCh38
rs782763580 Chr.X: 154542348-154542348 on Build GRCh38
rs889334371 Chr.X: 154542476-154542476 on Build GRCh38
rs920612647 Chr.X: 154542450-154542450 on Build GRCh38
rs782052842 Chr.X: 154542453-154542454 on Build GRCh38
rs201460468 Chr.X: 154542374-154542374 on Build GRCh38
rs781801725 Chr.X: 154542462-154542462 on Build GRCh38
rs782589734 Chr.X: 154542362-154542362 on Build GRCh38
rs782253817 Chr.X: 154542376-154542376 on Build GRCh38

View Full Product Details