Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122426882-122427147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
UniGene
Hs.161008
Gene Chromosome Location
Chr.3: 122421901-122514939 on Build GRCh38
Transcripts
NM_002264.3 NR_026698.1 XM_005247437.3 XM_005247439.3 XM_011512795.2 XM_011512797.2 XM_017006363.1 XM_017006364.1 XM_017006365.1 NR_125405.1

Gene Symbol
LOC102723582
Entrez Gene ID
102723582
Gene Name
uncharacterized LOC102723582
Gene Aliases
-
UniGene
Hs.675056
Gene Chromosome Location
Chr.3: 122416206-122443247 on Build GRCh38
Transcripts
NM_002264.3 NR_026698.1 XM_005247437.3 XM_005247439.3 XM_011512795.2 XM_011512797.2 XM_017006363.1 XM_017006364.1 XM_017006365.1 NR_125405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6697638 c.1518C>T p.T506T Chr.3: 122427084-122427084 on Build GRCh38
1616798 c.1481A>G p.Q494R Chr.3: 122427121-122427121 on Build GRCh38
3372982 c.1471G>A p.E491K Chr.3: 122427131-122427131 on Build GRCh38
5964051 c.1462G>A p.E488K Chr.3: 122427140-122427140 on Build GRCh38
1418268 c.1468C>T p.Q490* Chr.3: 122427134-122427134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746213432 Chr.3: 122427035-122427035 on Build GRCh38
rs980426413 Chr.3: 122426936-122426936 on Build GRCh38
rs368205703 Chr.3: 122426949-122426949 on Build GRCh38
rs143005503 Chr.3: 122427090-122427090 on Build GRCh38
rs15660 Chr.3: 122427008-122427008 on Build GRCh38
rs768710154 Chr.3: 122426960-122426960 on Build GRCh38
rs774261626 Chr.3: 122426961-122426961 on Build GRCh38
rs773781619 Chr.3: 122427087-122427087 on Build GRCh38
rs939941823 Chr.3: 122427042-122427042 on Build GRCh38
rs147574538 Chr.3: 122427063-122427063 on Build GRCh38
rs4677950 Chr.3: 122426963-122426963 on Build GRCh38
rs768068658 Chr.3: 122427065-122427065 on Build GRCh38
rs765354666 Chr.3: 122426967-122426967 on Build GRCh38
rs573159535 Chr.3: 122427054-122427054 on Build GRCh38
rs967746673 Chr.3: 122426932-122426932 on Build GRCh38
rs755834808 Chr.3: 122427045-122427045 on Build GRCh38
rs769282869 Chr.3: 122426948-122426948 on Build GRCh38
rs553380334 Chr.3: 122427029-122427029 on Build GRCh38
rs771945739 Chr.3: 122427110-122427110 on Build GRCh38
rs752118749 Chr.3: 122427011-122427011 on Build GRCh38
rs1021973965 Chr.3: 122426926-122426926 on Build GRCh38
rs745549838 Chr.3: 122426939-122426939 on Build GRCh38
rs762342906 Chr.3: 122426950-122426950 on Build GRCh38
rs200609980 Chr.3: 122427005-122427005 on Build GRCh38
rs969349638 Chr.3: 122427071-122427071 on Build GRCh38
rs781707034 Chr.3: 122427033-122427033 on Build GRCh38
rs758401352 Chr.3: 122426980-122426980 on Build GRCh38
rs773037206 Chr.3: 122427114-122427114 on Build GRCh38
rs149030683 Chr.3: 122427084-122427084 on Build GRCh38
rs368341068 Chr.3: 122426965-122426965 on Build GRCh38
rs1009790038 Chr.3: 122426912-122426912 on Build GRCh38
rs773782135 Chr.3: 122426940-122426941 on Build GRCh38
rs759822428 Chr.3: 122426966-122426966 on Build GRCh38
rs926394854 Chr.3: 122426937-122426937 on Build GRCh38
rs752861909 Chr.3: 122426970-122426970 on Build GRCh38

View Full Product Details