Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41352693-41352960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFB1
Entrez Gene ID
7040
Gene Name
transforming growth factor beta 1
Gene Aliases
CED, DPD1, LAP, TGFB, TGFbeta
UniGene
Hs.645227
Gene Chromosome Location
Chr.19: 41330531-41353933 on Build GRCh38
Transcripts
NM_000660.5 XM_011527242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681056 c.307T>C p.Y103H Chr.19: 41352738-41352738 on Build GRCh38
6999140 c.225G>T p.E75D Chr.19: 41352820-41352820 on Build GRCh38
3952972 c.99C>T p.C33C Chr.19: 41352946-41352946 on Build GRCh38
1481113 c.85G>A p.G29R Chr.19: 41352960-41352960 on Build GRCh38
5987372 c.223G>A p.E75K Chr.19: 41352822-41352822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137864931 Chr.19: 41352745-41352745 on Build GRCh38
rs199704558 Chr.19: 41352793-41352793 on Build GRCh38
rs913576317 Chr.19: 41352856-41352856 on Build GRCh38
rs201283333 Chr.19: 41352867-41352867 on Build GRCh38
rs748104908 Chr.19: 41352790-41352790 on Build GRCh38
rs200512008 Chr.19: 41352759-41352759 on Build GRCh38
rs13447341 Chr.19: 41352905-41352905 on Build GRCh38
rs751217033 Chr.19: 41352933-41352933 on Build GRCh38
rs986504298 Chr.19: 41352715-41352715 on Build GRCh38
rs938698778 Chr.19: 41352882-41352882 on Build GRCh38
rs764491141 Chr.19: 41352886-41352886 on Build GRCh38
rs950133017 Chr.19: 41352829-41352829 on Build GRCh38
rs751073303 Chr.19: 41352785-41352785 on Build GRCh38
rs745931726 Chr.19: 41352839-41352839 on Build GRCh38
rs754483500 Chr.19: 41352787-41352787 on Build GRCh38
rs201155269 Chr.19: 41352789-41352789 on Build GRCh38
rs777118195 Chr.19: 41352887-41352887 on Build GRCh38
rs750211892 Chr.19: 41352856-41352856 on Build GRCh38
rs149455461 Chr.19: 41352823-41352823 on Build GRCh38
rs777547933 Chr.19: 41352797-41352797 on Build GRCh38
rs111033611 Chr.19: 41352804-41352804 on Build GRCh38
rs145953838 Chr.19: 41352870-41352870 on Build GRCh38
rs945084245 Chr.19: 41352864-41352864 on Build GRCh38
rs762331262 Chr.19: 41352902-41352902 on Build GRCh38
rs766021098 Chr.19: 41352915-41352915 on Build GRCh38
rs1053427155 Chr.19: 41352842-41352842 on Build GRCh38
rs774505337 Chr.19: 41352830-41352830 on Build GRCh38
rs772095480 Chr.19: 41352847-41352847 on Build GRCh38
rs771172130 Chr.19: 41352817-41352817 on Build GRCh38
rs200230522 Chr.19: 41352834-41352834 on Build GRCh38
rs753186435 Chr.19: 41352750-41352750 on Build GRCh38
rs551552217 Chr.19: 41352937-41352937 on Build GRCh38
rs533204331 Chr.19: 41352854-41352854 on Build GRCh38
rs371589941 Chr.19: 41352779-41352779 on Build GRCh38
rs866580372 Chr.19: 41352743-41352743 on Build GRCh38
rs1017621656 Chr.19: 41352923-41352923 on Build GRCh38
rs761100949 Chr.19: 41352753-41352753 on Build GRCh38
rs758523150 Chr.19: 41352932-41352932 on Build GRCh38
rs942309616 Chr.19: 41352719-41352719 on Build GRCh38
rs910883327 Chr.19: 41352716-41352716 on Build GRCh38
rs199592796 Chr.19: 41352805-41352805 on Build GRCh38
rs147873920 Chr.19: 41352754-41352754 on Build GRCh38
rs759592536 Chr.19: 41352739-41352739 on Build GRCh38
rs774502862 Chr.19: 41352724-41352724 on Build GRCh38
rs758132393 Chr.19: 41352760-41352760 on Build GRCh38

View Full Product Details