Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204424723-204424972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3C2B
Entrez Gene ID
5287
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta
Gene Aliases
C2-PI3K
UniGene
Hs.497487
Gene Chromosome Location
Chr.1: 204422630-204494815 on Build GRCh38
Transcripts
NM_002646.3 XM_005245257.2 XM_005245258.3 XM_011509630.2 XM_011509631.2 XM_017001473.1 XM_017001474.1 XM_017001475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143253 c.4870T>G p.F1624V Chr.1: 204424887-204424887 on Build GRCh38
25371 c.4904G>C p.*1635S Chr.1: 204424853-204424853 on Build GRCh38
6358079 c.4829G>A p.R1610H Chr.1: 204424928-204424928 on Build GRCh38
5019460 c.4803C>T p.N1601N Chr.1: 204424954-204424954 on Build GRCh38
902590 c.4875C>A p.A1625A Chr.1: 204424882-204424882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754750718 Chr.1: 204424912-204424912 on Build GRCh38
rs746061962 Chr.1: 204424868-204424868 on Build GRCh38
rs376434764 Chr.1: 204424872-204424872 on Build GRCh38
rs570618115 Chr.1: 204424781-204424781 on Build GRCh38
rs921724766 Chr.1: 204424779-204424779 on Build GRCh38
rs1039763818 Chr.1: 204424841-204424841 on Build GRCh38
rs114464486 Chr.1: 204424894-204424894 on Build GRCh38
rs753566759 Chr.1: 204424907-204424907 on Build GRCh38
rs747145855 Chr.1: 204424848-204424848 on Build GRCh38
rs1040606553 Chr.1: 204424823-204424823 on Build GRCh38
rs767025111 Chr.1: 204424893-204424893 on Build GRCh38
rs61758020 Chr.1: 204424895-204424895 on Build GRCh38
rs575066961 Chr.1: 204424786-204424786 on Build GRCh38
rs907539091 Chr.1: 204424765-204424765 on Build GRCh38
rs373485341 Chr.1: 204424923-204424923 on Build GRCh38
rs1014880202 Chr.1: 204424914-204424914 on Build GRCh38
rs780418310 Chr.1: 204424933-204424933 on Build GRCh38
rs752647243 Chr.1: 204424922-204424922 on Build GRCh38
rs4403681 Chr.1: 204424769-204424769 on Build GRCh38
rs765203344 Chr.1: 204424918-204424918 on Build GRCh38
rs61758019 Chr.1: 204424945-204424945 on Build GRCh38
rs914721347 Chr.1: 204424817-204424817 on Build GRCh38
rs757335746 Chr.1: 204424861-204424861 on Build GRCh38
rs747003072 Chr.1: 204424929-204424929 on Build GRCh38
rs115140808 Chr.1: 204424953-204424953 on Build GRCh38
rs774986225 Chr.1: 204424887-204424887 on Build GRCh38
rs776170355 Chr.1: 204424750-204424750 on Build GRCh38
rs770160575 Chr.1: 204424749-204424749 on Build GRCh38
rs930769846 Chr.1: 204424795-204424795 on Build GRCh38
rs919360467 Chr.1: 204424785-204424785 on Build GRCh38
rs370055028 Chr.1: 204424819-204424819 on Build GRCh38
rs138229960 Chr.1: 204424897-204424897 on Build GRCh38
rs139295851 Chr.1: 204424950-204424950 on Build GRCh38
rs931709321 Chr.1: 204424791-204424791 on Build GRCh38
rs375764799 Chr.1: 204424815-204424815 on Build GRCh38
rs147192711 Chr.1: 204424911-204424911 on Build GRCh38
rs754855079 Chr.1: 204424824-204424824 on Build GRCh38
rs749816934 Chr.1: 204424874-204424874 on Build GRCh38
rs373230099 Chr.1: 204424865-204424865 on Build GRCh38
rs771774949 Chr.1: 204424759-204424759 on Build GRCh38
rs193298214 Chr.1: 204424928-204424928 on Build GRCh38
rs921569316 Chr.1: 204424930-204424930 on Build GRCh38
rs778814155 Chr.1: 204424832-204424832 on Build GRCh38
rs577562908 Chr.1: 204424885-204424885 on Build GRCh38
rs17847769 Chr.1: 204424792-204424792 on Build GRCh38
rs761280372 Chr.1: 204424892-204424892 on Build GRCh38
rs773931352 Chr.1: 204424763-204424763 on Build GRCh38
rs976149218 Chr.1: 204424778-204424778 on Build GRCh38
rs969286266 Chr.1: 204424908-204424908 on Build GRCh38
rs535230764 Chr.1: 204424774-204424774 on Build GRCh38
rs201442246 Chr.1: 204424804-204424804 on Build GRCh38
rs770075841 Chr.1: 204424871-204424871 on Build GRCh38
rs763503821 Chr.1: 204424751-204424751 on Build GRCh38

View Full Product Details