Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 41224667-41224932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
UniGene
Hs.476018
Gene Chromosome Location
Chr.3: 41199451-41240448 on Build GRCh38
Transcripts
NM_001098209.1 NM_001098210.1 NM_001904.3 XM_005264886.2 XM_006712983.1 XM_006712984.1 XM_006712985.1 XM_017005738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6941850 c.102_416del515 p.? Chr.3: 41224614-41225128 on Build GRCh38
6035 c.14_430del417 p.A5_Q143del Chr.3: 41224526-41225142 on Build GRCh38
24830 c.65_193del129 p.V22_Y64del Chr.3: 41224577-41224705 on Build GRCh38
6050509 c.14-21_242-57del393 p.? Chr.3: 41224505-41224897 on Build GRCh38
5707 c.160G>A p.E54K Chr.3: 41224672-41224672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375741392 Chr.3: 41224773-41224773 on Build GRCh38
rs747117715 Chr.3: 41224844-41224844 on Build GRCh38
rs755954409 Chr.3: 41224803-41224803 on Build GRCh38
rs745872861 Chr.3: 41224757-41224758 on Build GRCh38
rs770877302 Chr.3: 41224886-41224886 on Build GRCh38
rs774357411 Chr.3: 41224780-41224780 on Build GRCh38
rs149603705 Chr.3: 41224856-41224856 on Build GRCh38
rs764119976 Chr.3: 41224906-41224906 on Build GRCh38
rs564912197 Chr.3: 41224794-41224794 on Build GRCh38
rs113120762 Chr.3: 41224749-41224749 on Build GRCh38
rs886041553 Chr.3: 41224710-41224710 on Build GRCh38
rs748083020 Chr.3: 41224697-41224699 on Build GRCh38
rs371769678 Chr.3: 41224759-41224759 on Build GRCh38
rs752653613 Chr.3: 41224797-41224797 on Build GRCh38
rs909859735 Chr.3: 41224903-41224903 on Build GRCh38
rs759321007 Chr.3: 41224784-41224784 on Build GRCh38
rs183594226 Chr.3: 41224810-41224810 on Build GRCh38
rs199795087 Chr.3: 41224767-41224767 on Build GRCh38
rs997117183 Chr.3: 41224787-41224787 on Build GRCh38
rs953594154 Chr.3: 41224821-41224821 on Build GRCh38
rs993000676 Chr.3: 41224781-41224781 on Build GRCh38
rs138323374 Chr.3: 41224845-41224845 on Build GRCh38

View Full Product Details