Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48211811-48212044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
UniGene
Hs.446146
Gene Chromosome Location
Chr.19: 48203148-48256269 on Build GRCh38
Transcripts
NM_001184900.1 NM_001184901.1 NM_001184902.1 NM_001184903.1 NM_014959.3 NR_033678.1 NR_033679.1 NR_033680.1 XM_006723091.3 XM_006723092.3 XM_006723093.3 XM_006723094.3 XM_006723095.3 XM_006723096.3 XM_006723097.3 XM_006723098.3 XM_006723099.3 XM_006723101.2 XM_006723102.3 XM_006723104.3 XM_006723106.3 XM_006723107.3 XM_006723109.3 XM_011526641.2 XM_011526642.2 XM_011526643.2 XM_011526644.2 XM_011526646.2 XM_011526647.2 XM_011526648.2 XM_011526650.2 XM_011526653.2 XM_017026482.1 XM_017026483.1 XM_017026484.1 XM_017026485.1 XM_017026486.1 XM_017026487.1 XM_017026488.1 XM_017026489.1 XM_017026490.1 XM_017026492.1 XM_017026493.1 XM_017026494.1 XM_017026495.1 XM_017026496.1 XM_017026497.1 XM_017026498.1 XM_017026499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274779 c.1431C>A p.D477E Chr.19: 48211893-48211893 on Build GRCh38
1729866 c.1496A>G p.N499S Chr.19: 48211828-48211828 on Build GRCh38
6274780 c.1113C>A p.D371E Chr.19: 48211893-48211893 on Build GRCh38
4669254 c.1150G>A p.E384K Chr.19: 48211856-48211856 on Build GRCh38
3107029 c.1484G>A p.R495Q Chr.19: 48211840-48211840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754980203 Chr.19: 48211985-48211986 on Build GRCh38
rs149162452 Chr.19: 48211840-48211840 on Build GRCh38
rs3745718 Chr.19: 48211896-48211896 on Build GRCh38
rs777814650 Chr.19: 48212000-48212000 on Build GRCh38
rs754890431 Chr.19: 48211866-48211866 on Build GRCh38
rs780016656 Chr.19: 48211866-48211868 on Build GRCh38
rs1013415417 Chr.19: 48211868-48211868 on Build GRCh38
rs745911441 Chr.19: 48211907-48211907 on Build GRCh38
rs751317554 Chr.19: 48211956-48211956 on Build GRCh38
rs139483339 Chr.19: 48211950-48211950 on Build GRCh38
rs532121024 Chr.19: 48212000-48212001 on Build GRCh38
rs756028048 Chr.19: 48211987-48211987 on Build GRCh38
rs776993799 Chr.19: 48211935-48211935 on Build GRCh38
rs755632223 Chr.19: 48211929-48211929 on Build GRCh38
rs141801097 Chr.19: 48211948-48211948 on Build GRCh38
rs573785078 Chr.19: 48211893-48211893 on Build GRCh38
rs138779699 Chr.19: 48211875-48211875 on Build GRCh38
rs939770324 Chr.19: 48211979-48211979 on Build GRCh38
rs61755273 Chr.19: 48211908-48211908 on Build GRCh38
rs371795173 Chr.19: 48211927-48211927 on Build GRCh38
rs781123298 Chr.19: 48211882-48211882 on Build GRCh38
rs1032324679 Chr.19: 48211913-48211913 on Build GRCh38
rs770291118 Chr.19: 48211947-48211947 on Build GRCh38
rs756075950 Chr.19: 48211841-48211842 on Build GRCh38
rs749211389 Chr.19: 48211972-48211972 on Build GRCh38
rs1024589650 Chr.19: 48211906-48211906 on Build GRCh38
rs146199265 Chr.19: 48211859-48211859 on Build GRCh38
rs767222046 Chr.19: 48211969-48211969 on Build GRCh38
rs764409113 Chr.19: 48211831-48211831 on Build GRCh38
rs766186900 Chr.19: 48211862-48211862 on Build GRCh38
rs375457501 Chr.19: 48211981-48211981 on Build GRCh38
rs753686618 Chr.19: 48212008-48212008 on Build GRCh38
rs780293112 Chr.19: 48211914-48211914 on Build GRCh38
rs748050357 Chr.19: 48211890-48211890 on Build GRCh38
rs749876926 Chr.19: 48211856-48211856 on Build GRCh38
rs747242060 Chr.19: 48212011-48212011 on Build GRCh38
rs778506041 Chr.19: 48212001-48212001 on Build GRCh38
rs758539025 Chr.19: 48212010-48212010 on Build GRCh38
rs999862762 Chr.19: 48211837-48211837 on Build GRCh38
rs764663963 Chr.19: 48211848-48211848 on Build GRCh38
rs34632751 Chr.19: 48211939-48211939 on Build GRCh38
rs769063453 Chr.19: 48211934-48211934 on Build GRCh38
rs150504639 Chr.19: 48211949-48211949 on Build GRCh38
rs781437480 Chr.19: 48212014-48212014 on Build GRCh38
rs2008521 Chr.19: 48212012-48212012 on Build GRCh38
rs751229617 Chr.19: 48211863-48211863 on Build GRCh38
rs188716648 Chr.19: 48211965-48211965 on Build GRCh38
rs146657505 Chr.19: 48211841-48211841 on Build GRCh38
rs987648875 Chr.19: 48211964-48211964 on Build GRCh38

View Full Product Details