Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122427561-122427794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
UniGene
Hs.161008
Gene Chromosome Location
Chr.3: 122421901-122514939 on Build GRCh38
Transcripts
NM_002264.3 NR_026698.1 XM_005247437.3 XM_005247439.3 XM_011512795.2 XM_011512797.2 XM_017006363.1 XM_017006364.1 XM_017006365.1 NR_125405.1

Gene Symbol
LOC102723582
Entrez Gene ID
102723582
Gene Name
uncharacterized LOC102723582
Gene Aliases
-
UniGene
Hs.675056
Gene Chromosome Location
Chr.3: 122416206-122443247 on Build GRCh38
Transcripts
NM_002264.3 NR_026698.1 XM_005247437.3 XM_005247439.3 XM_011512795.2 XM_011512797.2 XM_017006363.1 XM_017006364.1 XM_017006365.1 NR_125405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5708445 c.1390G>A p.G464S Chr.3: 122427577-122427577 on Build GRCh38
5736727 c.1352T>C p.L451P Chr.3: 122427615-122427615 on Build GRCh38
6588936 c.1275C>G p.I425M Chr.3: 122427692-122427692 on Build GRCh38
3845988 c.1311T>C p.S437S Chr.3: 122427656-122427656 on Build GRCh38
4922309 c.1256T>C p.L419P Chr.3: 122427711-122427711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766681413 Chr.3: 122427687-122427687 on Build GRCh38
rs778503343 Chr.3: 122427714-122427714 on Build GRCh38
rs766984619 Chr.3: 122427722-122427723 on Build GRCh38
rs780898937 Chr.3: 122427746-122427746 on Build GRCh38
rs774614394 Chr.3: 122427604-122427604 on Build GRCh38
rs898848496 Chr.3: 122427703-122427703 on Build GRCh38
rs761983700 Chr.3: 122427659-122427659 on Build GRCh38
rs768198808 Chr.3: 122427663-122427663 on Build GRCh38
rs752589438 Chr.3: 122427750-122427751 on Build GRCh38
rs750911948 Chr.3: 122427668-122427668 on Build GRCh38
rs757888671 Chr.3: 122427732-122427732 on Build GRCh38
rs905020898 Chr.3: 122427755-122427755 on Build GRCh38
rs201121795 Chr.3: 122427716-122427716 on Build GRCh38
rs746924130 Chr.3: 122427745-122427745 on Build GRCh38
rs994581033 Chr.3: 122427739-122427739 on Build GRCh38
rs754671094 Chr.3: 122427700-122427700 on Build GRCh38
rs367980953 Chr.3: 122427721-122427721 on Build GRCh38
rs754178190 Chr.3: 122427697-122427697 on Build GRCh38
rs369415466 Chr.3: 122427669-122427669 on Build GRCh38
rs758999186 Chr.3: 122427718-122427719 on Build GRCh38
rs777206966 Chr.3: 122427737-122427737 on Build GRCh38
rs756132487 Chr.3: 122427750-122427755 on Build GRCh38
rs1036004077 Chr.3: 122427686-122427686 on Build GRCh38

View Full Product Details