Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 141427777-141428002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A3
Entrez Gene ID
11156
Gene Name
protein tyrosine phosphatase type IVA, member 3
Gene Aliases
PRL-3, PRL-R, PRL3
UniGene
Hs.43666
Gene Chromosome Location
Chr.8: 141391993-141432454 on Build GRCh38
Transcripts
NM_007079.3 NM_032611.2 XM_005250764.2 XM_006716500.2 XM_011516803.1 XM_011516805.1 XM_017012998.1 XM_017012999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569170 c.385G>A p.A129T Chr.8: 141427805-141427805 on Build GRCh38
4762192 c.401G>A p.R134H Chr.8: 141427821-141427821 on Build GRCh38
3367227 c.360T>C p.I120I Chr.8: 141427780-141427780 on Build GRCh38
7221327 c.404+10C>T p.? Chr.8: 141427834-141427834 on Build GRCh38
4825699 c.403C>A p.Q135K Chr.8: 141427823-141427823 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371605213 Chr.8: 141427858-141427858 on Build GRCh38
rs948698645 Chr.8: 141427958-141427958 on Build GRCh38
rs1035010555 Chr.8: 141427831-141427831 on Build GRCh38
rs563640852 Chr.8: 141427968-141427968 on Build GRCh38
rs974502057 Chr.8: 141427905-141427905 on Build GRCh38
rs116647247 Chr.8: 141427841-141427841 on Build GRCh38
rs894731929 Chr.8: 141427952-141427952 on Build GRCh38
rs998644727 Chr.8: 141427974-141427974 on Build GRCh38
rs989851176 Chr.8: 141427879-141427879 on Build GRCh38
rs374239991 Chr.8: 141427836-141427836 on Build GRCh38
rs544096433 Chr.8: 141427861-141427861 on Build GRCh38
rs542881966 Chr.8: 141427837-141427837 on Build GRCh38
rs943904954 Chr.8: 141427918-141427918 on Build GRCh38
rs564892631 Chr.8: 141427902-141427902 on Build GRCh38
rs914240512 Chr.8: 141427880-141427880 on Build GRCh38
rs1055933182 Chr.8: 141427937-141427937 on Build GRCh38
rs910470335 Chr.8: 141427900-141427900 on Build GRCh38
rs919923089 Chr.8: 141427920-141427920 on Build GRCh38
rs148274393 Chr.8: 141427955-141427955 on Build GRCh38
rs771529396 Chr.8: 141427804-141427804 on Build GRCh38
rs985923577 Chr.8: 141427898-141427898 on Build GRCh38
rs1021731667 Chr.8: 141427959-141427959 on Build GRCh38
rs7465654 Chr.8: 141427834-141427834 on Build GRCh38
rs977156008 Chr.8: 141427887-141427887 on Build GRCh38
rs1049072544 Chr.8: 141427940-141427940 on Build GRCh38
rs146352123 Chr.8: 141427907-141427908 on Build GRCh38
rs768285801 Chr.8: 141427934-141427934 on Build GRCh38
rs79585294 Chr.8: 141427909-141427909 on Build GRCh38
rs149204222 Chr.8: 141427820-141427820 on Build GRCh38
rs1031425620 Chr.8: 141427877-141427877 on Build GRCh38
rs929389972 Chr.8: 141427939-141427939 on Build GRCh38
rs531603839 Chr.8: 141427801-141427801 on Build GRCh38
rs182463694 Chr.8: 141427979-141427979 on Build GRCh38
rs758475446 Chr.8: 141427859-141427859 on Build GRCh38
rs759942603 Chr.8: 141427914-141427914 on Build GRCh38
rs930088581 Chr.8: 141427921-141427921 on Build GRCh38
rs532411348 Chr.8: 141427917-141427917 on Build GRCh38
rs1030205025 Chr.8: 141427896-141427896 on Build GRCh38
rs938854105 Chr.8: 141427936-141427936 on Build GRCh38
rs907418886 Chr.8: 141427933-141427933 on Build GRCh38
rs576630279 Chr.8: 141427845-141427845 on Build GRCh38
rs1047125514 Chr.8: 141427926-141427926 on Build GRCh38
rs954525632 Chr.8: 141427897-141427897 on Build GRCh38
rs371201500 Chr.8: 141427835-141427835 on Build GRCh38

View Full Product Details