Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6827997-6828242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV1
Entrez Gene ID
7409
Gene Name
vav guanine nucleotide exchange factor 1
Gene Aliases
VAV
UniGene
Hs.116237
Gene Chromosome Location
Chr.19: 6772668-6857366 on Build GRCh38
Transcripts
NM_001258206.1 NM_001258207.1 NM_005428.3 XM_005259642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7076537 c.941G>C p.R314T Chr.19: 6828089-6828089 on Build GRCh38
3541083 c.1017T>C p.L339L Chr.19: 6828165-6828165 on Build GRCh38
3541085 c.1020C>T p.L340L Chr.19: 6828168-6828168 on Build GRCh38
5365429 c.951C>T p.N317N Chr.19: 6828099-6828099 on Build GRCh38
4288671 c.964C>A p.L322M Chr.19: 6828112-6828112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367830862 Chr.19: 6828221-6828221 on Build GRCh38
rs779910201 Chr.19: 6828145-6828145 on Build GRCh38
rs773904821 Chr.19: 6828049-6828049 on Build GRCh38
rs759860276 Chr.19: 6828209-6828209 on Build GRCh38
rs200705988 Chr.19: 6828099-6828099 on Build GRCh38
rs763573602 Chr.19: 6828054-6828054 on Build GRCh38
rs751974047 Chr.19: 6828072-6828072 on Build GRCh38
rs148244465 Chr.19: 6828090-6828090 on Build GRCh38
rs776845862 Chr.19: 6828181-6828181 on Build GRCh38
rs756202184 Chr.19: 6828105-6828105 on Build GRCh38
rs1009235778 Chr.19: 6828115-6828115 on Build GRCh38
rs759883753 Chr.19: 6828085-6828085 on Build GRCh38
rs752959423 Chr.19: 6828215-6828215 on Build GRCh38
rs767905021 Chr.19: 6828213-6828213 on Build GRCh38
rs867909710 Chr.19: 6828141-6828141 on Build GRCh38
rs769397830 Chr.19: 6828042-6828042 on Build GRCh38
rs753876214 Chr.19: 6828120-6828120 on Build GRCh38
rs147266875 Chr.19: 6828134-6828134 on Build GRCh38
rs164022 Chr.19: 6828029-6828029 on Build GRCh38
rs869137960 Chr.19: 6828055-6828055 on Build GRCh38
rs766949121 Chr.19: 6828055-6828055 on Build GRCh38
rs865777450 Chr.19: 6828110-6828110 on Build GRCh38
rs768491466 Chr.19: 6828179-6828179 on Build GRCh38
rs541114512 Chr.19: 6828040-6828040 on Build GRCh38
rs774921585 Chr.19: 6828207-6828207 on Build GRCh38
rs141278487 Chr.19: 6828106-6828106 on Build GRCh38
rs373685773 Chr.19: 6828176-6828176 on Build GRCh38
rs777856537 Chr.19: 6828117-6828117 on Build GRCh38
rs772875776 Chr.19: 6828203-6828203 on Build GRCh38
rs747813611 Chr.19: 6828193-6828193 on Build GRCh38
rs916577588 Chr.19: 6828216-6828216 on Build GRCh38
rs769518456 Chr.19: 6828200-6828200 on Build GRCh38
rs995877473 Chr.19: 6828047-6828047 on Build GRCh38

View Full Product Details