Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 8890643-8890894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFHAS1
Entrez Gene ID
9258
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
LRRC65, MASL1
UniGene
Hs.379414
Gene Chromosome Location
Chr.8: 8783354-8893621 on Build GRCh38
Transcripts
NM_004225.2 XM_011543852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6711840 c.2292G>T p.G764G Chr.8: 8890767-8890767 on Build GRCh38
7255688 c.2308A>G p.M770V Chr.8: 8890751-8890751 on Build GRCh38
1662752 c.2192C>A p.T731N Chr.8: 8890867-8890867 on Build GRCh38
5966666 c.2332G>A p.E778K Chr.8: 8890727-8890727 on Build GRCh38
1214970 c.2311G>A p.A771T Chr.8: 8890748-8890748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896090504 Chr.8: 8890694-8890694 on Build GRCh38
rs745735831 Chr.8: 8890755-8890755 on Build GRCh38
rs953354074 Chr.8: 8890730-8890730 on Build GRCh38
rs750066878 Chr.8: 8890759-8890773 on Build GRCh38
rs780291310 Chr.8: 8890864-8890864 on Build GRCh38
rs753116083 Chr.8: 8890741-8890741 on Build GRCh38
rs747107868 Chr.8: 8890668-8890668 on Build GRCh38
rs747287530 Chr.8: 8890750-8890750 on Build GRCh38
rs777811486 Chr.8: 8890739-8890739 on Build GRCh38
rs968665596 Chr.8: 8890771-8890771 on Build GRCh38
rs751675269 Chr.8: 8890805-8890805 on Build GRCh38
rs762895293 Chr.8: 8890767-8890767 on Build GRCh38
rs775430527 Chr.8: 8890714-8890714 on Build GRCh38
rs768273360 Chr.8: 8890824-8890824 on Build GRCh38
rs749167233 Chr.8: 8890866-8890866 on Build GRCh38
rs61748276 Chr.8: 8890734-8890734 on Build GRCh38
rs775414136 Chr.8: 8890757-8890757 on Build GRCh38
rs145773634 Chr.8: 8890796-8890796 on Build GRCh38
rs773823568 Chr.8: 8890773-8890773 on Build GRCh38
rs746005927 Chr.8: 8890812-8890812 on Build GRCh38
rs750503759 Chr.8: 8890857-8890857 on Build GRCh38
rs769656186 Chr.8: 8890702-8890702 on Build GRCh38
rs771590776 Chr.8: 8890872-8890872 on Build GRCh38
rs570199760 Chr.8: 8890833-8890833 on Build GRCh38
rs764300346 Chr.8: 8890854-8890854 on Build GRCh38
rs766868644 Chr.8: 8890777-8890777 on Build GRCh38
rs149044537 Chr.8: 8890792-8890792 on Build GRCh38
rs756101224 Chr.8: 8890815-8890815 on Build GRCh38
rs573167495 Chr.8: 8890822-8890822 on Build GRCh38
rs759540678 Chr.8: 8890695-8890695 on Build GRCh38
rs946142241 Chr.8: 8890743-8890743 on Build GRCh38
rs937479354 Chr.8: 8890679-8890679 on Build GRCh38
rs772843408 Chr.8: 8890787-8890787 on Build GRCh38
rs777896379 Chr.8: 8890745-8890745 on Build GRCh38
rs765780162 Chr.8: 8890791-8890791 on Build GRCh38
rs370580365 Chr.8: 8890718-8890718 on Build GRCh38
rs751061437 Chr.8: 8890727-8890727 on Build GRCh38
rs200856967 Chr.8: 8890678-8890678 on Build GRCh38
rs756225996 Chr.8: 8890862-8890862 on Build GRCh38
rs767831198 Chr.8: 8890856-8890856 on Build GRCh38
rs747749180 Chr.8: 8890828-8890828 on Build GRCh38
rs762692803 Chr.8: 8890716-8890716 on Build GRCh38
rs765905032 Chr.8: 8890846-8890846 on Build GRCh38
rs779243826 Chr.8: 8890740-8890740 on Build GRCh38
rs780184943 Chr.8: 8890817-8890817 on Build GRCh38
rs746686373 Chr.8: 8890873-8890873 on Build GRCh38
rs776393845 Chr.8: 8890691-8890691 on Build GRCh38
rs1043082671 Chr.8: 8890672-8890672 on Build GRCh38
rs764542837 Chr.8: 8890800-8890800 on Build GRCh38
rs760218204 Chr.8: 8890836-8890836 on Build GRCh38
rs550424418 Chr.8: 8890717-8890717 on Build GRCh38
rs775927622 Chr.8: 8890849-8890849 on Build GRCh38
rs768396718 Chr.8: 8890768-8890768 on Build GRCh38
rs767072421 Chr.8: 8890735-8890735 on Build GRCh38
rs913234775 Chr.8: 8890759-8890759 on Build GRCh38
rs748022931 Chr.8: 8890870-8890870 on Build GRCh38
rs770867229 Chr.8: 8890751-8890751 on Build GRCh38
rs747043944 Chr.8: 8890790-8890790 on Build GRCh38
rs144886583 Chr.8: 8890753-8890753 on Build GRCh38
rs542844021 Chr.8: 8890756-8890756 on Build GRCh38
rs757507595 Chr.8: 8890808-8890808 on Build GRCh38
rs781129814 Chr.8: 8890810-8890810 on Build GRCh38
rs568897405 Chr.8: 8890772-8890772 on Build GRCh38
rs545693944 Chr.8: 8890851-8890851 on Build GRCh38
rs139308568 Chr.8: 8890834-8890834 on Build GRCh38
rs778556919 Chr.8: 8890868-8890868 on Build GRCh38
rs1029215004 Chr.8: 8890723-8890723 on Build GRCh38
rs754238229 Chr.8: 8890737-8890737 on Build GRCh38
rs749257203 Chr.8: 8890818-8890818 on Build GRCh38
rs777893240 Chr.8: 8890665-8890665 on Build GRCh38
rs761537037 Chr.8: 8890774-8890774 on Build GRCh38
rs757410849 Chr.8: 8890855-8890855 on Build GRCh38
rs755006853 Chr.8: 8890867-8890867 on Build GRCh38
rs898794374 Chr.8: 8890722-8890722 on Build GRCh38
rs150332195 Chr.8: 8890744-8890744 on Build GRCh38
rs774197423 Chr.8: 8890826-8890826 on Build GRCh38

View Full Product Details