Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63576868-63577109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTP4A1
Entrez Gene ID
7803
Gene Name
protein tyrosine phosphatase type IVA, member 1
Gene Aliases
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
UniGene
Hs.227777
Gene Chromosome Location
Chr.6: 63528021-63583588 on Build GRCh38
Transcripts
NM_003463.4 XM_011536111.1 XM_011536112.1 XM_017011270.1 XM_017011271.1 XM_017011272.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3875077 c.17G>A p.R6H Chr.6: 63576897-63576897 on Build GRCh38
5982613 c.21A>T p.P7P Chr.6: 63576901-63576901 on Build GRCh38
3875078 c.72T>A p.N24K Chr.6: 63576952-63576952 on Build GRCh38
1176584 c.95A>C p.K32T Chr.6: 63576975-63576975 on Build GRCh38
3160215 c.52A>C p.R18R Chr.6: 63576932-63576932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936867817 Chr.6: 63576999-63576999 on Build GRCh38
rs372864450 Chr.6: 63577033-63577033 on Build GRCh38
rs2075656 Chr.6: 63577080-63577080 on Build GRCh38
rs753010335 Chr.6: 63576965-63576965 on Build GRCh38
rs548254910 Chr.6: 63576964-63576964 on Build GRCh38
rs989664703 Chr.6: 63577000-63577001 on Build GRCh38
rs144613027 Chr.6: 63576946-63576946 on Build GRCh38
rs766268984 Chr.6: 63576955-63576955 on Build GRCh38
rs779426223 Chr.6: 63576972-63576972 on Build GRCh38
rs1029378671 Chr.6: 63576975-63576975 on Build GRCh38
rs1030865174 Chr.6: 63577073-63577073 on Build GRCh38
rs772599109 Chr.6: 63577021-63577021 on Build GRCh38
rs770801394 Chr.6: 63577027-63577027 on Build GRCh38
rs371837650 Chr.6: 63576925-63576925 on Build GRCh38
rs772464554 Chr.6: 63576981-63576981 on Build GRCh38
rs751346049 Chr.6: 63576960-63576960 on Build GRCh38
rs910691106 Chr.6: 63576986-63576986 on Build GRCh38
rs543945371 Chr.6: 63577016-63577016 on Build GRCh38
rs746337411 Chr.6: 63576980-63576980 on Build GRCh38
rs999287588 Chr.6: 63577031-63577031 on Build GRCh38
rs769357724 Chr.6: 63576896-63576896 on Build GRCh38
rs780389234 Chr.6: 63576982-63576982 on Build GRCh38
rs529722832 Chr.6: 63576963-63576963 on Build GRCh38
rs772692917 Chr.6: 63576898-63576898 on Build GRCh38
rs201234588 Chr.6: 63577025-63577025 on Build GRCh38
rs757709853 Chr.6: 63576968-63576968 on Build GRCh38
rs747853302 Chr.6: 63577012-63577012 on Build GRCh38
rs778059234 Chr.6: 63576967-63576967 on Build GRCh38
rs376423935 Chr.6: 63576991-63576991 on Build GRCh38
rs756403505 Chr.6: 63576966-63576966 on Build GRCh38
rs542556466 Chr.6: 63577036-63577036 on Build GRCh38

View Full Product Details