Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6828195-6828462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAV1
Entrez Gene ID
7409
Gene Name
vav guanine nucleotide exchange factor 1
Gene Aliases
VAV
UniGene
Hs.116237
Gene Chromosome Location
Chr.19: 6772668-6857366 on Build GRCh38
Transcripts
NM_001258206.1 NM_001258207.1 NM_005428.3 XM_005259642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3541087 c.1060G>A p.E354K Chr.19: 6828455-6828455 on Build GRCh38
5857048 c.1053G>A p.M351I Chr.19: 6828448-6828448 on Build GRCh38
7397229 c.1067T>G p.L356R Chr.19: 6828462-6828462 on Build GRCh38
4581563 c.1047G>A p.E349E Chr.19: 6828442-6828442 on Build GRCh38
3363075 c.1041G>A p.T347T Chr.19: 6828436-6828436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564391539 Chr.19: 6828296-6828296 on Build GRCh38
rs367830862 Chr.19: 6828221-6828221 on Build GRCh38
rs976505668 Chr.19: 6828287-6828287 on Build GRCh38
rs777506541 Chr.19: 6828430-6828430 on Build GRCh38
rs1037867473 Chr.19: 6828334-6828334 on Build GRCh38
rs949385272 Chr.19: 6828255-6828255 on Build GRCh38
rs753929575 Chr.19: 6828373-6828373 on Build GRCh38
rs867112305 Chr.19: 6828292-6828292 on Build GRCh38
rs754777718 Chr.19: 6828403-6828403 on Build GRCh38
rs752959423 Chr.19: 6828215-6828215 on Build GRCh38
rs563559141 Chr.19: 6828315-6828315 on Build GRCh38
rs376901761 Chr.19: 6828316-6828316 on Build GRCh38
rs543092434 Chr.19: 6828272-6828272 on Build GRCh38
rs921027296 Chr.19: 6828289-6828289 on Build GRCh38
rs528305636 Chr.19: 6828367-6828367 on Build GRCh38
rs781062965 Chr.19: 6828404-6828404 on Build GRCh38
rs751482177 Chr.19: 6828386-6828386 on Build GRCh38
rs940871327 Chr.19: 6828322-6828322 on Build GRCh38
rs943768769 Chr.19: 6828287-6828287 on Build GRCh38
rs529178627 Chr.19: 6828323-6828323 on Build GRCh38
rs111392665 Chr.19: 6828384-6828384 on Build GRCh38
rs755829677 Chr.19: 6828426-6828426 on Build GRCh38
rs780031060 Chr.19: 6828380-6828380 on Build GRCh38
rs898956049 Chr.19: 6828361-6828361 on Build GRCh38
rs61750001 Chr.19: 6828442-6828442 on Build GRCh38
rs757317714 Chr.19: 6828375-6828375 on Build GRCh38
rs781382160 Chr.19: 6828411-6828412 on Build GRCh38
rs908102516 Chr.19: 6828313-6828313 on Build GRCh38
rs764235143 Chr.19: 6828370-6828370 on Build GRCh38
rs371620071 Chr.19: 6828407-6828407 on Build GRCh38
rs916577588 Chr.19: 6828216-6828216 on Build GRCh38

View Full Product Details