Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47636940-47637213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4 XM_017029339.1

SNP Detail

SNP ID Chromosome Location
rs202001852 Chr.X: 47637044-47637044 on Build GRCh38
rs757490836 Chr.X: 47637009-47637009 on Build GRCh38
rs753234734 Chr.X: 47636991-47636991 on Build GRCh38
rs760578996 Chr.X: 47637192-47637192 on Build GRCh38
rs745709371 Chr.X: 47637039-47637039 on Build GRCh38
rs764659361 Chr.X: 47637160-47637160 on Build GRCh38
rs776005990 Chr.X: 47637110-47637110 on Build GRCh38
rs772827642 Chr.X: 47637133-47637133 on Build GRCh38
rs147990079 Chr.X: 47637043-47637043 on Build GRCh38
rs753050036 Chr.X: 47637143-47637143 on Build GRCh38
rs761092029 Chr.X: 47637154-47637154 on Build GRCh38
rs759879065 Chr.X: 47636973-47636973 on Build GRCh38
rs753928196 Chr.X: 47637162-47637162 on Build GRCh38
rs759932355 Chr.X: 47637134-47637134 on Build GRCh38
rs753889188 Chr.X: 47636998-47636998 on Build GRCh38
rs536399296 Chr.X: 47637004-47637004 on Build GRCh38
rs763209954 Chr.X: 47637038-47637038 on Build GRCh38
rs954379683 Chr.X: 47637181-47637181 on Build GRCh38
rs372203607 Chr.X: 47636987-47636987 on Build GRCh38
rs375698452 Chr.X: 47636995-47636995 on Build GRCh38
rs756596523 Chr.X: 47636994-47636994 on Build GRCh38
rs761381660 Chr.X: 47637120-47637120 on Build GRCh38
rs143440705 Chr.X: 47637031-47637031 on Build GRCh38
rs775377290 Chr.X: 47637058-47637058 on Build GRCh38
rs769494379 Chr.X: 47637123-47637123 on Build GRCh38

View Full Product Details